Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33442764-33443259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP3
Entrez Gene ID
360
Gene Name
aquaporin 3 (Gill blood group)
Gene Aliases
AQP-3, GIL
UniGene
Hs.234642
Gene Chromosome Location
Chr.9: 33441154-33447633 on Build GRCh38
Transcripts
NM_001318144.1 NM_004925.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
298549 c.403C>A p.L135I Chr.9: 33442941-33442941 on Build GRCh38
3848352 c.471G>T p.M157I Chr.9: 33442873-33442873 on Build GRCh38
3382518 c.392C>A p.A131D Chr.9: 33442952-33442952 on Build GRCh38
7222640 c.390C>T p.F130F Chr.9: 33442954-33442954 on Build GRCh38
3952663 c.374-7T>C p.? Chr.9: 33442977-33442977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780549945 Chr.9: 33443126-33443126 on Build GRCh38
rs372509995 Chr.9: 33443010-33443010 on Build GRCh38
rs36085726 Chr.9: 33443162-33443162 on Build GRCh38
rs2231231 Chr.9: 33442988-33442988 on Build GRCh38
rs56323059 Chr.9: 33442836-33442836 on Build GRCh38
rs369645288 Chr.9: 33442976-33442976 on Build GRCh38
rs758126339 Chr.9: 33442907-33442907 on Build GRCh38
rs769161680 Chr.9: 33442941-33442941 on Build GRCh38
rs756889869 Chr.9: 33442994-33442994 on Build GRCh38
rs903868255 Chr.9: 33443009-33443009 on Build GRCh38
rs2231230 Chr.9: 33443000-33443000 on Build GRCh38
rs774661867 Chr.9: 33442830-33442830 on Build GRCh38
rs751293190 Chr.9: 33442991-33442991 on Build GRCh38
rs766074740 Chr.9: 33443215-33443215 on Build GRCh38
rs777633399 Chr.9: 33442911-33442911 on Build GRCh38
rs891515641 Chr.9: 33443125-33443125 on Build GRCh38
rs760931129 Chr.9: 33442951-33442951 on Build GRCh38
rs369349773 Chr.9: 33443002-33443002 on Build GRCh38
rs756763951 Chr.9: 33442916-33442916 on Build GRCh38
rs774972543 Chr.9: 33442953-33442953 on Build GRCh38
rs886673852 Chr.9: 33443133-33443133 on Build GRCh38
rs1033818932 Chr.9: 33442822-33442822 on Build GRCh38
rs1021145087 Chr.9: 33443158-33443158 on Build GRCh38
rs775403743 Chr.9: 33442897-33442897 on Build GRCh38
rs777438913 Chr.9: 33442813-33442813 on Build GRCh38
rs376690273 Chr.9: 33442824-33442824 on Build GRCh38
rs777014963 Chr.9: 33442827-33442827 on Build GRCh38
rs529811466 Chr.9: 33442983-33442983 on Build GRCh38
rs765198766 Chr.9: 33442979-33442979 on Build GRCh38
rs895271570 Chr.9: 33443087-33443087 on Build GRCh38
rs775916392 Chr.9: 33442820-33442820 on Build GRCh38
rs2231234 Chr.9: 33442810-33442810 on Build GRCh38
rs144319749 Chr.9: 33442915-33442916 on Build GRCh38
rs56226536 Chr.9: 33442837-33442837 on Build GRCh38
rs2231233 Chr.9: 33442950-33442950 on Build GRCh38
rs376254828 Chr.9: 33443041-33443041 on Build GRCh38
rs143323747 Chr.9: 33442891-33442891 on Build GRCh38
rs760813059 Chr.9: 33442859-33442859 on Build GRCh38
rs56266335 Chr.9: 33443147-33443147 on Build GRCh38
rs2228332 Chr.9: 33442954-33442954 on Build GRCh38
rs746039157 Chr.9: 33442789-33442789 on Build GRCh38
rs753768669 Chr.9: 33442873-33442873 on Build GRCh38
rs539196703 Chr.9: 33443109-33443109 on Build GRCh38
rs754913764 Chr.9: 33442881-33442881 on Build GRCh38
rs749786864 Chr.9: 33442931-33442931 on Build GRCh38
rs905568958 Chr.9: 33443084-33443084 on Build GRCh38
rs766470887 Chr.9: 33442863-33442863 on Build GRCh38
rs372392094 Chr.9: 33442945-33442945 on Build GRCh38
rs547088243 Chr.9: 33442912-33442912 on Build GRCh38
rs560203097 Chr.9: 33443237-33443237 on Build GRCh38
rs770956892 Chr.9: 33442952-33442952 on Build GRCh38
rs762049664 Chr.9: 33442834-33442834 on Build GRCh38
rs778080331 Chr.9: 33443022-33443022 on Build GRCh38
rs964709949 Chr.9: 33443192-33443192 on Build GRCh38
rs574720068 Chr.9: 33443164-33443164 on Build GRCh38
rs750756179 Chr.9: 33443089-33443089 on Build GRCh38
rs367547761 Chr.9: 33442823-33442823 on Build GRCh38
rs765679172 Chr.9: 33443000-33443000 on Build GRCh38
rs746452381 Chr.9: 33442814-33442814 on Build GRCh38
rs1015187387 Chr.9: 33443167-33443167 on Build GRCh38
rs951030345 Chr.9: 33442871-33442871 on Build GRCh38
rs760295780 Chr.9: 33443171-33443171 on Build GRCh38
rs530939977 Chr.9: 33442799-33442799 on Build GRCh38
rs200617012 Chr.9: 33442809-33442809 on Build GRCh38
rs999781174 Chr.9: 33443101-33443101 on Build GRCh38
rs559298059 Chr.9: 33443115-33443115 on Build GRCh38
rs748446670 Chr.9: 33442946-33442946 on Build GRCh38
rs758074939 Chr.9: 33442811-33442811 on Build GRCh38
rs768715017 Chr.9: 33442959-33442959 on Build GRCh38
rs755447177 Chr.9: 33443004-33443004 on Build GRCh38
rs2231228 Chr.9: 33443108-33443108 on Build GRCh38
rs929399491 Chr.9: 33443231-33443231 on Build GRCh38
rs1046588881 Chr.9: 33443123-33443123 on Build GRCh38
rs1055218652 Chr.9: 33443037-33443037 on Build GRCh38
rs138550183 Chr.9: 33443016-33443016 on Build GRCh38
rs190117735 Chr.9: 33443159-33443159 on Build GRCh38
rs868475130 Chr.9: 33443091-33443091 on Build GRCh38
rs748642504 Chr.9: 33443015-33443015 on Build GRCh38
rs759881523 Chr.9: 33442999-33443000 on Build GRCh38
rs770467107 Chr.9: 33442816-33442816 on Build GRCh38
rs780717697 Chr.9: 33442922-33442922 on Build GRCh38
rs540767514 Chr.9: 33443214-33443214 on Build GRCh38
rs2231229 Chr.9: 33443033-33443033 on Build GRCh38
rs536650055 Chr.9: 33442999-33442999 on Build GRCh38
rs945359088 Chr.9: 33442784-33442784 on Build GRCh38
rs762895630 Chr.9: 33442987-33442987 on Build GRCh38
rs752168014 Chr.9: 33443200-33443200 on Build GRCh38

View Full Product Details