Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33443229-33443735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP3
Entrez Gene ID
360
Gene Name
aquaporin 3 (Gill blood group)
Gene Aliases
AQP-3, GIL
UniGene
Hs.234642
Gene Chromosome Location
Chr.9: 33441154-33447633 on Build GRCh38
Transcripts
NM_001318144.1 NM_004925.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5822571 c.256G>A p.V86M Chr.9: 33443438-33443438 on Build GRCh38
7270715 c.318G>A p.L106L Chr.9: 33443376-33443376 on Build GRCh38
455846 c.303G>C p.L101L Chr.9: 33443391-33443391 on Build GRCh38
5495073 c.356T>A p.V119D Chr.9: 33443338-33443338 on Build GRCh38
1108335 c.355G>T p.V119F Chr.9: 33443339-33443339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749257167 Chr.9: 33443474-33443474 on Build GRCh38
rs927229192 Chr.9: 33443652-33443652 on Build GRCh38
rs750516470 Chr.9: 33443624-33443625 on Build GRCh38
rs369321997 Chr.9: 33443283-33443283 on Build GRCh38
rs745993849 Chr.9: 33443698-33443698 on Build GRCh38
rs775667363 Chr.9: 33443464-33443464 on Build GRCh38
rs551112856 Chr.9: 33443461-33443461 on Build GRCh38
rs775614379 Chr.9: 33443309-33443309 on Build GRCh38
rs990801361 Chr.9: 33443616-33443616 on Build GRCh38
rs768660066 Chr.9: 33443479-33443479 on Build GRCh38
rs925183250 Chr.9: 33443611-33443611 on Build GRCh38
rs758695367 Chr.9: 33443300-33443300 on Build GRCh38
rs377229244 Chr.9: 33443501-33443501 on Build GRCh38
rs138913439 Chr.9: 33443519-33443519 on Build GRCh38
rs770031111 Chr.9: 33443462-33443462 on Build GRCh38
rs995119370 Chr.9: 33443533-33443533 on Build GRCh38
rs758863165 Chr.9: 33443437-33443437 on Build GRCh38
rs781376366 Chr.9: 33443510-33443510 on Build GRCh38
rs746128247 Chr.9: 33443688-33443688 on Build GRCh38
rs181185448 Chr.9: 33443563-33443563 on Build GRCh38
rs760291824 Chr.9: 33443497-33443497 on Build GRCh38
rs778212126 Chr.9: 33443455-33443455 on Build GRCh38
rs759726185 Chr.9: 33443288-33443288 on Build GRCh38
rs566526470 Chr.9: 33443575-33443575 on Build GRCh38
rs765905279 Chr.9: 33443499-33443499 on Build GRCh38
rs781532774 Chr.9: 33443456-33443456 on Build GRCh38
rs949923061 Chr.9: 33443278-33443278 on Build GRCh38
rs779519202 Chr.9: 33443408-33443408 on Build GRCh38
rs372865731 Chr.9: 33443293-33443293 on Build GRCh38
rs375757891 Chr.9: 33443439-33443439 on Build GRCh38
rs1027686364 Chr.9: 33443590-33443590 on Build GRCh38
rs757688068 Chr.9: 33443507-33443507 on Build GRCh38
rs758910108 Chr.9: 33443503-33443503 on Build GRCh38
rs751316681 Chr.9: 33443358-33443358 on Build GRCh38
rs755681406 Chr.9: 33443401-33443401 on Build GRCh38
rs1017864270 Chr.9: 33443554-33443554 on Build GRCh38
rs917283687 Chr.9: 33443596-33443596 on Build GRCh38
rs781103309 Chr.9: 33443627-33443627 on Build GRCh38
rs936652223 Chr.9: 33443367-33443367 on Build GRCh38
rs771195954 Chr.9: 33443450-33443450 on Build GRCh38
rs977958342 Chr.9: 33443608-33443608 on Build GRCh38
rs867265809 Chr.9: 33443424-33443424 on Build GRCh38
rs1046977287 Chr.9: 33443490-33443490 on Build GRCh38
rs751960030 Chr.9: 33443549-33443549 on Build GRCh38
rs762406625 Chr.9: 33443689-33443690 on Build GRCh38
rs776577569 Chr.9: 33443284-33443284 on Build GRCh38
rs1054285552 Chr.9: 33443713-33443713 on Build GRCh38
rs769979872 Chr.9: 33443305-33443305 on Build GRCh38
rs756352800 Chr.9: 33443706-33443706 on Build GRCh38
rs761737411 Chr.9: 33443481-33443481 on Build GRCh38
rs1055165073 Chr.9: 33443379-33443379 on Build GRCh38
rs1009440020 Chr.9: 33443541-33443541 on Build GRCh38
rs1027966719 Chr.9: 33443542-33443542 on Build GRCh38
rs776058631 Chr.9: 33443286-33443290 on Build GRCh38
rs957970172 Chr.9: 33443615-33443615 on Build GRCh38
rs371850737 Chr.9: 33443253-33443253 on Build GRCh38
rs375200684 Chr.9: 33443480-33443480 on Build GRCh38
rs778133652 Chr.9: 33443438-33443438 on Build GRCh38
rs186112815 Chr.9: 33443572-33443572 on Build GRCh38
rs772967421 Chr.9: 33443494-33443494 on Build GRCh38
rs111748641 Chr.9: 33443655-33443655 on Build GRCh38
rs767205817 Chr.9: 33443489-33443489 on Build GRCh38
rs567303194 Chr.9: 33443562-33443562 on Build GRCh38
rs767135708 Chr.9: 33443378-33443378 on Build GRCh38
rs201240634 Chr.9: 33443410-33443410 on Build GRCh38
rs559160241 Chr.9: 33443612-33443612 on Build GRCh38
rs750041458 Chr.9: 33443383-33443383 on Build GRCh38
rs761390305 Chr.9: 33443368-33443368 on Build GRCh38
rs916604623 Chr.9: 33443624-33443624 on Build GRCh38
rs2231227 Chr.9: 33443594-33443594 on Build GRCh38
rs780291400 Chr.9: 33443707-33443707 on Build GRCh38
rs370209890 Chr.9: 33443506-33443506 on Build GRCh38
rs905744374 Chr.9: 33443291-33443291 on Build GRCh38
rs764008707 Chr.9: 33443352-33443352 on Build GRCh38
rs925234297 Chr.9: 33443334-33443334 on Build GRCh38
rs747226845 Chr.9: 33443271-33443271 on Build GRCh38
rs762950611 Chr.9: 33443316-33443316 on Build GRCh38
rs1019831032 Chr.9: 33443565-33443565 on Build GRCh38
rs764728106 Chr.9: 33443505-33443505 on Build GRCh38

View Full Product Details