Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114336888-114337407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
UniGene
Hs.494895
Gene Chromosome Location
Chr.9: 114334149-114398498 on Build GRCh38
Transcripts
NM_001317950.1 NM_001317952.1 NM_030767.5 XM_005252244.2 XM_005252245.1 XM_005252247.4 XM_005252248.2 XM_006717294.1 XM_011519063.2 XM_011519065.2 XM_017015172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3903375 c.4284C>T p.R1428R Chr.9: 114337090-114337090 on Build GRCh38
6597915 c.4145G>A p.R1382Q Chr.9: 114337229-114337229 on Build GRCh38
1104377 c.4295G>T p.S1432I Chr.9: 114337079-114337079 on Build GRCh38
5988993 c.4222G>T p.A1408S Chr.9: 114337152-114337152 on Build GRCh38
3653349 c.4136C>A p.P1379Q Chr.9: 114337238-114337238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770972124 Chr.9: 114337209-114337209 on Build GRCh38
rs779099482 Chr.9: 114337025-114337025 on Build GRCh38
rs749224609 Chr.9: 114337157-114337157 on Build GRCh38
rs778267987 Chr.9: 114337014-114337014 on Build GRCh38
rs765791149 Chr.9: 114337169-114337169 on Build GRCh38
rs925297640 Chr.9: 114337297-114337297 on Build GRCh38
rs370544326 Chr.9: 114336916-114336916 on Build GRCh38
rs954709306 Chr.9: 114337230-114337230 on Build GRCh38
rs758418637 Chr.9: 114337136-114337136 on Build GRCh38
rs375487967 Chr.9: 114337331-114337331 on Build GRCh38
rs911196092 Chr.9: 114336991-114336991 on Build GRCh38
rs368911458 Chr.9: 114337328-114337328 on Build GRCh38
rs755221737 Chr.9: 114337184-114337184 on Build GRCh38
rs759313184 Chr.9: 114337163-114337163 on Build GRCh38
rs897233715 Chr.9: 114337041-114337041 on Build GRCh38
rs745888184 Chr.9: 114337042-114337042 on Build GRCh38
rs535703382 Chr.9: 114336911-114336911 on Build GRCh38
rs867340764 Chr.9: 114337280-114337280 on Build GRCh38
rs984421995 Chr.9: 114337010-114337010 on Build GRCh38
rs773745664 Chr.9: 114337370-114337370 on Build GRCh38
rs771760929 Chr.9: 114337226-114337226 on Build GRCh38
rs200451069 Chr.9: 114337330-114337330 on Build GRCh38
rs745385072 Chr.9: 114337298-114337298 on Build GRCh38
rs757676907 Chr.9: 114337021-114337021 on Build GRCh38
rs756326547 Chr.9: 114337273-114337273 on Build GRCh38
rs34983986 Chr.9: 114337385-114337385 on Build GRCh38
rs147478032 Chr.9: 114337140-114337140 on Build GRCh38
rs200387267 Chr.9: 114337183-114337183 on Build GRCh38
rs751639581 Chr.9: 114337254-114337254 on Build GRCh38
rs542469768 Chr.9: 114337238-114337238 on Build GRCh38
rs1014265419 Chr.9: 114336936-114336936 on Build GRCh38
rs747984154 Chr.9: 114337189-114337189 on Build GRCh38
rs781060930 Chr.9: 114337146-114337146 on Build GRCh38
rs752798540 Chr.9: 114337355-114337355 on Build GRCh38
rs868453226 Chr.9: 114337241-114337241 on Build GRCh38
rs202028720 Chr.9: 114337098-114337098 on Build GRCh38
rs770630821 Chr.9: 114337158-114337158 on Build GRCh38
rs768335978 Chr.9: 114337312-114337312 on Build GRCh38
rs762018030 Chr.9: 114337166-114337166 on Build GRCh38
rs986064576 Chr.9: 114336976-114336976 on Build GRCh38
rs149669785 Chr.9: 114337229-114337229 on Build GRCh38
rs375027878 Chr.9: 114337255-114337255 on Build GRCh38
rs549479330 Chr.9: 114337371-114337371 on Build GRCh38
rs568398137 Chr.9: 114337073-114337073 on Build GRCh38
rs774022950 Chr.9: 114337217-114337217 on Build GRCh38
rs769565776 Chr.9: 114337094-114337094 on Build GRCh38
rs745331774 Chr.9: 114337224-114337224 on Build GRCh38
rs777085814 Chr.9: 114337165-114337165 on Build GRCh38
rs766488777 Chr.9: 114337182-114337182 on Build GRCh38
rs781200093 Chr.9: 114337185-114337185 on Build GRCh38
rs771543595 Chr.9: 114337300-114337300 on Build GRCh38
rs145443430 Chr.9: 114337256-114337256 on Build GRCh38
rs745371860 Chr.9: 114337092-114337092 on Build GRCh38
rs759255976 Chr.9: 114337103-114337103 on Build GRCh38
rs144685609 Chr.9: 114337109-114337109 on Build GRCh38
rs145544680 Chr.9: 114337147-114337147 on Build GRCh38
rs780187746 Chr.9: 114337066-114337066 on Build GRCh38
rs944060257 Chr.9: 114336999-114336999 on Build GRCh38
rs377542846 Chr.9: 114337291-114337291 on Build GRCh38
rs375981274 Chr.9: 114337096-114337096 on Build GRCh38
rs774951126 Chr.9: 114337227-114337227 on Build GRCh38
rs779616230 Chr.9: 114337301-114337301 on Build GRCh38
rs78422742 Chr.9: 114337361-114337361 on Build GRCh38
rs886587360 Chr.9: 114337365-114337365 on Build GRCh38
rs772806799 Chr.9: 114337095-114337095 on Build GRCh38
rs551410698 Chr.9: 114337319-114337319 on Build GRCh38
rs375161600 Chr.9: 114337203-114337203 on Build GRCh38
rs866721340 Chr.9: 114337064-114337064 on Build GRCh38
rs776409662 Chr.9: 114337091-114337091 on Build GRCh38
rs754433727 Chr.9: 114337017-114337017 on Build GRCh38
rs148745966 Chr.9: 114337108-114337108 on Build GRCh38
rs368031001 Chr.9: 114337341-114337341 on Build GRCh38
rs758904162 Chr.9: 114337174-114337174 on Build GRCh38
rs752928987 Chr.9: 114337259-114337259 on Build GRCh38
rs777476762 Chr.9: 114337277-114337277 on Build GRCh38
rs896637695 Chr.9: 114336917-114336917 on Build GRCh38
rs950435034 Chr.9: 114336960-114336960 on Build GRCh38
rs779248491 Chr.9: 114337119-114337119 on Build GRCh38
rs772964645 Chr.9: 114337154-114337154 on Build GRCh38
rs1026541704 Chr.9: 114336959-114336959 on Build GRCh38
rs750953510 Chr.9: 114337171-114337171 on Build GRCh38
rs148856699 Chr.9: 114337294-114337294 on Build GRCh38
rs1041486676 Chr.9: 114337019-114337019 on Build GRCh38
rs369952317 Chr.9: 114337194-114337194 on Build GRCh38
rs532832550 Chr.9: 114337368-114337368 on Build GRCh38
rs767534600 Chr.9: 114337349-114337349 on Build GRCh38
rs763470796 Chr.9: 114337242-114337242 on Build GRCh38
rs915107255 Chr.9: 114336923-114336923 on Build GRCh38
rs369108374 Chr.9: 114337137-114337137 on Build GRCh38
rs774892836 Chr.9: 114337338-114337338 on Build GRCh38
rs976770558 Chr.9: 114337220-114337220 on Build GRCh38
rs768500725 Chr.9: 114337086-114337086 on Build GRCh38
rs750525549 Chr.9: 114337121-114337121 on Build GRCh38
rs151129184 Chr.9: 114337159-114337159 on Build GRCh38
rs140122242 Chr.9: 114337141-114337141 on Build GRCh38
rs376756585 Chr.9: 114337149-114337149 on Build GRCh38
rs764595457 Chr.9: 114337107-114337107 on Build GRCh38
rs141074312 Chr.9: 114337195-114337195 on Build GRCh38
rs886727153 Chr.9: 114337168-114337168 on Build GRCh38
rs762462373 Chr.9: 114337201-114337201 on Build GRCh38
rs746683066 Chr.9: 114337302-114337302 on Build GRCh38
rs370226733 Chr.9: 114337248-114337248 on Build GRCh38
rs949491018 Chr.9: 114337360-114337360 on Build GRCh38
rs144199745 Chr.9: 114337099-114337099 on Build GRCh38
rs757292584 Chr.9: 114337292-114337292 on Build GRCh38
rs578078726 Chr.9: 114337221-114337221 on Build GRCh38

View Full Product Details