Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124453287-124453777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRD2
Entrez Gene ID
347088
Gene Name
adhesion G protein-coupled receptor D2
Gene Aliases
GPR144, PGR24
UniGene
Hs.454099
Gene Chromosome Location
Chr.9: 124451087-124477795 on Build GRCh38
Transcripts
XM_006717348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372063 c.862C>T p.H288Y Chr.9: 124453559-124453559 on Build GRCh38
6398058 c.893A>T p.E298V Chr.9: 124453590-124453590 on Build GRCh38
3763626 c.796C>G p.H266D Chr.9: 124453493-124453493 on Build GRCh38
6517439 c.825C>A p.F275L Chr.9: 124453522-124453522 on Build GRCh38
1569433 c.802C>T p.L268F Chr.9: 124453499-124453499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955444358 Chr.9: 124453748-124453748 on Build GRCh38
rs868637546 Chr.9: 124453421-124453421 on Build GRCh38
rs759819122 Chr.9: 124453405-124453405 on Build GRCh38
rs1036459738 Chr.9: 124453680-124453680 on Build GRCh38
rs764112227 Chr.9: 124453471-124453471 on Build GRCh38
rs539411435 Chr.9: 124453523-124453523 on Build GRCh38
rs772400014 Chr.9: 124453579-124453579 on Build GRCh38
rs72616654 Chr.9: 124453493-124453493 on Build GRCh38
rs907864243 Chr.9: 124453751-124453751 on Build GRCh38
rs944709113 Chr.9: 124453730-124453730 on Build GRCh38
rs34667194 Chr.9: 124453623-124453624 on Build GRCh38
rs765088457 Chr.9: 124453702-124453702 on Build GRCh38
rs915589860 Chr.9: 124453455-124453455 on Build GRCh38
rs778617825 Chr.9: 124453494-124453494 on Build GRCh38
rs957388517 Chr.9: 124453696-124453696 on Build GRCh38
rs553341305 Chr.9: 124453541-124453541 on Build GRCh38
rs779669205 Chr.9: 124453582-124453582 on Build GRCh38
rs1019018658 Chr.9: 124453557-124453557 on Build GRCh38
rs542304419 Chr.9: 124453576-124453576 on Build GRCh38
rs998678601 Chr.9: 124453569-124453569 on Build GRCh38
rs575892742 Chr.9: 124453687-124453687 on Build GRCh38
rs760577316 Chr.9: 124453469-124453469 on Build GRCh38
rs971079870 Chr.9: 124453662-124453662 on Build GRCh38
rs1050350579 Chr.9: 124453525-124453525 on Build GRCh38
rs977690137 Chr.9: 124453414-124453414 on Build GRCh38
rs749692650 Chr.9: 124453532-124453532 on Build GRCh38
rs564742321 Chr.9: 124453729-124453729 on Build GRCh38
rs753676254 Chr.9: 124453491-124453491 on Build GRCh38
rs776641169 Chr.9: 124453666-124453666 on Build GRCh38
rs967231955 Chr.9: 124453413-124453413 on Build GRCh38
rs779193559 Chr.9: 124453682-124453683 on Build GRCh38
rs775802538 Chr.9: 124453614-124453614 on Build GRCh38
rs966577447 Chr.9: 124453649-124453649 on Build GRCh38
rs985039082 Chr.9: 124453676-124453676 on Build GRCh38
rs749150557 Chr.9: 124453683-124453683 on Build GRCh38
rs866667641 Chr.9: 124453418-124453418 on Build GRCh38
rs1036609523 Chr.9: 124453330-124453330 on Build GRCh38
rs908267203 Chr.9: 124453668-124453668 on Build GRCh38
rs758045979 Chr.9: 124453505-124453505 on Build GRCh38
rs953434144 Chr.9: 124453376-124453376 on Build GRCh38
rs1005500547 Chr.9: 124453312-124453312 on Build GRCh38
rs925844719 Chr.9: 124453415-124453415 on Build GRCh38
rs78331506 Chr.9: 124453519-124453519 on Build GRCh38
rs67997410 Chr.9: 124453499-124453499 on Build GRCh38
rs892165886 Chr.9: 124453593-124453593 on Build GRCh38
rs921358361 Chr.9: 124453707-124453707 on Build GRCh38
rs764009476 Chr.9: 124453656-124453656 on Build GRCh38
rs1007961694 Chr.9: 124453613-124453613 on Build GRCh38
rs145789254 Chr.9: 124453667-124453667 on Build GRCh38
rs1029635848 Chr.9: 124453657-124453657 on Build GRCh38
rs758099115 Chr.9: 124453714-124453714 on Build GRCh38
rs991126709 Chr.9: 124453435-124453435 on Build GRCh38
rs34747308 Chr.9: 124453742-124453743 on Build GRCh38
rs779582483 Chr.9: 124453511-124453511 on Build GRCh38
rs760837309 Chr.9: 124453639-124453639 on Build GRCh38
rs567913703 Chr.9: 124453306-124453306 on Build GRCh38
rs111489760 Chr.9: 124453643-124453643 on Build GRCh38
rs149830619 Chr.9: 124453550-124453550 on Build GRCh38
rs1019152464 Chr.9: 124453404-124453404 on Build GRCh38
rs1036961791 Chr.9: 124453518-124453518 on Build GRCh38
rs746508379 Chr.9: 124453516-124453516 on Build GRCh38
rs994778204 Chr.9: 124453351-124453351 on Build GRCh38
rs899453441 Chr.9: 124453350-124453350 on Build GRCh38
rs957295190 Chr.9: 124453434-124453434 on Build GRCh38
rs752791632 Chr.9: 124453423-124453423 on Build GRCh38
rs35394820 Chr.9: 124453729-124453730 on Build GRCh38
rs1000507818 Chr.9: 124453566-124453566 on Build GRCh38
rs75599067 Chr.9: 124453706-124453706 on Build GRCh38
rs1008989681 Chr.9: 124453384-124453384 on Build GRCh38
rs535738543 Chr.9: 124453440-124453440 on Build GRCh38
rs1028967935 Chr.9: 124453368-124453368 on Build GRCh38
rs370818119 Chr.9: 124453756-124453756 on Build GRCh38
rs767772696 Chr.9: 124453406-124453406 on Build GRCh38
rs375643116 Chr.9: 124453436-124453436 on Build GRCh38

View Full Product Details