Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124457892-124458385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRD2
Entrez Gene ID
347088
Gene Name
adhesion G protein-coupled receptor D2
Gene Aliases
GPR144, PGR24
UniGene
Hs.454099
Gene Chromosome Location
Chr.9: 124451087-124477795 on Build GRCh38
Transcripts
XM_006717348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4616964 c.1594T>C p.F532L Chr.9: 124458145-124458145 on Build GRCh38
4881343 c.1562G>T p.G521V Chr.9: 124458113-124458113 on Build GRCh38
1624530 c.1601C>A p.S534Y Chr.9: 124458152-124458152 on Build GRCh38
7225070 c.1562G>C p.G521A Chr.9: 124458113-124458113 on Build GRCh38
5410390 c.1682A>C p.Q561P Chr.9: 124458233-124458233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900441454 Chr.9: 124457969-124457969 on Build GRCh38
rs1008690503 Chr.9: 124457949-124457949 on Build GRCh38
rs556936012 Chr.9: 124458304-124458304 on Build GRCh38
rs545410352 Chr.9: 124458303-124458303 on Build GRCh38
rs1004921116 Chr.9: 124458035-124458035 on Build GRCh38
rs764384285 Chr.9: 124458139-124458139 on Build GRCh38
rs1001474624 Chr.9: 124457955-124457955 on Build GRCh38
rs766704522 Chr.9: 124458143-124458143 on Build GRCh38
rs925721868 Chr.9: 124458146-124458146 on Build GRCh38
rs754062201 Chr.9: 124458147-124458147 on Build GRCh38
rs58665806 Chr.9: 124458325-124458325 on Build GRCh38
rs111612783 Chr.9: 124458048-124458048 on Build GRCh38
rs1024940291 Chr.9: 124458030-124458030 on Build GRCh38
rs1012225141 Chr.9: 124458018-124458018 on Build GRCh38
rs115085739 Chr.9: 124458236-124458236 on Build GRCh38
rs376420490 Chr.9: 124458196-124458196 on Build GRCh38
rs4838178 Chr.9: 124458356-124458356 on Build GRCh38
rs754837399 Chr.9: 124458195-124458195 on Build GRCh38
rs1027259544 Chr.9: 124458033-124458033 on Build GRCh38
rs536407602 Chr.9: 124457937-124457937 on Build GRCh38
rs376058342 Chr.9: 124458079-124458079 on Build GRCh38
rs746881494 Chr.9: 124458193-124458193 on Build GRCh38
rs374542366 Chr.9: 124458179-124458179 on Build GRCh38
rs548060345 Chr.9: 124458183-124458183 on Build GRCh38
rs768032724 Chr.9: 124458128-124458128 on Build GRCh38
rs994742403 Chr.9: 124458026-124458026 on Build GRCh38
rs895410138 Chr.9: 124457987-124457987 on Build GRCh38
rs772776281 Chr.9: 124458116-124458116 on Build GRCh38
rs148254178 Chr.9: 124458168-124458168 on Build GRCh38
rs1017453367 Chr.9: 124457958-124457958 on Build GRCh38
rs757493051 Chr.9: 124458160-124458160 on Build GRCh38
rs963231960 Chr.9: 124458044-124458044 on Build GRCh38
rs112513707 Chr.9: 124458074-124458074 on Build GRCh38
rs58155983 Chr.9: 124458113-124458113 on Build GRCh38
rs1023135101 Chr.9: 124458355-124458355 on Build GRCh38
rs994712057 Chr.9: 124458296-124458296 on Build GRCh38
rs769458357 Chr.9: 124458252-124458252 on Build GRCh38
rs760003107 Chr.9: 124458117-124458117 on Build GRCh38
rs994653012 Chr.9: 124458247-124458247 on Build GRCh38
rs1012026189 Chr.9: 124458340-124458340 on Build GRCh38
rs369753254 Chr.9: 124458133-124458133 on Build GRCh38
rs553223903 Chr.9: 124458349-124458349 on Build GRCh38
rs765884645 Chr.9: 124458006-124458006 on Build GRCh38
rs574548728 Chr.9: 124458047-124458047 on Build GRCh38
rs1014997598 Chr.9: 124458042-124458042 on Build GRCh38
rs192479988 Chr.9: 124458091-124458091 on Build GRCh38
rs775933748 Chr.9: 124458130-124458130 on Build GRCh38
rs953429671 Chr.9: 124458078-124458078 on Build GRCh38
rs542313530 Chr.9: 124458359-124458359 on Build GRCh38
rs1038831782 Chr.9: 124458251-124458251 on Build GRCh38
rs758506837 Chr.9: 124458182-124458182 on Build GRCh38
rs960472823 Chr.9: 124458339-124458339 on Build GRCh38
rs184721177 Chr.9: 124458315-124458315 on Build GRCh38
rs776431357 Chr.9: 124458087-124458087 on Build GRCh38
rs368753968 Chr.9: 124458209-124458209 on Build GRCh38
rs944515291 Chr.9: 124458246-124458246 on Build GRCh38
rs369005526 Chr.9: 124458254-124458254 on Build GRCh38
rs953281570 Chr.9: 124458294-124458294 on Build GRCh38
rs936835610 Chr.9: 124458148-124458148 on Build GRCh38
rs529684784 Chr.9: 124457934-124457934 on Build GRCh38
rs375577630 Chr.9: 124458129-124458129 on Build GRCh38
rs879266222 Chr.9: 124458079-124458079 on Build GRCh38
rs1054028249 Chr.9: 124457960-124457960 on Build GRCh38
rs534551897 Chr.9: 124458170-124458170 on Build GRCh38
rs967022695 Chr.9: 124458316-124458316 on Build GRCh38

View Full Product Details