Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14844773-14845287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs749342600 Chr.X: 14845201-14845201 on Build GRCh38
rs144205393 Chr.X: 14845165-14845165 on Build GRCh38
rs775375428 Chr.X: 14845057-14845057 on Build GRCh38
rs375901101 Chr.X: 14845095-14845095 on Build GRCh38
rs894355393 Chr.X: 14844916-14844916 on Build GRCh38
rs986844479 Chr.X: 14844977-14844977 on Build GRCh38
rs756880603 Chr.X: 14845146-14845146 on Build GRCh38
rs1032099311 Chr.X: 14845252-14845252 on Build GRCh38
rs144764663 Chr.X: 14844945-14844945 on Build GRCh38
rs975151235 Chr.X: 14845018-14845018 on Build GRCh38
rs750639868 Chr.X: 14844937-14844937 on Build GRCh38
rs756702783 Chr.X: 14844997-14844997 on Build GRCh38
rs745690135 Chr.X: 14845149-14845149 on Build GRCh38
rs749159651 Chr.X: 14844807-14844807 on Build GRCh38
rs745472535 Chr.X: 14845028-14845028 on Build GRCh38
rs142959373 Chr.X: 14845014-14845014 on Build GRCh38
rs200948146 Chr.X: 14844883-14844883 on Build GRCh38
rs763366953 Chr.X: 14844942-14844942 on Build GRCh38
rs370248837 Chr.X: 14845213-14845213 on Build GRCh38
rs752804063 Chr.X: 14845173-14845173 on Build GRCh38
rs763699754 Chr.X: 14845143-14845143 on Build GRCh38
rs759926657 Chr.X: 14845100-14845100 on Build GRCh38
rs768004295 Chr.X: 14845234-14845234 on Build GRCh38
rs770548473 Chr.X: 14845027-14845027 on Build GRCh38
rs753355273 Chr.X: 14845145-14845145 on Build GRCh38
rs781690574 Chr.X: 14844972-14844972 on Build GRCh38
rs770615860 Chr.X: 14844816-14844816 on Build GRCh38
rs778604488 Chr.X: 14845147-14845147 on Build GRCh38
rs773221142 Chr.X: 14845064-14845064 on Build GRCh38
rs751885600 Chr.X: 14844949-14844949 on Build GRCh38
rs748958265 Chr.X: 14845024-14845024 on Build GRCh38
rs776713934 Chr.X: 14844871-14844871 on Build GRCh38
rs773313492 Chr.X: 14845177-14845177 on Build GRCh38
rs140470884 Chr.X: 14844799-14844799 on Build GRCh38
rs370765798 Chr.X: 14844859-14844859 on Build GRCh38
rs148560784 Chr.X: 14844966-14844966 on Build GRCh38
rs763091316 Chr.X: 14845081-14845081 on Build GRCh38
rs964099207 Chr.X: 14845016-14845016 on Build GRCh38
rs200303151 Chr.X: 14845063-14845063 on Build GRCh38
rs1021143964 Chr.X: 14844946-14844946 on Build GRCh38
rs754392611 Chr.X: 14845038-14845038 on Build GRCh38
rs868761818 Chr.X: 14845159-14845159 on Build GRCh38
rs1032511086 Chr.X: 14844910-14844910 on Build GRCh38
rs746962101 Chr.X: 14845172-14845172 on Build GRCh38
rs771950360 Chr.X: 14845047-14845047 on Build GRCh38
rs774133819 Chr.X: 14844847-14844847 on Build GRCh38
rs758665871 Chr.X: 14845202-14845202 on Build GRCh38
rs149387792 Chr.X: 14844889-14844889 on Build GRCh38
rs146157131 Chr.X: 14845125-14845125 on Build GRCh38
rs1055736273 Chr.X: 14844862-14844862 on Build GRCh38
rs767952529 Chr.X: 14845124-14845124 on Build GRCh38
rs756628396 Chr.X: 14845137-14845137 on Build GRCh38
rs752308935 Chr.X: 14844995-14844995 on Build GRCh38
rs780024809 Chr.X: 14845169-14845169 on Build GRCh38
rs905186149 Chr.X: 14845197-14845197 on Build GRCh38
rs772005838 Chr.X: 14844868-14844868 on Build GRCh38
rs772448306 Chr.X: 14845029-14845029 on Build GRCh38
rs766587734 Chr.X: 14845082-14845082 on Build GRCh38
rs976300441 Chr.X: 14845120-14845120 on Build GRCh38
rs373632536 Chr.X: 14845231-14845231 on Build GRCh38

View Full Product Details