Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18640187-18640690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

Gene Symbol
RS1
Entrez Gene ID
6247
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
UniGene
Hs.715725
Gene Chromosome Location
Chr.X: 18639688-18672103 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

SNP Detail

SNP ID Chromosome Location
rs749841836 Chr.X: 18640331-18640331 on Build GRCh38
rs753767629 Chr.X: 18640435-18640438 on Build GRCh38
rs978277274 Chr.X: 18640218-18640218 on Build GRCh38
rs764170478 Chr.X: 18640269-18640269 on Build GRCh38
rs886554786 Chr.X: 18640473-18640473 on Build GRCh38
rs561740701 Chr.X: 18640390-18640390 on Build GRCh38
rs190004049 Chr.X: 18640300-18640300 on Build GRCh38
rs184280844 Chr.X: 18640285-18640285 on Build GRCh38
rs987493516 Chr.X: 18640494-18640494 on Build GRCh38
rs907054071 Chr.X: 18640660-18640660 on Build GRCh38
rs766105401 Chr.X: 18640241-18640243 on Build GRCh38
rs926747589 Chr.X: 18640666-18640666 on Build GRCh38
rs915534896 Chr.X: 18640644-18640644 on Build GRCh38
rs988539778 Chr.X: 18640639-18640639 on Build GRCh38
rs867295613 Chr.X: 18640447-18640447 on Build GRCh38
rs1049022106 Chr.X: 18640379-18640379 on Build GRCh38
rs192216213 Chr.X: 18640217-18640217 on Build GRCh38
rs1004921095 Chr.X: 18640523-18640523 on Build GRCh38
rs781598838 Chr.X: 18640664-18640664 on Build GRCh38
rs898406894 Chr.X: 18640229-18640230 on Build GRCh38
rs1018903301 Chr.X: 18640573-18640573 on Build GRCh38
rs751705227 Chr.X: 18640636-18640636 on Build GRCh38
rs184482473 Chr.X: 18640662-18640662 on Build GRCh38
rs765044767 Chr.X: 18640434-18640434 on Build GRCh38
rs1039701169 Chr.X: 18640276-18640276 on Build GRCh38
rs976120077 Chr.X: 18640434-18640434 on Build GRCh38
rs946937888 Chr.X: 18640661-18640661 on Build GRCh38
rs756728161 Chr.X: 18640534-18640534 on Build GRCh38
rs966001582 Chr.X: 18640427-18640427 on Build GRCh38
rs192694551 Chr.X: 18640439-18640439 on Build GRCh38
rs912060482 Chr.X: 18640257-18640257 on Build GRCh38
rs774950331 Chr.X: 18640618-18640618 on Build GRCh38
rs956669644 Chr.X: 18640622-18640622 on Build GRCh38

View Full Product Details