Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18644340-18644835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

Gene Symbol
RS1
Entrez Gene ID
6247
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
UniGene
Hs.715725
Gene Chromosome Location
Chr.X: 18639688-18672103 on Build GRCh38
Transcripts
NM_001037343.1 NM_003159.2 NM_000330.3

SNP Detail

SNP ID Chromosome Location
rs754381417 Chr.X: 18644481-18644481 on Build GRCh38
rs780774646 Chr.X: 18644490-18644490 on Build GRCh38
rs1005590954 Chr.X: 18644641-18644641 on Build GRCh38
rs960643185 Chr.X: 18644739-18644739 on Build GRCh38
rs879212727 Chr.X: 18644366-18644366 on Build GRCh38
rs372728604 Chr.X: 18644631-18644631 on Build GRCh38
rs61752157 Chr.X: 18644533-18644533 on Build GRCh38
rs61752144 Chr.X: 18644616-18644616 on Build GRCh38
rs993922120 Chr.X: 18644684-18644684 on Build GRCh38
rs201856847 Chr.X: 18644396-18644396 on Build GRCh38
rs777847903 Chr.X: 18644677-18644677 on Build GRCh38
rs61752147 Chr.X: 18644586-18644586 on Build GRCh38
rs61752154 Chr.X: 18644540-18644540 on Build GRCh38
rs886041235 Chr.X: 18644432-18644432 on Build GRCh38
rs768571577 Chr.X: 18644607-18644607 on Build GRCh38
rs775507405 Chr.X: 18644520-18644520 on Build GRCh38
rs750755649 Chr.X: 18644459-18644459 on Build GRCh38
rs748127123 Chr.X: 18644391-18644391 on Build GRCh38
rs777902881 Chr.X: 18644496-18644496 on Build GRCh38
rs281865347 Chr.X: 18644559-18644560 on Build GRCh38
rs61753160 Chr.X: 18644525-18644526 on Build GRCh38
rs776962681 Chr.X: 18644671-18644671 on Build GRCh38
rs199469698 Chr.X: 18644576-18644576 on Build GRCh38
rs61752153 Chr.X: 18644545-18644545 on Build GRCh38
rs773769605 Chr.X: 18644630-18644630 on Build GRCh38
rs762034990 Chr.X: 18644700-18644700 on Build GRCh38
rs61752155 Chr.X: 18644536-18644536 on Build GRCh38
rs779660363 Chr.X: 18644580-18644580 on Build GRCh38
rs61753162 Chr.X: 18644516-18644516 on Build GRCh38
rs766007270 Chr.X: 18644474-18644474 on Build GRCh38
rs759260793 Chr.X: 18644658-18644658 on Build GRCh38
rs761024021 Chr.X: 18644413-18644413 on Build GRCh38
rs756084649 Chr.X: 18644495-18644495 on Build GRCh38
rs897002454 Chr.X: 18644670-18644670 on Build GRCh38
rs61752148 Chr.X: 18644574-18644577 on Build GRCh38
rs61752158 Chr.X: 18644531-18644531 on Build GRCh38
rs773168630 Chr.X: 18644395-18644395 on Build GRCh38
rs762407219 Chr.X: 18644430-18644430 on Build GRCh38
rs189534862 Chr.X: 18644805-18644805 on Build GRCh38
rs281865348 Chr.X: 18644429-18644429 on Build GRCh38
rs1012348843 Chr.X: 18644765-18644765 on Build GRCh38
rs751121023 Chr.X: 18644651-18644651 on Build GRCh38
rs199469695 Chr.X: 18644578-18644578 on Build GRCh38
rs41309707 Chr.X: 18644364-18644364 on Build GRCh38
rs61753161 Chr.X: 18644524-18644524 on Build GRCh38
rs776803555 Chr.X: 18644612-18644612 on Build GRCh38
rs61752146 Chr.X: 18644602-18644603 on Build GRCh38
rs753408808 Chr.X: 18644374-18644374 on Build GRCh38
rs61753167 Chr.X: 18644453-18644453 on Build GRCh38
rs1055486147 Chr.X: 18644652-18644652 on Build GRCh38
rs767111828 Chr.X: 18644669-18644669 on Build GRCh38
rs897733621 Chr.X: 18644727-18644727 on Build GRCh38
rs201159090 Chr.X: 18644412-18644412 on Build GRCh38
rs61753164 Chr.X: 18644492-18644492 on Build GRCh38
rs1800001 Chr.X: 18644526-18644526 on Build GRCh38
rs770056702 Chr.X: 18644663-18644663 on Build GRCh38
rs61752156 Chr.X: 18644534-18644534 on Build GRCh38
rs61753165 Chr.X: 18644488-18644488 on Build GRCh38
rs1057516744 Chr.X: 18644454-18644454 on Build GRCh38
rs61752075 Chr.X: 18644623-18644623 on Build GRCh38
rs184603580 Chr.X: 18644519-18644519 on Build GRCh38
rs775838179 Chr.X: 18644398-18644398 on Build GRCh38
rs61752145 Chr.X: 18644615-18644615 on Build GRCh38
rs61753168 Chr.X: 18644451-18644451 on Build GRCh38
rs113122924 Chr.X: 18644640-18644640 on Build GRCh38
rs1800002 Chr.X: 18644480-18644480 on Build GRCh38
rs755743775 Chr.X: 18644675-18644675 on Build GRCh38
rs61753163 Chr.X: 18644514-18644514 on Build GRCh38
rs769660191 Chr.X: 18644393-18644393 on Build GRCh38
rs749081937 Chr.X: 18644517-18644517 on Build GRCh38
rs375701039 Chr.X: 18644649-18644649 on Build GRCh38
rs766773788 Chr.X: 18644650-18644650 on Build GRCh38
rs61752151 Chr.X: 18644555-18644555 on Build GRCh38
rs770267626 Chr.X: 18644624-18644624 on Build GRCh38
rs1023296481 Chr.X: 18644792-18644792 on Build GRCh38
rs1035366970 Chr.X: 18644685-18644685 on Build GRCh38
rs61752149 Chr.X: 18644572-18644572 on Build GRCh38
rs758816133 Chr.X: 18644473-18644473 on Build GRCh38
rs199469696 Chr.X: 18644603-18644603 on Build GRCh38
rs765547068 Chr.X: 18644431-18644431 on Build GRCh38
rs61752152 Chr.X: 18644548-18644548 on Build GRCh38
rs1801161 Chr.X: 18644622-18644622 on Build GRCh38
rs61753166 Chr.X: 18644463-18644463 on Build GRCh38
rs61752159 Chr.X: 18644530-18644530 on Build GRCh38
rs200074383 Chr.X: 18644425-18644425 on Build GRCh38
rs199469699 Chr.X: 18644517-18644518 on Build GRCh38

View Full Product Details