Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22090345-22090849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_011545533.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs1011534888 Chr.X: 22090613-22090613 on Build GRCh38
rs973741547 Chr.X: 22090417-22090417 on Build GRCh38
rs905947020 Chr.X: 22090791-22090791 on Build GRCh38
rs776273230 Chr.X: 22090504-22090504 on Build GRCh38
rs912760008 Chr.X: 22090564-22090564 on Build GRCh38
rs918788970 Chr.X: 22090367-22090367 on Build GRCh38
rs750344488 Chr.X: 22090541-22090541 on Build GRCh38
rs376291775 Chr.X: 22090456-22090456 on Build GRCh38
rs763517570 Chr.X: 22090495-22090495 on Build GRCh38
rs761216664 Chr.X: 22090518-22090518 on Build GRCh38
rs760003019 Chr.X: 22090773-22090773 on Build GRCh38
rs776019107 Chr.X: 22090410-22090411 on Build GRCh38
rs1028948612 Chr.X: 22090620-22090620 on Build GRCh38
rs760319054 Chr.X: 22090489-22090489 on Build GRCh38
rs769778878 Chr.X: 22090411-22090411 on Build GRCh38
rs199881071 Chr.X: 22090521-22090521 on Build GRCh38
rs1057515843 Chr.X: 22090465-22090465 on Build GRCh38
rs190038809 Chr.X: 22090823-22090823 on Build GRCh38
rs138952285 Chr.X: 22090565-22090565 on Build GRCh38
rs766607982 Chr.X: 22090709-22090709 on Build GRCh38
rs781597549 Chr.X: 22090379-22090379 on Build GRCh38
rs889032314 Chr.X: 22090634-22090634 on Build GRCh38
rs149411968 Chr.X: 22090807-22090807 on Build GRCh38
rs768117057 Chr.X: 22090780-22090780 on Build GRCh38
rs775148284 Chr.X: 22090475-22090475 on Build GRCh38
rs1051252517 Chr.X: 22090556-22090556 on Build GRCh38
rs763370653 Chr.X: 22090535-22090537 on Build GRCh38
rs373157206 Chr.X: 22090648-22090648 on Build GRCh38
rs144007621 Chr.X: 22090490-22090490 on Build GRCh38
rs948246694 Chr.X: 22090435-22090435 on Build GRCh38
rs770065098 Chr.X: 22090578-22090578 on Build GRCh38
rs771529993 Chr.X: 22090443-22090443 on Build GRCh38
rs752313250 Chr.X: 22090803-22090803 on Build GRCh38
rs773352589 Chr.X: 22090580-22090580 on Build GRCh38
rs934844711 Chr.X: 22090571-22090571 on Build GRCh38
rs186702335 Chr.X: 22090767-22090767 on Build GRCh38
rs749327791 Chr.X: 22090438-22090438 on Build GRCh38
rs931463073 Chr.X: 22090376-22090376 on Build GRCh38
rs886041529 Chr.X: 22090428-22090428 on Build GRCh38
rs932424245 Chr.X: 22090519-22090519 on Build GRCh38
rs780814767 Chr.X: 22090618-22090618 on Build GRCh38
rs935085485 Chr.X: 22090531-22090531 on Build GRCh38
rs145091318 Chr.X: 22090380-22090380 on Build GRCh38
rs763851520 Chr.X: 22090817-22090817 on Build GRCh38
rs149487757 Chr.X: 22090455-22090455 on Build GRCh38
rs758289641 Chr.X: 22090543-22090543 on Build GRCh38
rs893388927 Chr.X: 22090610-22090610 on Build GRCh38
rs777845328 Chr.X: 22090426-22090426 on Build GRCh38
rs952950805 Chr.X: 22090824-22090824 on Build GRCh38

View Full Product Details