Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107650247-107650748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs748667318 Chr.X: 107650483-107650483 on Build GRCh38
rs868718698 Chr.X: 107650558-107650558 on Build GRCh38
rs946064751 Chr.X: 107650352-107650352 on Build GRCh38
rs1041792781 Chr.X: 107650353-107650353 on Build GRCh38
rs5962870 Chr.X: 107650421-107650421 on Build GRCh38
rs770220766 Chr.X: 107650712-107650712 on Build GRCh38
rs6622249 Chr.X: 107650556-107650556 on Build GRCh38
rs1000255630 Chr.X: 107650641-107650641 on Build GRCh38
rs180980520 Chr.X: 107650683-107650683 on Build GRCh38
rs760953456 Chr.X: 107650504-107650504 on Build GRCh38
rs189408679 Chr.X: 107650591-107650591 on Build GRCh38
rs370453137 Chr.X: 107650571-107650571 on Build GRCh38
rs777180568 Chr.X: 107650467-107650467 on Build GRCh38
rs758574318 Chr.X: 107650285-107650287 on Build GRCh38
rs6622250 Chr.X: 107650557-107650557 on Build GRCh38
rs113130873 Chr.X: 107650583-107650583 on Build GRCh38
rs1057515727 Chr.X: 107650570-107650570 on Build GRCh38

View Full Product Details