Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107650637-107651147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs1010764279 Chr.X: 107651059-107651059 on Build GRCh38
rs765995437 Chr.X: 107651082-107651082 on Build GRCh38
rs983877312 Chr.X: 107650996-107650996 on Build GRCh38
rs1025326110 Chr.X: 107650900-107650900 on Build GRCh38
rs1053513497 Chr.X: 107650725-107650725 on Build GRCh38
rs183744100 Chr.X: 107650757-107650757 on Build GRCh38
rs1012116674 Chr.X: 107650758-107650758 on Build GRCh38
rs374457825 Chr.X: 107650728-107650728 on Build GRCh38
rs777186638 Chr.X: 107651120-107651120 on Build GRCh38
rs368782406 Chr.X: 107650876-107650876 on Build GRCh38
rs902446648 Chr.X: 107651089-107651089 on Build GRCh38
rs1025333373 Chr.X: 107650771-107650771 on Build GRCh38
rs768310830 Chr.X: 107650794-107650794 on Build GRCh38
rs770220766 Chr.X: 107650712-107650712 on Build GRCh38
rs748141034 Chr.X: 107651074-107651074 on Build GRCh38
rs1057515728 Chr.X: 107650970-107650971 on Build GRCh38
rs1804226 Chr.X: 107650954-107650954 on Build GRCh38
rs180980520 Chr.X: 107650683-107650683 on Build GRCh38
rs763326742 Chr.X: 107650877-107650877 on Build GRCh38
rs773964270 Chr.X: 107651034-107651034 on Build GRCh38

View Full Product Details