Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185734783-185735290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
UniGene
Hs.58877
Gene Chromosome Location
Chr.1: 185734526-186190953 on Build GRCh38
Transcripts
NM_031935.2 XM_011510038.2 XM_011510041.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6849269 c.80A>G p.Q27R Chr.1: 185734859-185734859 on Build GRCh38
6035025 c.177G>T p.V59V Chr.1: 185734956-185734956 on Build GRCh38
6459810 c.110A>G p.E37G Chr.1: 185734889-185734889 on Build GRCh38
3360543 c.202G>C p.E68Q Chr.1: 185734981-185734981 on Build GRCh38
900944 c.229C>A p.P77T Chr.1: 185735008-185735008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004238153 Chr.1: 185735063-185735063 on Build GRCh38
rs776032407 Chr.1: 185735033-185735033 on Build GRCh38
rs1037195160 Chr.1: 185735233-185735233 on Build GRCh38
rs768644658 Chr.1: 185734841-185734841 on Build GRCh38
rs754826733 Chr.1: 185734914-185734914 on Build GRCh38
rs375537965 Chr.1: 185734829-185734829 on Build GRCh38
rs778944028 Chr.1: 185734933-185734933 on Build GRCh38
rs977263406 Chr.1: 185735131-185735131 on Build GRCh38
rs776798391 Chr.1: 185734987-185734987 on Build GRCh38
rs528525287 Chr.1: 185735144-185735145 on Build GRCh38
rs770572008 Chr.1: 185734986-185734986 on Build GRCh38
rs920452583 Chr.1: 185735182-185735182 on Build GRCh38
rs751696136 Chr.1: 185735077-185735077 on Build GRCh38
rs543568589 Chr.1: 185735012-185735012 on Build GRCh38
rs1048410667 Chr.1: 185735202-185735202 on Build GRCh38
rs757621363 Chr.1: 185734905-185734905 on Build GRCh38
rs114170176 Chr.1: 185734836-185734836 on Build GRCh38
rs749461334 Chr.1: 185735010-185735012 on Build GRCh38
rs888816751 Chr.1: 185734837-185734837 on Build GRCh38
rs538326290 Chr.1: 185735149-185735149 on Build GRCh38
rs201562174 Chr.1: 185734855-185734855 on Build GRCh38
rs762069813 Chr.1: 185735079-185735079 on Build GRCh38
rs879814580 Chr.1: 185735155-185735156 on Build GRCh38
rs772823654 Chr.1: 185734854-185734854 on Build GRCh38
rs771269323 Chr.1: 185734857-185734857 on Build GRCh38
rs570884458 Chr.1: 185735142-185735142 on Build GRCh38
rs753051051 Chr.1: 185735125-185735125 on Build GRCh38
rs770930436 Chr.1: 185735095-185735098 on Build GRCh38
rs763601734 Chr.1: 185735046-185735046 on Build GRCh38
rs753681391 Chr.1: 185734906-185734906 on Build GRCh38
rs138241069 Chr.1: 185734894-185734894 on Build GRCh38
rs771676142 Chr.1: 185734851-185734851 on Build GRCh38
rs115169621 Chr.1: 185734893-185734893 on Build GRCh38
rs199574143 Chr.1: 185734888-185734888 on Build GRCh38
rs960780930 Chr.1: 185735043-185735043 on Build GRCh38
rs547291068 Chr.1: 185734833-185734833 on Build GRCh38
rs752822947 Chr.1: 185734807-185734807 on Build GRCh38
rs568468627 Chr.1: 185735248-185735248 on Build GRCh38
rs747719975 Chr.1: 185734844-185734844 on Build GRCh38
rs148198045 Chr.1: 185735236-185735236 on Build GRCh38
rs376088038 Chr.1: 185735010-185735010 on Build GRCh38
rs1048441564 Chr.1: 185734952-185734952 on Build GRCh38
rs376373045 Chr.1: 185734979-185734979 on Build GRCh38
rs778228244 Chr.1: 185734842-185734842 on Build GRCh38
rs777116837 Chr.1: 185734868-185734868 on Build GRCh38
rs985915465 Chr.1: 185735218-185735218 on Build GRCh38
rs943976154 Chr.1: 185734873-185734873 on Build GRCh38
rs1033945381 Chr.1: 185735066-185735066 on Build GRCh38
rs1040456551 Chr.1: 185735260-185735260 on Build GRCh38
rs758634864 Chr.1: 185734811-185734811 on Build GRCh38
rs556642334 Chr.1: 185735198-185735198 on Build GRCh38
rs751820820 Chr.1: 185734899-185734899 on Build GRCh38
rs76631922 Chr.1: 185734815-185734815 on Build GRCh38
rs777319602 Chr.1: 185734969-185734969 on Build GRCh38
rs745617416 Chr.1: 185734820-185734820 on Build GRCh38
rs754774775 Chr.1: 185735090-185735090 on Build GRCh38
rs992572610 Chr.1: 185735085-185735085 on Build GRCh38
rs551524001 Chr.1: 185735084-185735084 on Build GRCh38
rs765941663 Chr.1: 185735220-185735220 on Build GRCh38
rs753540305 Chr.1: 185735086-185735086 on Build GRCh38
rs930453086 Chr.1: 185735194-185735194 on Build GRCh38
rs759967411 Chr.1: 185735006-185735006 on Build GRCh38
rs139597135 Chr.1: 185735148-185735149 on Build GRCh38
rs143214070 Chr.1: 185735031-185735031 on Build GRCh38
rs906784787 Chr.1: 185735040-185735040 on Build GRCh38
rs971954274 Chr.1: 185735171-185735171 on Build GRCh38
rs1004371882 Chr.1: 185735008-185735008 on Build GRCh38
rs777890155 Chr.1: 185735002-185735002 on Build GRCh38
rs764266638 Chr.1: 185735075-185735075 on Build GRCh38
rs771551161 Chr.1: 185734964-185734964 on Build GRCh38
rs759890607 Chr.1: 185734869-185734869 on Build GRCh38
rs944435978 Chr.1: 185735250-185735250 on Build GRCh38
rs748127350 Chr.1: 185734942-185734942 on Build GRCh38

View Full Product Details