Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185845933-185846455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
UniGene
Hs.58877
Gene Chromosome Location
Chr.1: 185734526-186190953 on Build GRCh38
Transcripts
NM_031935.2 XM_011510038.2 XM_011510041.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6123447 c.285A>T p.T95T Chr.1: 185846042-185846042 on Build GRCh38
5922037 c.299C>T p.P100L Chr.1: 185846056-185846056 on Build GRCh38
5754288 c.295G>A p.D99N Chr.1: 185846052-185846052 on Build GRCh38
6042363 c.270A>C p.E90D Chr.1: 185846027-185846027 on Build GRCh38
5922038 c.298C>T p.P100S Chr.1: 185846055-185846055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985632302 Chr.1: 185846229-185846229 on Build GRCh38
rs755727356 Chr.1: 185846144-185846144 on Build GRCh38
rs980879698 Chr.1: 185846204-185846204 on Build GRCh38
rs760866986 Chr.1: 185846095-185846095 on Build GRCh38
rs758394023 Chr.1: 185845987-185845987 on Build GRCh38
rs774524448 Chr.1: 185846059-185846059 on Build GRCh38
rs750874103 Chr.1: 185845994-185845994 on Build GRCh38
rs956158312 Chr.1: 185846127-185846127 on Build GRCh38
rs770170640 Chr.1: 185846009-185846009 on Build GRCh38
rs777194932 Chr.1: 185845996-185845996 on Build GRCh38
rs570410605 Chr.1: 185846195-185846195 on Build GRCh38
rs187109431 Chr.1: 185846364-185846364 on Build GRCh38
rs764038380 Chr.1: 185846119-185846119 on Build GRCh38
rs746445269 Chr.1: 185846124-185846124 on Build GRCh38
rs34513507 Chr.1: 185846259-185846259 on Build GRCh38
rs955431441 Chr.1: 185846227-185846227 on Build GRCh38
rs545310205 Chr.1: 185846312-185846312 on Build GRCh38
rs940308233 Chr.1: 185846279-185846279 on Build GRCh38
rs745546948 Chr.1: 185846006-185846006 on Build GRCh38
rs941638602 Chr.1: 185846265-185846265 on Build GRCh38
rs756807021 Chr.1: 185846003-185846003 on Build GRCh38
rs189502409 Chr.1: 185846126-185846126 on Build GRCh38
rs776168925 Chr.1: 185846355-185846355 on Build GRCh38
rs554711052 Chr.1: 185846264-185846264 on Build GRCh38
rs1008497796 Chr.1: 185846099-185846099 on Build GRCh38
rs1028100946 Chr.1: 185846055-185846055 on Build GRCh38
rs1026052018 Chr.1: 185846069-185846069 on Build GRCh38
rs10911779 Chr.1: 185846304-185846304 on Build GRCh38
rs898196561 Chr.1: 185846290-185846290 on Build GRCh38
rs184714882 Chr.1: 185845971-185845971 on Build GRCh38
rs1038004174 Chr.1: 185845967-185845967 on Build GRCh38
rs762018712 Chr.1: 185846074-185846074 on Build GRCh38
rs765073432 Chr.1: 185846096-185846096 on Build GRCh38
rs764993236 Chr.1: 185845978-185845978 on Build GRCh38
rs572890563 Chr.1: 185846266-185846266 on Build GRCh38
rs571498680 Chr.1: 185846080-185846080 on Build GRCh38
rs746531214 Chr.1: 185846002-185846002 on Build GRCh38
rs758270694 Chr.1: 185846118-185846118 on Build GRCh38
rs553317695 Chr.1: 185846284-185846284 on Build GRCh38
rs550547960 Chr.1: 185846334-185846334 on Build GRCh38
rs749747591 Chr.1: 185846016-185846016 on Build GRCh38
rs550562248 Chr.1: 185846192-185846192 on Build GRCh38
rs1049445884 Chr.1: 185846313-185846313 on Build GRCh38
rs776053732 Chr.1: 185846014-185846014 on Build GRCh38
rs752621121 Chr.1: 185845981-185845981 on Build GRCh38
rs745519109 Chr.1: 185846140-185846140 on Build GRCh38
rs536309215 Chr.1: 185846263-185846263 on Build GRCh38
rs773464453 Chr.1: 185846089-185846089 on Build GRCh38
rs762996198 Chr.1: 185846411-185846411 on Build GRCh38
rs769299998 Chr.1: 185846050-185846050 on Build GRCh38
rs1008026115 Chr.1: 185846339-185846339 on Build GRCh38
rs890884232 Chr.1: 185846338-185846338 on Build GRCh38
rs988118368 Chr.1: 185846128-185846128 on Build GRCh38
rs752490646 Chr.1: 185846102-185846102 on Build GRCh38
rs915284233 Chr.1: 185846177-185846177 on Build GRCh38
rs79340647 Chr.1: 185846268-185846268 on Build GRCh38
rs760155300 Chr.1: 185846205-185846205 on Build GRCh38
rs568895774 Chr.1: 185846194-185846194 on Build GRCh38
rs780928104 Chr.1: 185846005-185846005 on Build GRCh38
rs780614798 Chr.1: 185846133-185846133 on Build GRCh38
rs780510179 Chr.1: 185846145-185846145 on Build GRCh38
rs905662985 Chr.1: 185846409-185846409 on Build GRCh38

View Full Product Details