Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109643564-109644096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs763872669 Chr.X: 109643781-109643781 on Build GRCh38
rs987258549 Chr.X: 109643620-109643620 on Build GRCh38
rs769828137 Chr.X: 109644001-109644001 on Build GRCh38
rs756342527 Chr.X: 109644011-109644012 on Build GRCh38
rs906608123 Chr.X: 109643835-109643835 on Build GRCh38
rs745454032 Chr.X: 109643676-109643676 on Build GRCh38
rs1033848425 Chr.X: 109643934-109643934 on Build GRCh38
rs770153222 Chr.X: 109643749-109643749 on Build GRCh38
rs1038836909 Chr.X: 109643699-109643699 on Build GRCh38
rs766299247 Chr.X: 109644036-109644036 on Build GRCh38
rs774434074 Chr.X: 109643638-109643639 on Build GRCh38
rs763217379 Chr.X: 109644007-109644007 on Build GRCh38
rs147143931 Chr.X: 109643610-109643610 on Build GRCh38
rs765006032 Chr.X: 109643917-109643917 on Build GRCh38
rs1802109 Chr.X: 109643975-109643975 on Build GRCh38
rs987531901 Chr.X: 109644069-109644069 on Build GRCh38
rs935936652 Chr.X: 109643733-109643733 on Build GRCh38
rs773057024 Chr.X: 109644015-109644015 on Build GRCh38
rs192073376 Chr.X: 109643755-109643755 on Build GRCh38
rs943236853 Chr.X: 109643659-109643659 on Build GRCh38
rs760480845 Chr.X: 109644019-109644019 on Build GRCh38
rs184672678 Chr.X: 109643992-109643992 on Build GRCh38
rs1056037005 Chr.X: 109643977-109643977 on Build GRCh38
rs925904776 Chr.X: 109643862-109643867 on Build GRCh38
rs946083809 Chr.X: 109643756-109643756 on Build GRCh38
rs777069871 Chr.X: 109643994-109643994 on Build GRCh38
rs764281998 Chr.X: 109644008-109644008 on Build GRCh38
rs150760891 Chr.X: 109644002-109644002 on Build GRCh38
rs189248428 Chr.X: 109643744-109643744 on Build GRCh38
rs936026399 Chr.X: 109643907-109643907 on Build GRCh38
rs977510367 Chr.X: 109643854-109643854 on Build GRCh38
rs776499674 Chr.X: 109643820-109643820 on Build GRCh38
rs10126612 Chr.X: 109643871-109643871 on Build GRCh38
rs973692121 Chr.X: 109643981-109643981 on Build GRCh38
rs898008437 Chr.X: 109644070-109644070 on Build GRCh38
rs765895091 Chr.X: 109643999-109643999 on Build GRCh38
rs904489120 Chr.X: 109643714-109643714 on Build GRCh38
rs911625706 Chr.X: 109643633-109643633 on Build GRCh38
rs1025093017 Chr.X: 109643760-109643760 on Build GRCh38

View Full Product Details