Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109665328-109665764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs964324471 Chr.X: 109665544-109665544 on Build GRCh38
rs190547829 Chr.X: 109665642-109665642 on Build GRCh38
rs776149298 Chr.X: 109665387-109665387 on Build GRCh38
rs369861977 Chr.X: 109665540-109665540 on Build GRCh38
rs200793312 Chr.X: 109665483-109665483 on Build GRCh38
rs751721921 Chr.X: 109665538-109665538 on Build GRCh38
rs759883850 Chr.X: 109665398-109665398 on Build GRCh38
rs369016368 Chr.X: 109665543-109665543 on Build GRCh38
rs66800257 Chr.X: 109665717-109665718 on Build GRCh38
rs751702873 Chr.X: 109665365-109665365 on Build GRCh38
rs766121234 Chr.X: 109665382-109665382 on Build GRCh38
rs780987569 Chr.X: 109665554-109665554 on Build GRCh38
rs1032488001 Chr.X: 109665478-109665478 on Build GRCh38
rs970578229 Chr.X: 109665698-109665698 on Build GRCh38
rs397748898 Chr.X: 109665720-109665721 on Build GRCh38
rs753212400 Chr.X: 109665444-109665444 on Build GRCh38
rs745837370 Chr.X: 109665583-109665583 on Build GRCh38
rs200451158 Chr.X: 109665485-109665485 on Build GRCh38
rs3831071 Chr.X: 109665721-109665722 on Build GRCh38
rs112027806 Chr.X: 109665518-109665518 on Build GRCh38
rs980650780 Chr.X: 109665721-109665721 on Build GRCh38
rs765715088 Chr.X: 109665430-109665430 on Build GRCh38
rs753562929 Chr.X: 109665705-109665705 on Build GRCh38
rs113688695 Chr.X: 109665723-109665723 on Build GRCh38
rs760229827 Chr.X: 109665375-109665375 on Build GRCh38
rs869146951 Chr.X: 109665722-109665723 on Build GRCh38
rs1025708750 Chr.X: 109665400-109665400 on Build GRCh38
rs183575068 Chr.X: 109665662-109665662 on Build GRCh38
rs771538928 Chr.X: 109665374-109665374 on Build GRCh38
rs186584278 Chr.X: 109665616-109665616 on Build GRCh38

View Full Product Details