Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130137279-130137784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM1
Entrez Gene ID
9131
Gene Name
apoptosis inducing factor, mitochondria associated 1
Gene Aliases
AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8
UniGene
Hs.424932
Gene Chromosome Location
Chr.X: 130129362-130165887 on Build GRCh38
Transcripts
NM_001130846.3 NM_001130847.3 NM_004208.3 NM_145812.2 NR_132647.1 XM_017029963.1

Gene Symbol
RAB33A
Entrez Gene ID
9363
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
RabS10
UniGene
Hs.654356
Gene Chromosome Location
Chr.X: 130110633-130184872 on Build GRCh38
Transcripts
NM_001130846.3 NM_001130847.3 NM_004208.3 NM_145812.2 NR_132647.1 XM_017029963.1

SNP Detail

SNP ID Chromosome Location
rs1009099745 Chr.X: 130137661-130137661 on Build GRCh38
rs927781083 Chr.X: 130137430-130137430 on Build GRCh38
rs969719481 Chr.X: 130137381-130137381 on Build GRCh38
rs928159906 Chr.X: 130137390-130137390 on Build GRCh38
rs935187661 Chr.X: 130137525-130137525 on Build GRCh38
rs934602986 Chr.X: 130137624-130137624 on Build GRCh38
rs904408118 Chr.X: 130137583-130137583 on Build GRCh38
rs771487502 Chr.X: 130137462-130137462 on Build GRCh38
rs191791972 Chr.X: 130137635-130137635 on Build GRCh38
rs1042745217 Chr.X: 130137558-130137558 on Build GRCh38
rs771849452 Chr.X: 130137328-130137328 on Build GRCh38
rs890290003 Chr.X: 130137672-130137672 on Build GRCh38
rs964695152 Chr.X: 130137313-130137313 on Build GRCh38
rs587780847 Chr.X: 130137543-130137543 on Build GRCh38
rs951010474 Chr.X: 130137345-130137345 on Build GRCh38
rs889332980 Chr.X: 130137638-130137638 on Build GRCh38
rs749543402 Chr.X: 130137534-130137534 on Build GRCh38
rs868579519 Chr.X: 130137410-130137410 on Build GRCh38
rs746018824 Chr.X: 130137725-130137725 on Build GRCh38
rs188923962 Chr.X: 130137346-130137346 on Build GRCh38
rs1017740634 Chr.X: 130137302-130137302 on Build GRCh38
rs747357783 Chr.X: 130137416-130137416 on Build GRCh38
rs1008769715 Chr.X: 130137713-130137713 on Build GRCh38
rs915327675 Chr.X: 130137535-130137535 on Build GRCh38
rs993869709 Chr.X: 130137308-130137308 on Build GRCh38
rs1053323840 Chr.X: 130137636-130137636 on Build GRCh38
rs1050863436 Chr.X: 130137611-130137611 on Build GRCh38
rs745718398 Chr.X: 130137324-130137324 on Build GRCh38
rs916473650 Chr.X: 130137367-130137367 on Build GRCh38
rs992530966 Chr.X: 130137438-130137438 on Build GRCh38
rs878874453 Chr.X: 130137700-130137700 on Build GRCh38
rs1025376221 Chr.X: 130137325-130137325 on Build GRCh38

View Full Product Details