Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 213004908-213005419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGEL2
Entrez Gene ID
90806
Gene Name
angel homolog 2
Gene Aliases
Ccr4d, KIAA0759L
UniGene
Hs.157078
Gene Chromosome Location
Chr.1: 212992182-213015875 on Build GRCh38
Transcripts
NM_001300753.1 NM_001300755.1 NM_001300757.1 NM_001300758.1 NM_144567.4 NR_125333.1 XM_005273344.1 XM_005273345.1 XM_005273346.2 XM_005273347.3 XM_017002774.1 XM_017002775.1 XM_017002776.1 XM_017002777.1 XM_017002778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599601 c.1115A>C p.E372A Chr.1: 213005052-213005052 on Build GRCh38
5820996 c.956A>T p.D319V Chr.1: 213005211-213005211 on Build GRCh38
1243073 c.927G>A p.T309T Chr.1: 213005240-213005240 on Build GRCh38
3803828 c.852C>T p.D284D Chr.1: 213005315-213005315 on Build GRCh38
1338701 c.950G>A p.R317Q Chr.1: 213005217-213005217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372396987 Chr.1: 213005150-213005150 on Build GRCh38
rs371485272 Chr.1: 213005230-213005230 on Build GRCh38
rs775674752 Chr.1: 213005284-213005284 on Build GRCh38
rs981435567 Chr.1: 213005370-213005370 on Build GRCh38
rs753936578 Chr.1: 213005101-213005101 on Build GRCh38
rs754570840 Chr.1: 213005366-213005366 on Build GRCh38
rs867851951 Chr.1: 213005167-213005167 on Build GRCh38
rs765456910 Chr.1: 213005217-213005217 on Build GRCh38
rs1014546231 Chr.1: 213004972-213004972 on Build GRCh38
rs766416721 Chr.1: 213005094-213005094 on Build GRCh38
rs763259803 Chr.1: 213005287-213005287 on Build GRCh38
rs149002480 Chr.1: 213005157-213005157 on Build GRCh38
rs760692360 Chr.1: 213005290-213005295 on Build GRCh38
rs991722841 Chr.1: 213004933-213004933 on Build GRCh38
rs768074662 Chr.1: 213005071-213005071 on Build GRCh38
rs749880684 Chr.1: 213005249-213005249 on Build GRCh38
rs777802669 Chr.1: 213005134-213005134 on Build GRCh38
rs553085890 Chr.1: 213005226-213005226 on Build GRCh38
rs549664993 Chr.1: 213004967-213004967 on Build GRCh38
rs911593065 Chr.1: 213005116-213005116 on Build GRCh38
rs143952778 Chr.1: 213005223-213005223 on Build GRCh38
rs778100901 Chr.1: 213005263-213005263 on Build GRCh38
rs201289618 Chr.1: 213005392-213005392 on Build GRCh38
rs201831288 Chr.1: 213005209-213005209 on Build GRCh38
rs202178970 Chr.1: 213005330-213005330 on Build GRCh38
rs757588975 Chr.1: 213005379-213005379 on Build GRCh38
rs371318328 Chr.1: 213005087-213005087 on Build GRCh38
rs1046235474 Chr.1: 213005018-213005018 on Build GRCh38
rs143448361 Chr.1: 213005241-213005241 on Build GRCh38
rs781653829 Chr.1: 213005385-213005385 on Build GRCh38
rs781550335 Chr.1: 213005128-213005128 on Build GRCh38
rs767973346 Chr.1: 213005064-213005064 on Build GRCh38
rs41277166 Chr.1: 213005041-213005041 on Build GRCh38
rs11120076 Chr.1: 213004998-213004998 on Build GRCh38
rs191902583 Chr.1: 213005030-213005030 on Build GRCh38
rs779665981 Chr.1: 213005252-213005252 on Build GRCh38
rs768365552 Chr.1: 213005262-213005262 on Build GRCh38
rs766633395 Chr.1: 213004968-213004968 on Build GRCh38
rs374763589 Chr.1: 213005240-213005240 on Build GRCh38
rs779426660 Chr.1: 213005349-213005349 on Build GRCh38
rs750306674 Chr.1: 213005344-213005344 on Build GRCh38
rs199975471 Chr.1: 213005266-213005266 on Build GRCh38
rs35494356 Chr.1: 213005132-213005132 on Build GRCh38
rs778546219 Chr.1: 213005372-213005372 on Build GRCh38
rs775524555 Chr.1: 213005106-213005107 on Build GRCh38
rs1033203178 Chr.1: 213005280-213005280 on Build GRCh38
rs898314460 Chr.1: 213005124-213005124 on Build GRCh38
rs752882028 Chr.1: 213005218-213005218 on Build GRCh38
rs771363320 Chr.1: 213005271-213005271 on Build GRCh38
rs779576156 Chr.1: 213005166-213005166 on Build GRCh38
rs1023642942 Chr.1: 213004982-213004982 on Build GRCh38
rs201180365 Chr.1: 213005343-213005343 on Build GRCh38
rs767439809 Chr.1: 213005062-213005075 on Build GRCh38
rs770583817 Chr.1: 213005007-213005007 on Build GRCh38
rs759608394 Chr.1: 213005275-213005275 on Build GRCh38
rs748908379 Chr.1: 213005255-213005255 on Build GRCh38
rs780748334 Chr.1: 213005153-213005153 on Build GRCh38
rs200576994 Chr.1: 213005131-213005131 on Build GRCh38
rs751342083 Chr.1: 213005321-213005321 on Build GRCh38
rs773732795 Chr.1: 213005085-213005085 on Build GRCh38
rs369696781 Chr.1: 213005391-213005391 on Build GRCh38
rs957517748 Chr.1: 213005288-213005288 on Build GRCh38
rs916126504 Chr.1: 213005341-213005341 on Build GRCh38
rs368451930 Chr.1: 213005148-213005148 on Build GRCh38
rs755603082 Chr.1: 213005251-213005251 on Build GRCh38
rs368842471 Chr.1: 213005294-213005294 on Build GRCh38
rs139660446 Chr.1: 213005274-213005274 on Build GRCh38
rs148141428 Chr.1: 213005198-213005198 on Build GRCh38
rs772405308 Chr.1: 213005191-213005191 on Build GRCh38
rs758301052 Chr.1: 213004991-213004991 on Build GRCh38
rs776719513 Chr.1: 213005208-213005208 on Build GRCh38
rs755219220 Chr.1: 213005113-213005113 on Build GRCh38
rs776326168 Chr.1: 213005008-213005008 on Build GRCh38
rs368001839 Chr.1: 213005183-213005183 on Build GRCh38
rs771489874 Chr.1: 213005199-213005199 on Build GRCh38
rs375445649 Chr.1: 213005129-213005129 on Build GRCh38
rs915946683 Chr.1: 213004941-213004941 on Build GRCh38
rs991778465 Chr.1: 213005334-213005334 on Build GRCh38
rs373505616 Chr.1: 213005296-213005296 on Build GRCh38
rs746475061 Chr.1: 213004999-213004999 on Build GRCh38
rs375384442 Chr.1: 213005156-213005156 on Build GRCh38
rs769737729 Chr.1: 213005279-213005279 on Build GRCh38
rs753404842 Chr.1: 213005360-213005360 on Build GRCh38
rs762097562 Chr.1: 213005057-213005057 on Build GRCh38

View Full Product Details