Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215083118-215083609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK2
Entrez Gene ID
3776
Gene Name
potassium two pore domain channel subfamily K member 2
Gene Aliases
K2p2.1, TPKC1, TREK, TREK-1, TREK1, hTREK-1c, hTREK-1e
UniGene
Hs.497745
Gene Chromosome Location
Chr.1: 215002920-215237093 on Build GRCh38
Transcripts
NM_001017424.2 NM_001017425.2 NM_014217.3 XM_011509522.2 XM_011509524.2 XM_017001248.1 XM_017001249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548541 c.44G>A p.G15E Chr.1: 215083429-215083429 on Build GRCh38
1255369 c.42A>T p.A14A Chr.1: 215083427-215083427 on Build GRCh38
1930923 c.44G>C p.G15A Chr.1: 215083429-215083429 on Build GRCh38
1741887 c.17C>T p.S6L Chr.1: 215083402-215083402 on Build GRCh38
6691322 c.34T>C p.Y12H Chr.1: 215083419-215083419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570938656 Chr.1: 215083488-215083488 on Build GRCh38
rs746866510 Chr.1: 215083337-215083337 on Build GRCh38
rs911254851 Chr.1: 215083195-215083195 on Build GRCh38
rs370361294 Chr.1: 215083283-215083283 on Build GRCh38
rs371595283 Chr.1: 215083384-215083384 on Build GRCh38
rs978248553 Chr.1: 215083171-215083171 on Build GRCh38
rs1047597144 Chr.1: 215083222-215083222 on Build GRCh38
rs757502103 Chr.1: 215083476-215083476 on Build GRCh38
rs994653586 Chr.1: 215083206-215083206 on Build GRCh38
rs368385202 Chr.1: 215083330-215083330 on Build GRCh38
rs774194497 Chr.1: 215083444-215083444 on Build GRCh38
rs971304469 Chr.1: 215083154-215083154 on Build GRCh38
rs201512454 Chr.1: 215083303-215083303 on Build GRCh38
rs761577134 Chr.1: 215083446-215083446 on Build GRCh38
rs61818297 Chr.1: 215083545-215083545 on Build GRCh38
rs762979273 Chr.1: 215083443-215083444 on Build GRCh38
rs200437003 Chr.1: 215083292-215083292 on Build GRCh38
rs998529159 Chr.1: 215083571-215083571 on Build GRCh38
rs752718371 Chr.1: 215083459-215083459 on Build GRCh38
rs572329515 Chr.1: 215083530-215083530 on Build GRCh38
rs778126382 Chr.1: 215083416-215083416 on Build GRCh38
rs763863451 Chr.1: 215083397-215083397 on Build GRCh38
rs757093767 Chr.1: 215083423-215083423 on Build GRCh38
rs769784134 Chr.1: 215083441-215083441 on Build GRCh38
rs773317322 Chr.1: 215083438-215083438 on Build GRCh38
rs539971501 Chr.1: 215083503-215083503 on Build GRCh38
rs762453632 Chr.1: 215083389-215083389 on Build GRCh38
rs775055288 Chr.1: 215083264-215083264 on Build GRCh38
rs574383045 Chr.1: 215083437-215083438 on Build GRCh38
rs533944882 Chr.1: 215083225-215083225 on Build GRCh38
rs751326472 Chr.1: 215083420-215083420 on Build GRCh38
rs750813191 Chr.1: 215083270-215083270 on Build GRCh38
rs371383172 Chr.1: 215083467-215083467 on Build GRCh38
rs764840280 Chr.1: 215083257-215083257 on Build GRCh38
rs1054021355 Chr.1: 215083195-215083196 on Build GRCh38
rs1006124241 Chr.1: 215083234-215083234 on Build GRCh38
rs775069221 Chr.1: 215083387-215083387 on Build GRCh38
rs765834306 Chr.1: 215083457-215083457 on Build GRCh38
rs370946670 Chr.1: 215083201-215083202 on Build GRCh38
rs1041153417 Chr.1: 215083201-215083201 on Build GRCh38
rs766967584 Chr.1: 215083402-215083402 on Build GRCh38
rs745765366 Chr.1: 215083440-215083440 on Build GRCh38
rs759906872 Chr.1: 215083449-215083449 on Build GRCh38
rs376231574 Chr.1: 215083512-215083512 on Build GRCh38
rs764202201 Chr.1: 215083471-215083471 on Build GRCh38
rs755673578 Chr.1: 215083556-215083556 on Build GRCh38
rs372387400 Chr.1: 215083342-215083342 on Build GRCh38
rs199698339 Chr.1: 215083415-215083415 on Build GRCh38
rs751731854 Chr.1: 215083472-215083472 on Build GRCh38
rs557555919 Chr.1: 215083202-215083202 on Build GRCh38
rs936977997 Chr.1: 215083185-215083185 on Build GRCh38
rs752839443 Chr.1: 215083413-215083413 on Build GRCh38
rs376060078 Chr.1: 215083310-215083310 on Build GRCh38
rs368043036 Chr.1: 215083367-215083367 on Build GRCh38
rs755999506 Chr.1: 215083477-215083477 on Build GRCh38
rs551109554 Chr.1: 215083532-215083532 on Build GRCh38
rs771379034 Chr.1: 215083448-215083448 on Build GRCh38
rs147376791 Chr.1: 215083378-215083378 on Build GRCh38
rs765168645 Chr.1: 215083407-215083407 on Build GRCh38
rs578217149 Chr.1: 215083577-215083577 on Build GRCh38
rs878998176 Chr.1: 215083204-215083204 on Build GRCh38
rs777210116 Chr.1: 215083357-215083358 on Build GRCh38
rs375480163 Chr.1: 215083363-215083363 on Build GRCh38
rs947728631 Chr.1: 215083322-215083322 on Build GRCh38
rs779416464 Chr.1: 215083442-215083442 on Build GRCh38
rs745929534 Chr.1: 215083496-215083496 on Build GRCh38
rs747364165 Chr.1: 215083303-215083305 on Build GRCh38
rs924095339 Chr.1: 215083173-215083173 on Build GRCh38
rs748893772 Chr.1: 215083443-215083443 on Build GRCh38
rs371586961 Chr.1: 215083301-215083301 on Build GRCh38
rs377170755 Chr.1: 215083268-215083268 on Build GRCh38
rs1024680747 Chr.1: 215083514-215083514 on Build GRCh38
rs773706433 Chr.1: 215083398-215083398 on Build GRCh38
rs758655657 Chr.1: 215083319-215083319 on Build GRCh38
rs1025150623 Chr.1: 215083323-215083323 on Build GRCh38
rs766580620 Chr.1: 215083456-215083457 on Build GRCh38
rs980403919 Chr.1: 215083340-215083340 on Build GRCh38
rs201482650 Chr.1: 215083195-215083195 on Build GRCh38
rs777377008 Chr.1: 215083321-215083321 on Build GRCh38
rs376084267 Chr.1: 215083295-215083295 on Build GRCh38
rs780988028 Chr.1: 215083431-215083431 on Build GRCh38
rs548262217 Chr.1: 215083199-215083199 on Build GRCh38
rs889164193 Chr.1: 215083497-215083497 on Build GRCh38
rs767708308 Chr.1: 215083269-215083269 on Build GRCh38
rs769156553 Chr.1: 215083329-215083330 on Build GRCh38
rs754344732 Chr.1: 215083403-215083403 on Build GRCh38
rs989743403 Chr.1: 215083410-215083410 on Build GRCh38
rs755569227 Chr.1: 215083404-215083404 on Build GRCh38

View Full Product Details