Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226165623-226166122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD3
Entrez Gene ID
64746
Gene Name
acyl-CoA binding domain containing 3
Gene Aliases
GCP60, GOCAP1, GOLPH1, PAP7
UniGene
Hs.520207
Gene Chromosome Location
Chr.1: 226144679-226186722 on Build GRCh38
Transcripts
NM_022735.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
273843 c.373A>C p.N125H Chr.1: 226165914-226165914 on Build GRCh38
4609598 c.351G>T p.K117N Chr.1: 226165936-226165936 on Build GRCh38
4849699 c.406G>C p.D136H Chr.1: 226165881-226165881 on Build GRCh38
5845644 c.376C>T p.P126S Chr.1: 226165911-226165911 on Build GRCh38
1626845 c.371A>G p.Y124C Chr.1: 226165916-226165916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043459509 Chr.1: 226165745-226165746 on Build GRCh38
rs764093578 Chr.1: 226165873-226165873 on Build GRCh38
rs753476995 Chr.1: 226165969-226165969 on Build GRCh38
rs982660147 Chr.1: 226165895-226165895 on Build GRCh38
rs1052389485 Chr.1: 226165775-226165775 on Build GRCh38
rs998988395 Chr.1: 226165768-226165768 on Build GRCh38
rs534406005 Chr.1: 226165689-226165689 on Build GRCh38
rs1043501446 Chr.1: 226166091-226166091 on Build GRCh38
rs376041082 Chr.1: 226165809-226165809 on Build GRCh38
rs964146951 Chr.1: 226165817-226165817 on Build GRCh38
rs983283710 Chr.1: 226165960-226165960 on Build GRCh38
rs949824406 Chr.1: 226166070-226166070 on Build GRCh38
rs200429229 Chr.1: 226166095-226166095 on Build GRCh38
rs555922789 Chr.1: 226165840-226165840 on Build GRCh38
rs893774757 Chr.1: 226165800-226165800 on Build GRCh38
rs762954434 Chr.1: 226165868-226165868 on Build GRCh38
rs780178479 Chr.1: 226166040-226166040 on Build GRCh38
rs781702827 Chr.1: 226166025-226166025 on Build GRCh38
rs909559800 Chr.1: 226165945-226165945 on Build GRCh38
rs906815560 Chr.1: 226165715-226165715 on Build GRCh38
rs768850622 Chr.1: 226166049-226166049 on Build GRCh38
rs1027657404 Chr.1: 226165903-226165903 on Build GRCh38
rs552026804 Chr.1: 226165673-226165673 on Build GRCh38
rs900152951 Chr.1: 226165709-226165709 on Build GRCh38
rs9725713 Chr.1: 226165815-226165815 on Build GRCh38
rs911724137 Chr.1: 226166016-226166017 on Build GRCh38
rs376339787 Chr.1: 226165894-226165894 on Build GRCh38
rs999730899 Chr.1: 226165736-226165736 on Build GRCh38
rs78902019 Chr.1: 226166075-226166075 on Build GRCh38
rs747455259 Chr.1: 226165811-226165811 on Build GRCh38
rs962346460 Chr.1: 226165994-226165994 on Build GRCh38
rs904693241 Chr.1: 226165757-226165757 on Build GRCh38
rs577545559 Chr.1: 226165936-226165936 on Build GRCh38
rs567023025 Chr.1: 226165683-226165683 on Build GRCh38
rs754543231 Chr.1: 226165973-226165973 on Build GRCh38
rs950561110 Chr.1: 226165918-226165918 on Build GRCh38
rs761794296 Chr.1: 226165893-226165893 on Build GRCh38
rs1029494871 Chr.1: 226165826-226165826 on Build GRCh38
rs752243790 Chr.1: 226166003-226166003 on Build GRCh38
rs750222474 Chr.1: 226165900-226165900 on Build GRCh38
rs774408681 Chr.1: 226165875-226165875 on Build GRCh38
rs757889298 Chr.1: 226166013-226166013 on Build GRCh38
rs770137831 Chr.1: 226166038-226166038 on Build GRCh38
rs778635797 Chr.1: 226165997-226165997 on Build GRCh38
rs775731925 Chr.1: 226165866-226165866 on Build GRCh38
rs750323420 Chr.1: 226165852-226165853 on Build GRCh38
rs746249713 Chr.1: 226166037-226166037 on Build GRCh38
rs2306121 Chr.1: 226165820-226165820 on Build GRCh38
rs769950649 Chr.1: 226165851-226165851 on Build GRCh38
rs915676264 Chr.1: 226166061-226166061 on Build GRCh38
rs755895070 Chr.1: 226165925-226165925 on Build GRCh38
rs749577222 Chr.1: 226166044-226166044 on Build GRCh38

View Full Product Details