Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243286192-243286690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
UniGene
Hs.591530
Gene Chromosome Location
Chr.1: 243255328-243500092 on Build GRCh38
Transcripts
NM_006642.3 XM_005273013.4 XM_005273018.1 XM_005273021.4 XM_005273022.3 XM_005273023.4 XM_006711727.3 XM_006711728.3 XM_011544021.2 XM_011544022.2 XM_011544023.2 XM_011544024.2 XM_011544026.2 XM_011544028.2 XM_011544029.2 XM_017000103.1 XM_017000104.1 XM_017000105.1 XM_017000106.1 XM_017000107.1 XM_017000108.1 XM_017000109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5480347 c.494A>G p.K165R Chr.1: 243286345-243286345 on Build GRCh38
1265104 c.499C>G p.Q167E Chr.1: 243286350-243286350 on Build GRCh38
425784 c.431C>G p.S144C Chr.1: 243286282-243286282 on Build GRCh38
679632 c.440A>T p.K147I Chr.1: 243286291-243286291 on Build GRCh38
5380281 c.470A>G p.N157S Chr.1: 243286321-243286321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769881390 Chr.1: 243286322-243286322 on Build GRCh38
rs921608200 Chr.1: 243286616-243286616 on Build GRCh38
rs977621690 Chr.1: 243286553-243286553 on Build GRCh38
rs779296720 Chr.1: 243286408-243286408 on Build GRCh38
rs1021763396 Chr.1: 243286537-243286537 on Build GRCh38
rs753201767 Chr.1: 243286258-243286261 on Build GRCh38
rs192491820 Chr.1: 243286610-243286610 on Build GRCh38
rs746241693 Chr.1: 243286424-243286424 on Build GRCh38
rs755711791 Chr.1: 243286277-243286277 on Build GRCh38
rs547499797 Chr.1: 243286667-243286667 on Build GRCh38
rs564399164 Chr.1: 243286397-243286397 on Build GRCh38
rs1036411079 Chr.1: 243286431-243286431 on Build GRCh38
rs767375284 Chr.1: 243286374-243286374 on Build GRCh38
rs143941525 Chr.1: 243286648-243286648 on Build GRCh38
rs913885292 Chr.1: 243286471-243286471 on Build GRCh38
rs374580113 Chr.1: 243286305-243286305 on Build GRCh38
rs762183907 Chr.1: 243286349-243286349 on Build GRCh38
rs751500755 Chr.1: 243286240-243286240 on Build GRCh38
rs759961252 Chr.1: 243286657-243286657 on Build GRCh38
rs568066096 Chr.1: 243286621-243286621 on Build GRCh38
rs907330422 Chr.1: 243286451-243286451 on Build GRCh38
rs371179612 Chr.1: 243286278-243286278 on Build GRCh38
rs1034794653 Chr.1: 243286509-243286509 on Build GRCh38
rs749010399 Chr.1: 243286281-243286281 on Build GRCh38
rs371348715 Chr.1: 243286331-243286331 on Build GRCh38
rs754160992 Chr.1: 243286228-243286228 on Build GRCh38
rs368200875 Chr.1: 243286372-243286372 on Build GRCh38
rs752723111 Chr.1: 243286391-243286391 on Build GRCh38
rs764307400 Chr.1: 243286341-243286341 on Build GRCh38
rs766344027 Chr.1: 243286237-243286237 on Build GRCh38
rs756907665 Chr.1: 243286398-243286398 on Build GRCh38
rs766215446 Chr.1: 243286361-243286361 on Build GRCh38
rs3818802 Chr.1: 243286579-243286579 on Build GRCh38
rs781229537 Chr.1: 243286598-243286599 on Build GRCh38
rs942494761 Chr.1: 243286612-243286612 on Build GRCh38
rs797045947 Chr.1: 243286332-243286332 on Build GRCh38
rs749550494 Chr.1: 243286526-243286526 on Build GRCh38
rs868051741 Chr.1: 243286302-243286302 on Build GRCh38
rs764435907 Chr.1: 243286224-243286224 on Build GRCh38
rs773249871 Chr.1: 243286384-243286384 on Build GRCh38
rs375641930 Chr.1: 243286394-243286394 on Build GRCh38
rs946837069 Chr.1: 243286500-243286500 on Build GRCh38
rs760647221 Chr.1: 243286335-243286335 on Build GRCh38
rs137862192 Chr.1: 243286231-243286231 on Build GRCh38
rs367590060 Chr.1: 243286323-243286323 on Build GRCh38
rs966210234 Chr.1: 243286597-243286597 on Build GRCh38
rs551943112 Chr.1: 243286472-243286472 on Build GRCh38
rs372403021 Chr.1: 243286393-243286393 on Build GRCh38
rs532369360 Chr.1: 243286463-243286463 on Build GRCh38
rs894700289 Chr.1: 243286511-243286511 on Build GRCh38
rs771275385 Chr.1: 243286283-243286283 on Build GRCh38
rs775828115 Chr.1: 243286444-243286444 on Build GRCh38
rs780682849 Chr.1: 243286256-243286256 on Build GRCh38
rs754881873 Chr.1: 243286249-243286249 on Build GRCh38
rs541533278 Chr.1: 243286369-243286369 on Build GRCh38
rs776989165 Chr.1: 243286348-243286348 on Build GRCh38
rs772538806 Chr.1: 243286425-243286425 on Build GRCh38
rs373877739 Chr.1: 243286368-243286368 on Build GRCh38
rs370587270 Chr.1: 243286549-243286549 on Build GRCh38
rs1025419081 Chr.1: 243286248-243286248 on Build GRCh38
rs747639788 Chr.1: 243286265-243286265 on Build GRCh38

View Full Product Details