Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243503961-243504464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
UniGene
Hs.498292
Gene Chromosome Location
Chr.1: 243488233-243851079 on Build GRCh38
Transcripts
NM_001206729.1 NM_005465.4 NM_181690.2 XM_005272994.4 XM_005272995.2 XM_011544014.2 XM_016999985.1

SNP Detail

SNP ID Chromosome Location
rs927552543 Chr.1: 243504395-243504395 on Build GRCh38
rs1027869355 Chr.1: 243504190-243504190 on Build GRCh38
rs1049467029 Chr.1: 243504221-243504221 on Build GRCh38
rs573066313 Chr.1: 243504303-243504303 on Build GRCh38
rs982118932 Chr.1: 243504123-243504123 on Build GRCh38
rs186169175 Chr.1: 243504316-243504316 on Build GRCh38
rs914377541 Chr.1: 243504196-243504196 on Build GRCh38
rs1018883443 Chr.1: 243504171-243504171 on Build GRCh38
rs765267766 Chr.1: 243504353-243504353 on Build GRCh38
rs937559398 Chr.1: 243504420-243504420 on Build GRCh38
rs543218384 Chr.1: 243504347-243504347 on Build GRCh38
rs543442802 Chr.1: 243504189-243504189 on Build GRCh38
rs3203274 Chr.1: 243504121-243504121 on Build GRCh38
rs900399497 Chr.1: 243504175-243504175 on Build GRCh38
rs61833175 Chr.1: 243504202-243504202 on Build GRCh38
rs879697511 Chr.1: 243504376-243504377 on Build GRCh38
rs1012328485 Chr.1: 243504009-243504009 on Build GRCh38
rs1023763028 Chr.1: 243504011-243504011 on Build GRCh38
rs749778855 Chr.1: 243504027-243504027 on Build GRCh38
rs1008552448 Chr.1: 243504111-243504111 on Build GRCh38
rs796769872 Chr.1: 243504160-243504160 on Build GRCh38
rs983981409 Chr.1: 243504302-243504302 on Build GRCh38
rs981383829 Chr.1: 243504379-243504379 on Build GRCh38
rs140237232 Chr.1: 243504218-243504218 on Build GRCh38
rs759359511 Chr.1: 243504080-243504081 on Build GRCh38

View Full Product Details