Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 1177604-1178099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB2
Entrez Gene ID
105
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Gene Aliases
ADAR3, RED2
UniGene
Hs.586663
Gene Chromosome Location
Chr.10: 1177313-1737476 on Build GRCh38
Transcripts
NM_018702.3

SNP Detail

SNP ID Chromosome Location
rs1050905751 Chr.10: 1177789-1177789 on Build GRCh38
rs4880783 Chr.10: 1177802-1177802 on Build GRCh38
rs889821571 Chr.10: 1177999-1177999 on Build GRCh38
rs965866234 Chr.10: 1178034-1178034 on Build GRCh38
rs1041897683 Chr.10: 1177806-1177806 on Build GRCh38
rs976596829 Chr.10: 1177963-1177963 on Build GRCh38
rs776816770 Chr.10: 1177939-1177939 on Build GRCh38
rs775838983 Chr.10: 1177859-1177859 on Build GRCh38
rs1026828072 Chr.10: 1177662-1177662 on Build GRCh38
rs10903397 Chr.10: 1177861-1177861 on Build GRCh38
rs908875855 Chr.10: 1178065-1178065 on Build GRCh38
rs965252328 Chr.10: 1177710-1177710 on Build GRCh38
rs185287284 Chr.10: 1177821-1177821 on Build GRCh38
rs1011537524 Chr.10: 1177785-1177785 on Build GRCh38
rs975991343 Chr.10: 1177853-1177853 on Build GRCh38
rs544649685 Chr.10: 1177912-1177912 on Build GRCh38
rs984730495 Chr.10: 1178056-1178056 on Build GRCh38
rs772343778 Chr.10: 1177858-1177858 on Build GRCh38
rs537528255 Chr.10: 1177639-1177639 on Build GRCh38
rs34033894 Chr.10: 1178078-1178079 on Build GRCh38
rs560693711 Chr.10: 1178026-1178026 on Build GRCh38
rs984467670 Chr.10: 1177860-1177860 on Build GRCh38
rs938858820 Chr.10: 1177926-1177926 on Build GRCh38
rs879244704 Chr.10: 1177929-1177929 on Build GRCh38
rs923270439 Chr.10: 1177744-1177744 on Build GRCh38
rs948433194 Chr.10: 1177891-1177891 on Build GRCh38
rs1013043771 Chr.10: 1177658-1177658 on Build GRCh38
rs867459519 Chr.10: 1177907-1177907 on Build GRCh38
rs1044558211 Chr.10: 1177734-1177734 on Build GRCh38
rs1017432885 Chr.10: 1177819-1177819 on Build GRCh38
rs1048803546 Chr.10: 1177634-1177634 on Build GRCh38
rs908489766 Chr.10: 1177707-1177707 on Build GRCh38
rs190884476 Chr.10: 1177993-1177993 on Build GRCh38
rs1044577655 Chr.10: 1177902-1177902 on Build GRCh38
rs142218058 Chr.10: 1177771-1177771 on Build GRCh38
rs764483743 Chr.10: 1177921-1177921 on Build GRCh38
rs1003124485 Chr.10: 1177826-1177826 on Build GRCh38
rs1021642364 Chr.10: 1177799-1177799 on Build GRCh38
rs182486608 Chr.10: 1177759-1177759 on Build GRCh38
rs546396561 Chr.10: 1178048-1178048 on Build GRCh38
rs770723966 Chr.10: 1177766-1177766 on Build GRCh38
rs977628249 Chr.10: 1178007-1178007 on Build GRCh38
rs976086296 Chr.10: 1177720-1177720 on Build GRCh38
rs749139938 Chr.10: 1177700-1177700 on Build GRCh38
rs543882229 Chr.10: 1177988-1177988 on Build GRCh38
rs982680651 Chr.10: 1177692-1177692 on Build GRCh38
rs1006965124 Chr.10: 1178020-1178020 on Build GRCh38
rs950601316 Chr.10: 1178049-1178049 on Build GRCh38
rs965302536 Chr.10: 1177959-1177959 on Build GRCh38
rs762255962 Chr.10: 1178018-1178018 on Build GRCh38
rs938923798 Chr.10: 1178022-1178022 on Build GRCh38
rs141625067 Chr.10: 1177980-1177980 on Build GRCh38
rs985531990 Chr.10: 1177810-1177810 on Build GRCh38
rs958521707 Chr.10: 1177636-1177636 on Build GRCh38
rs199681628 Chr.10: 1177850-1177850 on Build GRCh38
rs969745994 Chr.10: 1178074-1178074 on Build GRCh38
rs947488896 Chr.10: 1178003-1178003 on Build GRCh38
rs948838664 Chr.10: 1177674-1177674 on Build GRCh38
rs924735090 Chr.10: 1178008-1178008 on Build GRCh38
rs947567246 Chr.10: 1177805-1177805 on Build GRCh38
rs764615138 Chr.10: 1177829-1177829 on Build GRCh38
rs137862805 Chr.10: 1177977-1177977 on Build GRCh38
rs1053337325 Chr.10: 1177976-1177976 on Build GRCh38
rs891766067 Chr.10: 1177777-1177777 on Build GRCh38
rs1032376578 Chr.10: 1177775-1177775 on Build GRCh38
rs1035347560 Chr.10: 1177630-1177630 on Build GRCh38
rs986132494 Chr.10: 1177981-1177981 on Build GRCh38
rs543986571 Chr.10: 1177772-1177772 on Build GRCh38
rs879327000 Chr.10: 1178066-1178066 on Build GRCh38
rs183891805 Chr.10: 1178046-1178046 on Build GRCh38

View Full Product Details