Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 1179570-1180065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB2
Entrez Gene ID
105
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Gene Aliases
ADAR3, RED2
UniGene
Hs.586663
Gene Chromosome Location
Chr.10: 1177313-1737476 on Build GRCh38
Transcripts
NM_018702.3

SNP Detail

SNP ID Chromosome Location
rs192514006 Chr.10: 1179728-1179728 on Build GRCh38
rs900568919 Chr.10: 1180040-1180040 on Build GRCh38
rs918546577 Chr.10: 1179873-1179874 on Build GRCh38
rs17156051 Chr.10: 1180034-1180034 on Build GRCh38
rs981068625 Chr.10: 1179661-1179661 on Build GRCh38
rs1042654337 Chr.10: 1180010-1180010 on Build GRCh38
rs933639655 Chr.10: 1179801-1179801 on Build GRCh38
rs1983027 Chr.10: 1179817-1179817 on Build GRCh38
rs1025168770 Chr.10: 1179957-1179958 on Build GRCh38
rs555659422 Chr.10: 1179940-1179940 on Build GRCh38
rs374467841 Chr.10: 1179812-1179812 on Build GRCh38
rs1055359729 Chr.10: 1179831-1179831 on Build GRCh38
rs996297220 Chr.10: 1180044-1180044 on Build GRCh38
rs980615014 Chr.10: 1179822-1179822 on Build GRCh38
rs957467918 Chr.10: 1179768-1179768 on Build GRCh38
rs1007153185 Chr.10: 1179748-1179748 on Build GRCh38
rs148702791 Chr.10: 1179887-1179887 on Build GRCh38
rs539145052 Chr.10: 1179879-1179879 on Build GRCh38
rs189199431 Chr.10: 1179725-1179725 on Build GRCh38
rs530096923 Chr.10: 1179646-1179646 on Build GRCh38
rs572250648 Chr.10: 1179977-1179977 on Build GRCh38
rs1038160172 Chr.10: 1179816-1179816 on Build GRCh38
rs1038528335 Chr.10: 1179658-1179658 on Build GRCh38
rs761563663 Chr.10: 1179948-1179948 on Build GRCh38
rs973779019 Chr.10: 1179949-1179949 on Build GRCh38
rs1055689546 Chr.10: 1179985-1179985 on Build GRCh38
rs939748985 Chr.10: 1179815-1179815 on Build GRCh38
rs1002450578 Chr.10: 1179699-1179699 on Build GRCh38
rs1028184806 Chr.10: 1179923-1179923 on Build GRCh38
rs201644925 Chr.10: 1179995-1179996 on Build GRCh38
rs975324698 Chr.10: 1179783-1179783 on Build GRCh38
rs936903067 Chr.10: 1179939-1179939 on Build GRCh38
rs190488894 Chr.10: 1179794-1179794 on Build GRCh38
rs1000826970 Chr.10: 1180028-1180028 on Build GRCh38
rs1032806636 Chr.10: 1179759-1179759 on Build GRCh38
rs1050807590 Chr.10: 1179612-1179612 on Build GRCh38
rs1024822753 Chr.10: 1179813-1179813 on Build GRCh38
rs902491538 Chr.10: 1180022-1180022 on Build GRCh38
rs1013356647 Chr.10: 1179951-1179951 on Build GRCh38
rs918706985 Chr.10: 1179722-1179722 on Build GRCh38
rs188846984 Chr.10: 1179995-1179995 on Build GRCh38
rs966752723 Chr.10: 1179730-1179730 on Build GRCh38
rs1013718952 Chr.10: 1179800-1179800 on Build GRCh38
rs552701851 Chr.10: 1179726-1179726 on Build GRCh38
rs908198813 Chr.10: 1179932-1179932 on Build GRCh38
rs919688675 Chr.10: 1179976-1179976 on Build GRCh38
rs1016050316 Chr.10: 1179998-1179998 on Build GRCh38
rs570494968 Chr.10: 1179878-1179878 on Build GRCh38
rs933651837 Chr.10: 1179600-1179600 on Build GRCh38
rs560583968 Chr.10: 1179706-1179706 on Build GRCh38
rs986754205 Chr.10: 1179926-1179926 on Build GRCh38
rs1042353296 Chr.10: 1179866-1179866 on Build GRCh38
rs1013821408 Chr.10: 1179854-1179854 on Build GRCh38
rs142773112 Chr.10: 1179924-1179924 on Build GRCh38
rs774841822 Chr.10: 1179811-1179811 on Build GRCh38
rs896744951 Chr.10: 1179833-1179833 on Build GRCh38
rs773526266 Chr.10: 1179613-1179613 on Build GRCh38
rs553510570 Chr.10: 1179855-1179855 on Build GRCh38
rs763717260 Chr.10: 1179828-1179828 on Build GRCh38
rs771371390 Chr.10: 1179682-1179682 on Build GRCh38
rs185633219 Chr.10: 1179787-1179787 on Build GRCh38
rs949637052 Chr.10: 1180007-1180007 on Build GRCh38
rs969341394 Chr.10: 1179650-1179650 on Build GRCh38
rs186088520 Chr.10: 1179917-1179917 on Build GRCh38
rs181695444 Chr.10: 1179906-1179906 on Build GRCh38
rs540592421 Chr.10: 1179993-1179993 on Build GRCh38
rs540273820 Chr.10: 1179700-1179700 on Build GRCh38
rs568426262 Chr.10: 1179805-1179805 on Build GRCh38
rs763332279 Chr.10: 1179628-1179628 on Build GRCh38
rs921195816 Chr.10: 1179785-1179785 on Build GRCh38
rs753409293 Chr.10: 1179905-1179905 on Build GRCh38
rs902563365 Chr.10: 1179876-1179876 on Build GRCh38
rs891253387 Chr.10: 1179746-1179746 on Build GRCh38
rs956429537 Chr.10: 1179598-1179598 on Build GRCh38
rs147638824 Chr.10: 1179751-1179751 on Build GRCh38
rs938181788 Chr.10: 1179829-1179829 on Build GRCh38

View Full Product Details