Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 1179833-1180326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB2
Entrez Gene ID
105
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Gene Aliases
ADAR3, RED2
UniGene
Hs.586663
Gene Chromosome Location
Chr.10: 1177313-1737476 on Build GRCh38
Transcripts
NM_018702.3

SNP Detail

SNP ID Chromosome Location
rs900568919 Chr.10: 1180040-1180040 on Build GRCh38
rs918546577 Chr.10: 1179873-1179874 on Build GRCh38
rs1027064643 Chr.10: 1180289-1180289 on Build GRCh38
rs918072943 Chr.10: 1180153-1180153 on Build GRCh38
rs17156051 Chr.10: 1180034-1180034 on Build GRCh38
rs1042654337 Chr.10: 1180010-1180010 on Build GRCh38
rs548105681 Chr.10: 1180247-1180247 on Build GRCh38
rs900537642 Chr.10: 1180251-1180251 on Build GRCh38
rs1025168770 Chr.10: 1179957-1179958 on Build GRCh38
rs555659422 Chr.10: 1179940-1179940 on Build GRCh38
rs890924637 Chr.10: 1180256-1180256 on Build GRCh38
rs897061844 Chr.10: 1180283-1180283 on Build GRCh38
rs996297220 Chr.10: 1180044-1180044 on Build GRCh38
rs148702791 Chr.10: 1179887-1179887 on Build GRCh38
rs780138234 Chr.10: 1180094-1180094 on Build GRCh38
rs539145052 Chr.10: 1179879-1179879 on Build GRCh38
rs572250648 Chr.10: 1179977-1179977 on Build GRCh38
rs1052664983 Chr.10: 1180158-1180158 on Build GRCh38
rs761563663 Chr.10: 1179948-1179948 on Build GRCh38
rs973779019 Chr.10: 1179949-1179949 on Build GRCh38
rs1055689546 Chr.10: 1179985-1179985 on Build GRCh38
rs951433660 Chr.10: 1180070-1180070 on Build GRCh38
rs879342402 Chr.10: 1180284-1180284 on Build GRCh38
rs942790590 Chr.10: 1180131-1180131 on Build GRCh38
rs1028184806 Chr.10: 1179923-1179923 on Build GRCh38
rs532100641 Chr.10: 1180164-1180164 on Build GRCh38
rs201644925 Chr.10: 1179995-1179996 on Build GRCh38
rs761533243 Chr.10: 1180112-1180112 on Build GRCh38
rs936903067 Chr.10: 1179939-1179939 on Build GRCh38
rs562684482 Chr.10: 1180083-1180083 on Build GRCh38
rs1000826970 Chr.10: 1180028-1180028 on Build GRCh38
rs902491538 Chr.10: 1180022-1180022 on Build GRCh38
rs1013356647 Chr.10: 1179951-1179951 on Build GRCh38
rs188846984 Chr.10: 1179995-1179995 on Build GRCh38
rs539463908 Chr.10: 1180300-1180300 on Build GRCh38
rs995461491 Chr.10: 1180288-1180288 on Build GRCh38
rs1054176621 Chr.10: 1180248-1180248 on Build GRCh38
rs973747874 Chr.10: 1180118-1180118 on Build GRCh38
rs908198813 Chr.10: 1179932-1179932 on Build GRCh38
rs527995105 Chr.10: 1180187-1180187 on Build GRCh38
rs750191081 Chr.10: 1180060-1180060 on Build GRCh38
rs919688675 Chr.10: 1179976-1179976 on Build GRCh38
rs570494968 Chr.10: 1179878-1179878 on Build GRCh38
rs1016050316 Chr.10: 1179998-1179998 on Build GRCh38
rs570332612 Chr.10: 1180255-1180255 on Build GRCh38
rs986754205 Chr.10: 1179926-1179926 on Build GRCh38
rs1042353296 Chr.10: 1179866-1179866 on Build GRCh38
rs1013821408 Chr.10: 1179854-1179854 on Build GRCh38
rs559915579 Chr.10: 1180135-1180135 on Build GRCh38
rs7081909 Chr.10: 1180066-1180066 on Build GRCh38
rs142773112 Chr.10: 1179924-1179924 on Build GRCh38
rs7097304 Chr.10: 1180133-1180133 on Build GRCh38
rs1015522406 Chr.10: 1180102-1180102 on Build GRCh38
rs924206942 Chr.10: 1180204-1180204 on Build GRCh38
rs936921956 Chr.10: 1180145-1180145 on Build GRCh38
rs553510570 Chr.10: 1179855-1179855 on Build GRCh38
rs949637052 Chr.10: 1180007-1180007 on Build GRCh38
rs1015491481 Chr.10: 1180273-1180273 on Build GRCh38
rs186088520 Chr.10: 1179917-1179917 on Build GRCh38
rs181695444 Chr.10: 1179906-1179906 on Build GRCh38
rs531751120 Chr.10: 1180111-1180111 on Build GRCh38
rs540592421 Chr.10: 1179993-1179993 on Build GRCh38
rs912666818 Chr.10: 1180125-1180125 on Build GRCh38
rs932050517 Chr.10: 1180253-1180253 on Build GRCh38
rs990625948 Chr.10: 1180141-1180141 on Build GRCh38
rs753409293 Chr.10: 1179905-1179905 on Build GRCh38
rs925409040 Chr.10: 1180144-1180144 on Build GRCh38
rs902563365 Chr.10: 1179876-1179876 on Build GRCh38
rs949565533 Chr.10: 1180176-1180176 on Build GRCh38
rs1008082680 Chr.10: 1180257-1180257 on Build GRCh38

View Full Product Details