Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70674661-70675189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
-
UniGene
Hs.352156
Gene Chromosome Location
Chr.10: 70672518-70762441 on Build GRCh38
Transcripts
NM_080722.4 NM_139155.3 XM_011539300.2 XM_011539303.2 XM_011539305.2 XM_011539306.2 XM_011539307.2 XM_011539308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354272 c.235C>T p.H79Y Chr.10: 70674708-70674708 on Build GRCh38
2160552 c.517G>A p.G173R Chr.10: 70674990-70674990 on Build GRCh38
4144822 c.413A>G p.E138G Chr.10: 70674886-70674886 on Build GRCh38
5663599 c.277G>T p.G93* Chr.10: 70674750-70674750 on Build GRCh38
1348852 c.384A>G p.V128V Chr.10: 70674857-70674857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147702966 Chr.10: 70674919-70674919 on Build GRCh38
rs147532816 Chr.10: 70675033-70675033 on Build GRCh38
rs1013819811 Chr.10: 70675141-70675141 on Build GRCh38
rs773646424 Chr.10: 70674957-70674957 on Build GRCh38
rs757990147 Chr.10: 70674736-70674736 on Build GRCh38
rs776807900 Chr.10: 70674948-70674948 on Build GRCh38
rs748048717 Chr.10: 70674759-70674759 on Build GRCh38
rs756040828 Chr.10: 70674842-70674842 on Build GRCh38
rs1025076597 Chr.10: 70675112-70675112 on Build GRCh38
rs773558030 Chr.10: 70674890-70674890 on Build GRCh38
rs751131063 Chr.10: 70674836-70674836 on Build GRCh38
rs200138396 Chr.10: 70674889-70674889 on Build GRCh38
rs769866318 Chr.10: 70674762-70674762 on Build GRCh38
rs144062605 Chr.10: 70674886-70674887 on Build GRCh38
rs1026831167 Chr.10: 70674774-70674774 on Build GRCh38
rs565065762 Chr.10: 70675075-70675075 on Build GRCh38
rs760436249 Chr.10: 70675032-70675032 on Build GRCh38
rs149675451 Chr.10: 70674824-70674824 on Build GRCh38
rs145448662 Chr.10: 70674831-70674831 on Build GRCh38
rs1032561028 Chr.10: 70675081-70675081 on Build GRCh38
rs776686112 Chr.10: 70674693-70674693 on Build GRCh38
rs554496606 Chr.10: 70674688-70674688 on Build GRCh38
rs757243783 Chr.10: 70674926-70674926 on Build GRCh38
rs140711390 Chr.10: 70674918-70674918 on Build GRCh38
rs755746039 Chr.10: 70675043-70675043 on Build GRCh38
rs747153190 Chr.10: 70674872-70674872 on Build GRCh38
rs376781600 Chr.10: 70675031-70675031 on Build GRCh38
rs760999643 Chr.10: 70674778-70674778 on Build GRCh38
rs1049554519 Chr.10: 70674689-70674689 on Build GRCh38
rs772253184 Chr.10: 70674773-70674773 on Build GRCh38
rs140980259 Chr.10: 70674956-70674956 on Build GRCh38
rs144956824 Chr.10: 70674989-70674989 on Build GRCh38
rs775672699 Chr.10: 70674855-70674855 on Build GRCh38
rs779844010 Chr.10: 70674683-70674683 on Build GRCh38
rs147256643 Chr.10: 70674837-70674837 on Build GRCh38
rs777841514 Chr.10: 70674844-70674844 on Build GRCh38
rs556871391 Chr.10: 70674817-70674817 on Build GRCh38
rs61754837 Chr.10: 70674733-70674733 on Build GRCh38
rs768795403 Chr.10: 70674945-70674945 on Build GRCh38
rs762148250 Chr.10: 70674888-70674888 on Build GRCh38
rs746336683 Chr.10: 70675005-70675006 on Build GRCh38
rs142923634 Chr.10: 70675138-70675138 on Build GRCh38
rs780648554 Chr.10: 70675110-70675110 on Build GRCh38
rs773117486 Chr.10: 70674812-70674813 on Build GRCh38
rs778909297 Chr.10: 70675035-70675035 on Build GRCh38
rs754610594 Chr.10: 70674897-70674897 on Build GRCh38
rs761881397 Chr.10: 70674694-70674694 on Build GRCh38
rs139509859 Chr.10: 70674812-70674812 on Build GRCh38
rs368175978 Chr.10: 70674757-70674757 on Build GRCh38
rs774787276 Chr.10: 70674975-70674975 on Build GRCh38
rs777004859 Chr.10: 70674785-70674785 on Build GRCh38
rs147617019 Chr.10: 70674846-70674846 on Build GRCh38
rs768301923 Chr.10: 70674750-70674750 on Build GRCh38
rs757796491 Chr.10: 70674732-70674732 on Build GRCh38
rs371895859 Chr.10: 70674780-70674780 on Build GRCh38
rs538508973 Chr.10: 70674705-70674705 on Build GRCh38
rs148577695 Chr.10: 70675129-70675129 on Build GRCh38
rs753031516 Chr.10: 70674720-70674720 on Build GRCh38
rs760936209 Chr.10: 70674722-70674722 on Build GRCh38
rs936513162 Chr.10: 70674686-70674686 on Build GRCh38
rs202169822 Chr.10: 70674886-70674886 on Build GRCh38
rs767186205 Chr.10: 70675022-70675022 on Build GRCh38
rs771093112 Chr.10: 70674704-70674704 on Build GRCh38
rs145276866 Chr.10: 70674838-70674838 on Build GRCh38
rs1025192331 Chr.10: 70675152-70675152 on Build GRCh38
rs190475211 Chr.10: 70675163-70675163 on Build GRCh38
rs1040971063 Chr.10: 70674930-70674930 on Build GRCh38
rs932455101 Chr.10: 70674847-70674847 on Build GRCh38
rs753803274 Chr.10: 70674850-70674850 on Build GRCh38
rs774505808 Chr.10: 70674765-70674765 on Build GRCh38
rs757735745 Chr.10: 70674889-70674894 on Build GRCh38
rs372200693 Chr.10: 70674770-70674770 on Build GRCh38
rs779942556 Chr.10: 70674927-70674927 on Build GRCh38
rs142118917 Chr.10: 70674881-70674881 on Build GRCh38
rs71477504 Chr.10: 70674940-70674940 on Build GRCh38
rs749142773 Chr.10: 70674925-70674925 on Build GRCh38
rs370245201 Chr.10: 70674746-70674746 on Build GRCh38
rs781758593 Chr.10: 70674943-70674944 on Build GRCh38
rs767274241 Chr.10: 70674906-70674906 on Build GRCh38
rs772015923 Chr.10: 70674852-70674852 on Build GRCh38
rs746771070 Chr.10: 70674684-70674684 on Build GRCh38
rs1047684991 Chr.10: 70674990-70674990 on Build GRCh38
rs201449180 Chr.10: 70674706-70674706 on Build GRCh38
rs983420240 Chr.10: 70674789-70674789 on Build GRCh38
rs764571352 Chr.10: 70674723-70674723 on Build GRCh38
rs373362179 Chr.10: 70674972-70674972 on Build GRCh38
rs186907048 Chr.10: 70674813-70674813 on Build GRCh38
rs752458774 Chr.10: 70674737-70674737 on Build GRCh38
rs184307876 Chr.10: 70675128-70675128 on Build GRCh38
rs145982854 Chr.10: 70674751-70674751 on Build GRCh38
rs866835304 Chr.10: 70674884-70674885 on Build GRCh38
rs150199494 Chr.10: 70674969-70674969 on Build GRCh38
rs765756863 Chr.10: 70674895-70674895 on Build GRCh38
rs895299900 Chr.10: 70675140-70675140 on Build GRCh38
rs748405704 Chr.10: 70674954-70674954 on Build GRCh38
rs749952998 Chr.10: 70674734-70674734 on Build GRCh38
rs548297532 Chr.10: 70675165-70675165 on Build GRCh38
rs775554366 Chr.10: 70674775-70674775 on Build GRCh38
rs74141809 Chr.10: 70674708-70674708 on Build GRCh38
rs752456214 Chr.10: 70674907-70674907 on Build GRCh38
rs754314763 Chr.10: 70674724-70674724 on Build GRCh38
rs763545321 Chr.10: 70674894-70674894 on Build GRCh38
rs778768541 Chr.10: 70674807-70674808 on Build GRCh38
rs747120060 Chr.10: 70674943-70674943 on Build GRCh38
rs959531027 Chr.10: 70675094-70675094 on Build GRCh38

View Full Product Details