Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72334268-72334808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB12
Entrez Gene ID
54788
Gene Name
DnaJ heat shock protein family (Hsp40) member B12
Gene Aliases
DJ10
UniGene
Hs.500013
Gene Chromosome Location
Chr.10: 72332830-72355149 on Build GRCh38
Transcripts
NM_001002762.2 NM_017626.4 XM_005269931.1 XM_005269932.3

SNP Detail

SNP ID Chromosome Location
rs759589609 Chr.10: 72334344-72334344 on Build GRCh38
rs1007274814 Chr.10: 72334380-72334380 on Build GRCh38
rs959770577 Chr.10: 72334602-72334602 on Build GRCh38
rs983986841 Chr.10: 72334646-72334646 on Build GRCh38
rs760673581 Chr.10: 72334663-72334663 on Build GRCh38
rs368219491 Chr.10: 72334657-72334657 on Build GRCh38
rs1032214900 Chr.10: 72334697-72334697 on Build GRCh38
rs1004098781 Chr.10: 72334336-72334336 on Build GRCh38
rs905442003 Chr.10: 72334754-72334754 on Build GRCh38
rs975248721 Chr.10: 72334368-72334368 on Build GRCh38
rs929776928 Chr.10: 72334627-72334627 on Build GRCh38
rs992544435 Chr.10: 72334607-72334607 on Build GRCh38
rs1048625792 Chr.10: 72334569-72334569 on Build GRCh38
rs1028086135 Chr.10: 72334682-72334682 on Build GRCh38
rs1035280373 Chr.10: 72334764-72334764 on Build GRCh38
rs532860635 Chr.10: 72334474-72334474 on Build GRCh38
rs901246114 Chr.10: 72334389-72334389 on Build GRCh38
rs755592333 Chr.10: 72334769-72334769 on Build GRCh38
rs998262828 Chr.10: 72334402-72334402 on Build GRCh38
rs948853519 Chr.10: 72334533-72334533 on Build GRCh38
rs899349161 Chr.10: 72334537-72334540 on Build GRCh38
rs968532404 Chr.10: 72334686-72334686 on Build GRCh38
rs539554486 Chr.10: 72334321-72334321 on Build GRCh38
rs1039710011 Chr.10: 72334381-72334381 on Build GRCh38
rs933999107 Chr.10: 72334704-72334704 on Build GRCh38
rs748761786 Chr.10: 72334578-72334578 on Build GRCh38
rs72808118 Chr.10: 72334414-72334414 on Build GRCh38
rs900863912 Chr.10: 72334782-72334782 on Build GRCh38
rs929834305 Chr.10: 72334334-72334334 on Build GRCh38
rs191074448 Chr.10: 72334335-72334335 on Build GRCh38
rs1022489509 Chr.10: 72334732-72334732 on Build GRCh38
rs888391239 Chr.10: 72334373-72334373 on Build GRCh38
rs543382585 Chr.10: 72334310-72334310 on Build GRCh38
rs965179086 Chr.10: 72334362-72334362 on Build GRCh38
rs918457361 Chr.10: 72334330-72334330 on Build GRCh38
rs188359793 Chr.10: 72334504-72334504 on Build GRCh38
rs559026522 Chr.10: 72334415-72334415 on Build GRCh38
rs573541156 Chr.10: 72334383-72334383 on Build GRCh38
rs979751724 Chr.10: 72334540-72334540 on Build GRCh38
rs1048751448 Chr.10: 72334351-72334351 on Build GRCh38
rs766547155 Chr.10: 72334668-72334668 on Build GRCh38
rs969679484 Chr.10: 72334508-72334508 on Build GRCh38
rs1011463472 Chr.10: 72334728-72334728 on Build GRCh38
rs41282262 Chr.10: 72334299-72334299 on Build GRCh38
rs892568298 Chr.10: 72334727-72334727 on Build GRCh38
rs968752013 Chr.10: 72334404-72334404 on Build GRCh38
rs376921588 Chr.10: 72334550-72334550 on Build GRCh38
rs530523217 Chr.10: 72334511-72334511 on Build GRCh38
rs918347470 Chr.10: 72334615-72334615 on Build GRCh38
rs145166873 Chr.10: 72334770-72334770 on Build GRCh38
rs995154224 Chr.10: 72334322-72334322 on Build GRCh38
rs182796644 Chr.10: 72334497-72334497 on Build GRCh38
rs770511144 Chr.10: 72334462-72334462 on Build GRCh38
rs778586065 Chr.10: 72334711-72334711 on Build GRCh38
rs750457437 Chr.10: 72334647-72334647 on Build GRCh38
rs1034055279 Chr.10: 72334559-72334559 on Build GRCh38
rs1011077037 Chr.10: 72334501-72334501 on Build GRCh38
rs754286951 Chr.10: 72334712-72334712 on Build GRCh38
rs113225492 Chr.10: 72334536-72334536 on Build GRCh38
rs1056987193 Chr.10: 72334314-72334314 on Build GRCh38
rs904972762 Chr.10: 72334319-72334319 on Build GRCh38
rs939687430 Chr.10: 72334776-72334776 on Build GRCh38
rs1002490980 Chr.10: 72334755-72334755 on Build GRCh38
rs777272112 Chr.10: 72334542-72334542 on Build GRCh38
rs1000248148 Chr.10: 72334687-72334687 on Build GRCh38
rs901215030 Chr.10: 72334664-72334664 on Build GRCh38
rs1028349594 Chr.10: 72334377-72334377 on Build GRCh38
rs770569178 Chr.10: 72334626-72334626 on Build GRCh38
rs745406603 Chr.10: 72334777-72334777 on Build GRCh38
rs761980407 Chr.10: 72334430-72334430 on Build GRCh38
rs1026626794 Chr.10: 72334326-72334326 on Build GRCh38
rs141378115 Chr.10: 72334719-72334719 on Build GRCh38
rs1019952622 Chr.10: 72334356-72334356 on Build GRCh38
rs938436314 Chr.10: 72334301-72334301 on Build GRCh38
rs942655040 Chr.10: 72334658-72334658 on Build GRCh38

View Full Product Details