Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72340897-72341403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB12
Entrez Gene ID
54788
Gene Name
DnaJ heat shock protein family (Hsp40) member B12
Gene Aliases
DJ10
UniGene
Hs.500013
Gene Chromosome Location
Chr.10: 72332830-72355149 on Build GRCh38
Transcripts
NM_001002762.2 NM_017626.4 XM_005269931.1 XM_005269932.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390820 c.628G>A p.D210N Chr.10: 72341102-72341102 on Build GRCh38
7416277 c.623T>A p.F208Y Chr.10: 72341005-72341005 on Build GRCh38
7416278 c.725T>A p.F242Y Chr.10: 72341005-72341005 on Build GRCh38
4390821 c.526G>A p.D176N Chr.10: 72341102-72341102 on Build GRCh38
7365048 c.625G>A p.D209N Chr.10: 72341105-72341105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370734886 Chr.10: 72341103-72341103 on Build GRCh38
rs946853153 Chr.10: 72341284-72341284 on Build GRCh38
rs550917850 Chr.10: 72341237-72341237 on Build GRCh38
rs764910483 Chr.10: 72341203-72341203 on Build GRCh38
rs777664658 Chr.10: 72341082-72341082 on Build GRCh38
rs753361361 Chr.10: 72341107-72341107 on Build GRCh38
rs182924547 Chr.10: 72341040-72341040 on Build GRCh38
rs146585391 Chr.10: 72341105-72341105 on Build GRCh38
rs766025466 Chr.10: 72341210-72341210 on Build GRCh38
rs763447858 Chr.10: 72341065-72341065 on Build GRCh38
rs1053232927 Chr.10: 72341236-72341236 on Build GRCh38
rs922694021 Chr.10: 72340987-72340987 on Build GRCh38
rs938294828 Chr.10: 72341336-72341336 on Build GRCh38
rs955410888 Chr.10: 72341217-72341217 on Build GRCh38
rs144190908 Chr.10: 72341167-72341167 on Build GRCh38
rs763369473 Chr.10: 72340947-72340947 on Build GRCh38
rs150571106 Chr.10: 72341370-72341370 on Build GRCh38
rs72808129 Chr.10: 72341342-72341342 on Build GRCh38
rs747223755 Chr.10: 72340965-72340965 on Build GRCh38
rs761416324 Chr.10: 72341185-72341185 on Build GRCh38
rs576377084 Chr.10: 72340945-72340945 on Build GRCh38
rs191338296 Chr.10: 72341242-72341242 on Build GRCh38
rs926420300 Chr.10: 72340968-72340968 on Build GRCh38
rs759934125 Chr.10: 72341054-72341054 on Build GRCh38
rs779020651 Chr.10: 72341222-72341222 on Build GRCh38
rs753403780 Chr.10: 72341068-72341068 on Build GRCh38
rs147993058 Chr.10: 72340998-72340998 on Build GRCh38
rs751390287 Chr.10: 72341135-72341135 on Build GRCh38
rs147227290 Chr.10: 72341039-72341039 on Build GRCh38
rs777469200 Chr.10: 72341000-72341000 on Build GRCh38
rs536978112 Chr.10: 72341285-72341285 on Build GRCh38
rs555241425 Chr.10: 72341302-72341302 on Build GRCh38
rs756039593 Chr.10: 72341221-72341221 on Build GRCh38
rs202089929 Chr.10: 72341001-72341001 on Build GRCh38
rs779652414 Chr.10: 72341131-72341131 on Build GRCh38
rs373452284 Chr.10: 72340945-72340945 on Build GRCh38
rs41282266 Chr.10: 72341073-72341073 on Build GRCh38
rs751932092 Chr.10: 72340954-72340954 on Build GRCh38
rs988300628 Chr.10: 72341031-72341031 on Build GRCh38
rs375528007 Chr.10: 72341004-72341004 on Build GRCh38
rs773259182 Chr.10: 72341214-72341215 on Build GRCh38
rs749399633 Chr.10: 72341083-72341083 on Build GRCh38
rs750215643 Chr.10: 72341071-72341071 on Build GRCh38
rs143406248 Chr.10: 72341032-72341032 on Build GRCh38
rs200575874 Chr.10: 72341030-72341030 on Build GRCh38
rs375106554 Chr.10: 72341125-72341125 on Build GRCh38
rs914000027 Chr.10: 72341259-72341259 on Build GRCh38
rs749348077 Chr.10: 72341140-72341140 on Build GRCh38
rs764785110 Chr.10: 72341106-72341106 on Build GRCh38
rs762565615 Chr.10: 72341209-72341209 on Build GRCh38
rs542360918 Chr.10: 72341234-72341234 on Build GRCh38
rs755428025 Chr.10: 72340962-72340962 on Build GRCh38
rs775670938 Chr.10: 72341174-72341174 on Build GRCh38
rs767474761 Chr.10: 72341286-72341286 on Build GRCh38
rs771056666 Chr.10: 72341142-72341142 on Build GRCh38
rs369942027 Chr.10: 72341016-72341016 on Build GRCh38
rs561511838 Chr.10: 72341151-72341151 on Build GRCh38
rs752220148 Chr.10: 72340997-72340997 on Build GRCh38
rs764965547 Chr.10: 72341109-72341109 on Build GRCh38
rs139558574 Chr.10: 72341137-72341137 on Build GRCh38
rs138088607 Chr.10: 72341215-72341215 on Build GRCh38
rs906093337 Chr.10: 72341317-72341317 on Build GRCh38
rs754830639 Chr.10: 72341136-72341136 on Build GRCh38
rs752564005 Chr.10: 72341219-72341219 on Build GRCh38
rs763101111 Chr.10: 72341049-72341049 on Build GRCh38
rs765471839 Chr.10: 72340940-72340940 on Build GRCh38
rs139788055 Chr.10: 72341084-72341084 on Build GRCh38
rs143893741 Chr.10: 72341089-72341089 on Build GRCh38
rs760229144 Chr.10: 72341057-72341057 on Build GRCh38
rs769016982 Chr.10: 72341181-72341181 on Build GRCh38
rs768042650 Chr.10: 72340975-72340975 on Build GRCh38
rs891791777 Chr.10: 72341247-72341248 on Build GRCh38
rs746023978 Chr.10: 72341157-72341157 on Build GRCh38
rs148088395 Chr.10: 72341078-72341078 on Build GRCh38
rs375107524 Chr.10: 72341079-72341079 on Build GRCh38
rs528685833 Chr.10: 72341184-72341184 on Build GRCh38
rs536864860 Chr.10: 72341007-72341007 on Build GRCh38
rs772316600 Chr.10: 72341215-72341215 on Build GRCh38
rs778742260 Chr.10: 72341070-72341070 on Build GRCh38
rs111276924 Chr.10: 72341207-72341207 on Build GRCh38
rs11552373 Chr.10: 72341053-72341053 on Build GRCh38
rs753202870 Chr.10: 72341055-72341055 on Build GRCh38
rs761452946 Chr.10: 72341108-72341108 on Build GRCh38
rs375426252 Chr.10: 72341086-72341086 on Build GRCh38
rs372789312 Chr.10: 72341214-72341214 on Build GRCh38
rs143603767 Chr.10: 72341360-72341360 on Build GRCh38
rs761136354 Chr.10: 72341178-72341178 on Build GRCh38
rs1046388873 Chr.10: 72341294-72341294 on Build GRCh38

View Full Product Details