Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87094791-87095289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
UniGene
Hs.500419
Gene Chromosome Location
Chr.10: 87094478-87191468 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_011539877.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1 NM_001318904.1 NM_001318905.1 NM_001318906.1 NM_005271.4

Gene Symbol
GLUD1
Entrez Gene ID
2746
Gene Name
glutamate dehydrogenase 1
Gene Aliases
GDH, GDH1, GLUD
UniGene
Hs.500409
Gene Chromosome Location
Chr.10: 87050202-87095047 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_011539877.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1 NM_001318904.1 NM_001318905.1 NM_001318906.1 NM_005271.4

SNP Detail

SNP ID Chromosome Location
rs886047379 Chr.10: 87094813-87094813 on Build GRCh38
rs949637358 Chr.10: 87095226-87095226 on Build GRCh38
rs983042837 Chr.10: 87094853-87094853 on Build GRCh38
rs551417750 Chr.10: 87094850-87094850 on Build GRCh38
rs954116156 Chr.10: 87095183-87095183 on Build GRCh38
rs966044577 Chr.10: 87095163-87095163 on Build GRCh38
rs1015830747 Chr.10: 87095154-87095154 on Build GRCh38
rs1027186321 Chr.10: 87095121-87095121 on Build GRCh38
rs998169783 Chr.10: 87095173-87095173 on Build GRCh38
rs527251901 Chr.10: 87095032-87095032 on Build GRCh38
rs980743839 Chr.10: 87095227-87095227 on Build GRCh38
rs973121849 Chr.10: 87095108-87095108 on Build GRCh38
rs926604563 Chr.10: 87095234-87095234 on Build GRCh38
rs920051565 Chr.10: 87094839-87094839 on Build GRCh38
rs867264837 Chr.10: 87094979-87094979 on Build GRCh38
rs893429196 Chr.10: 87095020-87095030 on Build GRCh38
rs905296305 Chr.10: 87094912-87094912 on Build GRCh38
rs886047382 Chr.10: 87094965-87094965 on Build GRCh38
rs1045163127 Chr.10: 87094899-87094899 on Build GRCh38
rs918170554 Chr.10: 87095206-87095206 on Build GRCh38
rs886047380 Chr.10: 87094869-87094869 on Build GRCh38
rs1030227413 Chr.10: 87095026-87095026 on Build GRCh38
rs532878121 Chr.10: 87094847-87094847 on Build GRCh38
rs951560408 Chr.10: 87095127-87095127 on Build GRCh38
rs750771951 Chr.10: 87094810-87094810 on Build GRCh38
rs888612087 Chr.10: 87095050-87095050 on Build GRCh38
rs917935529 Chr.10: 87094854-87094854 on Build GRCh38
rs989532231 Chr.10: 87095189-87095189 on Build GRCh38
rs936720497 Chr.10: 87094972-87094972 on Build GRCh38
rs1028761395 Chr.10: 87095179-87095179 on Build GRCh38
rs886047381 Chr.10: 87094948-87094948 on Build GRCh38
rs936769558 Chr.10: 87095262-87095262 on Build GRCh38
rs1053566933 Chr.10: 87094974-87094974 on Build GRCh38
rs983478341 Chr.10: 87095178-87095179 on Build GRCh38
rs963125250 Chr.10: 87095096-87095096 on Build GRCh38
rs892269951 Chr.10: 87094997-87094997 on Build GRCh38
rs548584083 Chr.10: 87095061-87095061 on Build GRCh38
rs949410982 Chr.10: 87094869-87094869 on Build GRCh38
rs192909203 Chr.10: 87094851-87094851 on Build GRCh38

View Full Product Details