Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87151391-87151894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
UniGene
Hs.500419
Gene Chromosome Location
Chr.10: 87094478-87191468 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_006717905.3 XM_011539876.2 XM_011539877.2 XM_011539878.2 XM_011539879.2 XM_011539881.2 XM_011539882.2 XM_011539883.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371244 c.343A>T p.I115L Chr.10: 87151697-87151697 on Build GRCh38
3441257 c.368G>A p.C123Y Chr.10: 87151722-87151722 on Build GRCh38
6659605 c.78A>G p.T26T Chr.10: 87151432-87151432 on Build GRCh38
6659603 c.385G>A p.V129I Chr.10: 87151739-87151739 on Build GRCh38
3441255 c.231C>T p.F77F Chr.10: 87151585-87151585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768110510 Chr.10: 87151581-87151581 on Build GRCh38
rs79425056 Chr.10: 87151438-87151438 on Build GRCh38
rs749451832 Chr.10: 87151612-87151612 on Build GRCh38
rs117866241 Chr.10: 87151570-87151570 on Build GRCh38
rs749768385 Chr.10: 87151668-87151670 on Build GRCh38
rs750642041 Chr.10: 87151728-87151728 on Build GRCh38
rs775988391 Chr.10: 87151775-87151775 on Build GRCh38
rs749541799 Chr.10: 87151484-87151484 on Build GRCh38
rs192846566 Chr.10: 87151737-87151737 on Build GRCh38
rs756572167 Chr.10: 87151522-87151522 on Build GRCh38
rs780539827 Chr.10: 87151637-87151638 on Build GRCh38
rs761176344 Chr.10: 87151777-87151777 on Build GRCh38
rs758155969 Chr.10: 87151736-87151736 on Build GRCh38
rs745712637 Chr.10: 87151496-87151496 on Build GRCh38
rs372992990 Chr.10: 87151606-87151606 on Build GRCh38
rs756305590 Chr.10: 87151458-87151458 on Build GRCh38
rs755350958 Chr.10: 87151580-87151580 on Build GRCh38
rs3129519 Chr.10: 87151835-87151835 on Build GRCh38
rs566910680 Chr.10: 87151802-87151802 on Build GRCh38
rs781148725 Chr.10: 87151850-87151850 on Build GRCh38
rs768089724 Chr.10: 87151658-87151658 on Build GRCh38
rs770570795 Chr.10: 87151487-87151487 on Build GRCh38
rs759908160 Chr.10: 87151693-87151693 on Build GRCh38
rs763617741 Chr.10: 87151540-87151540 on Build GRCh38
rs368234758 Chr.10: 87151442-87151442 on Build GRCh38
rs898297395 Chr.10: 87151681-87151681 on Build GRCh38
rs781159357 Chr.10: 87151748-87151748 on Build GRCh38
rs1046763310 Chr.10: 87151722-87151722 on Build GRCh38
rs138542761 Chr.10: 87151670-87151670 on Build GRCh38
rs754811742 Chr.10: 87151849-87151849 on Build GRCh38
rs368795591 Chr.10: 87151827-87151827 on Build GRCh38
rs779061912 Chr.10: 87151631-87151631 on Build GRCh38
rs1023249725 Chr.10: 87151804-87151804 on Build GRCh38
rs747667298 Chr.10: 87151753-87151753 on Build GRCh38
rs541793920 Chr.10: 87151480-87151480 on Build GRCh38
rs926096323 Chr.10: 87151474-87151474 on Build GRCh38
rs534044465 Chr.10: 87151830-87151830 on Build GRCh38
rs550130200 Chr.10: 87151597-87151597 on Build GRCh38
rs896242774 Chr.10: 87151801-87151801 on Build GRCh38
rs375287268 Chr.10: 87151625-87151625 on Build GRCh38
rs769470354 Chr.10: 87151679-87151679 on Build GRCh38
rs145480311 Chr.10: 87151678-87151678 on Build GRCh38
rs768018782 Chr.10: 87151694-87151694 on Build GRCh38
rs751334486 Chr.10: 87151846-87151846 on Build GRCh38
rs980409152 Chr.10: 87151465-87151465 on Build GRCh38
rs3129521 Chr.10: 87151696-87151696 on Build GRCh38
rs769221597 Chr.10: 87151755-87151755 on Build GRCh38
rs772495709 Chr.10: 87151772-87151772 on Build GRCh38
rs571453392 Chr.10: 87151654-87151654 on Build GRCh38
rs778104235 Chr.10: 87151483-87151483 on Build GRCh38
rs761532885 Chr.10: 87151560-87151560 on Build GRCh38
rs376932843 Chr.10: 87151635-87151635 on Build GRCh38
rs768807234 Chr.10: 87151694-87151699 on Build GRCh38
rs772032737 Chr.10: 87151499-87151499 on Build GRCh38
rs774197213 Chr.10: 87151491-87151491 on Build GRCh38
rs764675285 Chr.10: 87151791-87151791 on Build GRCh38
rs143725514 Chr.10: 87151754-87151754 on Build GRCh38
rs762523029 Chr.10: 87151762-87151762 on Build GRCh38
rs781516860 Chr.10: 87151463-87151463 on Build GRCh38
rs750856045 Chr.10: 87151462-87151463 on Build GRCh38
rs768568606 Chr.10: 87151744-87151744 on Build GRCh38
rs371944597 Chr.10: 87151615-87151615 on Build GRCh38
rs140993693 Chr.10: 87151743-87151743 on Build GRCh38
rs775453459 Chr.10: 87151522-87151522 on Build GRCh38
rs772783439 Chr.10: 87151680-87151680 on Build GRCh38
rs761911237 Chr.10: 87151810-87151810 on Build GRCh38
rs753194156 Chr.10: 87151695-87151695 on Build GRCh38
rs3129520 Chr.10: 87151750-87151750 on Build GRCh38
rs776210406 Chr.10: 87151552-87151552 on Build GRCh38
rs137989844 Chr.10: 87151707-87151707 on Build GRCh38
rs760144773 Chr.10: 87151535-87151535 on Build GRCh38
rs942596676 Chr.10: 87151656-87151656 on Build GRCh38
rs756862299 Chr.10: 87151713-87151713 on Build GRCh38
rs753936276 Chr.10: 87151711-87151711 on Build GRCh38
rs778010684 Chr.10: 87151609-87151609 on Build GRCh38
rs765252814 Chr.10: 87151813-87151813 on Build GRCh38
rs779165002 Chr.10: 87151743-87151748 on Build GRCh38
rs779924621 Chr.10: 87151651-87151651 on Build GRCh38
rs750593174 Chr.10: 87151815-87151815 on Build GRCh38
rs755506139 Chr.10: 87151433-87151433 on Build GRCh38
rs745653598 Chr.10: 87151634-87151634 on Build GRCh38
rs745863745 Chr.10: 87151565-87151565 on Build GRCh38
rs757329100 Chr.10: 87151725-87151725 on Build GRCh38
rs751942922 Chr.10: 87151577-87151577 on Build GRCh38

View Full Product Details