Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87157956-87158389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
UniGene
Hs.500419
Gene Chromosome Location
Chr.10: 87094478-87191468 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_006717905.3 XM_011539876.2 XM_011539877.2 XM_011539878.2 XM_011539879.2 XM_011539881.2 XM_011539882.2 XM_011539883.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6257042 c.1528G>T p.V510F Chr.10: 87158050-87158050 on Build GRCh38
3441260 c.1608T>C p.S536S Chr.10: 87158130-87158130 on Build GRCh38
6257041 c.1528G>T p.V510F Chr.10: 87158050-87158050 on Build GRCh38
1349439 c.1542G>T p.E514D Chr.10: 87158064-87158064 on Build GRCh38
5088705 c.1542G>T p.E514D Chr.10: 87158064-87158064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761378237 Chr.10: 87158143-87158143 on Build GRCh38
rs768200675 Chr.10: 87158099-87158099 on Build GRCh38
rs12251817 Chr.10: 87158258-87158258 on Build GRCh38
rs749075667 Chr.10: 87158003-87158003 on Build GRCh38
rs561121893 Chr.10: 87158190-87158190 on Build GRCh38
rs3129493 Chr.10: 87158224-87158224 on Build GRCh38
rs760755506 Chr.10: 87158187-87158187 on Build GRCh38
rs752893852 Chr.10: 87157999-87157999 on Build GRCh38
rs142064377 Chr.10: 87158119-87158119 on Build GRCh38
rs531648617 Chr.10: 87158207-87158207 on Build GRCh38
rs4933430 Chr.10: 87158217-87158217 on Build GRCh38
rs187130729 Chr.10: 87158001-87158001 on Build GRCh38
rs3129495 Chr.10: 87158079-87158079 on Build GRCh38
rs374592298 Chr.10: 87158031-87158031 on Build GRCh38
rs780014562 Chr.10: 87158137-87158137 on Build GRCh38
rs763871341 Chr.10: 87158185-87158186 on Build GRCh38
rs942428466 Chr.10: 87158270-87158270 on Build GRCh38
rs989094140 Chr.10: 87158206-87158206 on Build GRCh38
rs200039650 Chr.10: 87158042-87158042 on Build GRCh38
rs376872876 Chr.10: 87158060-87158060 on Build GRCh38
rs577639245 Chr.10: 87158088-87158088 on Build GRCh38
rs993799111 Chr.10: 87158346-87158346 on Build GRCh38
rs766669099 Chr.10: 87158084-87158084 on Build GRCh38
rs3129496 Chr.10: 87158030-87158030 on Build GRCh38
rs778007749 Chr.10: 87158115-87158115 on Build GRCh38
rs560075779 Chr.10: 87158153-87158153 on Build GRCh38
rs776168090 Chr.10: 87158186-87158186 on Build GRCh38
rs140423511 Chr.10: 87158133-87158133 on Build GRCh38
rs3129494 Chr.10: 87158185-87158185 on Build GRCh38
rs776180006 Chr.10: 87158076-87158076 on Build GRCh38
rs993010492 Chr.10: 87158045-87158045 on Build GRCh38
rs372447614 Chr.10: 87158172-87158172 on Build GRCh38
rs562956269 Chr.10: 87158081-87158081 on Build GRCh38
rs771502249 Chr.10: 87158051-87158051 on Build GRCh38
rs778858312 Chr.10: 87158044-87158044 on Build GRCh38
rs760119008 Chr.10: 87158093-87158093 on Build GRCh38
rs752805811 Chr.10: 87158100-87158100 on Build GRCh38
rs765499914 Chr.10: 87158194-87158194 on Build GRCh38
rs551617411 Chr.10: 87158291-87158291 on Build GRCh38
rs771984382 Chr.10: 87158136-87158136 on Build GRCh38
rs756184116 Chr.10: 87158105-87158105 on Build GRCh38
rs571377498 Chr.10: 87158219-87158219 on Build GRCh38
rs768073132 Chr.10: 87158140-87158140 on Build GRCh38
rs148634820 Chr.10: 87158049-87158049 on Build GRCh38
rs768110784 Chr.10: 87158177-87158177 on Build GRCh38
rs746484005 Chr.10: 87158138-87158138 on Build GRCh38
rs146657981 Chr.10: 87158132-87158132 on Build GRCh38
rs549293304 Chr.10: 87158189-87158189 on Build GRCh38
rs898134550 Chr.10: 87158323-87158323 on Build GRCh38
rs889639223 Chr.10: 87158269-87158269 on Build GRCh38
rs781649531 Chr.10: 87158064-87158064 on Build GRCh38
rs371636091 Chr.10: 87158000-87158000 on Build GRCh38
rs769560246 Chr.10: 87158151-87158151 on Build GRCh38
rs757016279 Chr.10: 87158125-87158125 on Build GRCh38
rs774974544 Chr.10: 87158053-87158053 on Build GRCh38

View Full Product Details