Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87170144-87170674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
UniGene
Hs.500419
Gene Chromosome Location
Chr.10: 87094478-87191468 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_006717905.3 XM_011539876.2 XM_011539877.2 XM_011539878.2 XM_011539879.2 XM_011539881.2 XM_011539882.2 XM_011539883.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3675897 c.1648A>G p.S550G Chr.10: 87170492-87170492 on Build GRCh38
6659608 c.1793C>T p.A598V Chr.10: 87170637-87170637 on Build GRCh38
3935192 c.1756G>A p.E586K Chr.10: 87170600-87170600 on Build GRCh38
6659607 c.1793C>T p.A598V Chr.10: 87170637-87170637 on Build GRCh38
6348171 c.1657G>T p.V553L Chr.10: 87170501-87170501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531777516 Chr.10: 87170470-87170470 on Build GRCh38
rs146511329 Chr.10: 87170504-87170504 on Build GRCh38
rs888952582 Chr.10: 87170363-87170363 on Build GRCh38
rs147849599 Chr.10: 87170635-87170635 on Build GRCh38
rs1050245766 Chr.10: 87170362-87170362 on Build GRCh38
rs1052887209 Chr.10: 87170272-87170272 on Build GRCh38
rs979494703 Chr.10: 87170173-87170173 on Build GRCh38
rs925406436 Chr.10: 87170180-87170180 on Build GRCh38
rs923282750 Chr.10: 87170585-87170585 on Build GRCh38
rs1035779136 Chr.10: 87170454-87170454 on Build GRCh38
rs375357078 Chr.10: 87170631-87170631 on Build GRCh38
rs372338504 Chr.10: 87170615-87170615 on Build GRCh38
rs777514637 Chr.10: 87170587-87170587 on Build GRCh38
rs529413432 Chr.10: 87170624-87170624 on Build GRCh38
rs184722375 Chr.10: 87170354-87170354 on Build GRCh38
rs751441138 Chr.10: 87170562-87170562 on Build GRCh38
rs1005939500 Chr.10: 87170412-87170414 on Build GRCh38
rs141059392 Chr.10: 87170563-87170563 on Build GRCh38
rs766168388 Chr.10: 87170561-87170561 on Build GRCh38
rs779918276 Chr.10: 87170433-87170433 on Build GRCh38
rs762001305 Chr.10: 87170527-87170527 on Build GRCh38
rs781241618 Chr.10: 87170566-87170566 on Build GRCh38
rs566293742 Chr.10: 87170636-87170636 on Build GRCh38
rs758229862 Chr.10: 87170429-87170429 on Build GRCh38
rs761643329 Chr.10: 87170537-87170537 on Build GRCh38
rs751172169 Chr.10: 87170432-87170434 on Build GRCh38
rs532872611 Chr.10: 87170523-87170523 on Build GRCh38
rs775990308 Chr.10: 87170612-87170612 on Build GRCh38
rs756438954 Chr.10: 87170449-87170452 on Build GRCh38
rs552991945 Chr.10: 87170502-87170502 on Build GRCh38
rs368077394 Chr.10: 87170500-87170500 on Build GRCh38
rs766648156 Chr.10: 87170640-87170640 on Build GRCh38
rs754329562 Chr.10: 87170577-87170577 on Build GRCh38
rs552767285 Chr.10: 87170351-87170351 on Build GRCh38
rs956814744 Chr.10: 87170559-87170559 on Build GRCh38
rs925353113 Chr.10: 87170532-87170532 on Build GRCh38
rs11202365 Chr.10: 87170492-87170492 on Build GRCh38
rs772495492 Chr.10: 87170593-87170593 on Build GRCh38
rs561116306 Chr.10: 87170444-87170444 on Build GRCh38
rs761088281 Chr.10: 87170503-87170503 on Build GRCh38
rs777234889 Chr.10: 87170626-87170626 on Build GRCh38
rs370350253 Chr.10: 87170484-87170484 on Build GRCh38
rs755721302 Chr.10: 87170581-87170581 on Build GRCh38
rs540791344 Chr.10: 87170271-87170271 on Build GRCh38
rs773043851 Chr.10: 87170477-87170477 on Build GRCh38
rs746963777 Chr.10: 87170437-87170437 on Build GRCh38
rs754445962 Chr.10: 87170440-87170440 on Build GRCh38
rs766675425 Chr.10: 87170462-87170463 on Build GRCh38
rs747584230 Chr.10: 87170576-87170576 on Build GRCh38
rs748958433 Chr.10: 87170592-87170592 on Build GRCh38
rs747754735 Chr.10: 87170445-87170445 on Build GRCh38
rs765413835 Chr.10: 87170529-87170529 on Build GRCh38
rs878938676 Chr.10: 87170625-87170625 on Build GRCh38
rs758132082 Chr.10: 87170545-87170545 on Build GRCh38

View Full Product Details