Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87170827-87171335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
UniGene
Hs.500419
Gene Chromosome Location
Chr.10: 87094478-87191468 on Build GRCh38
Transcripts
NM_019054.2 XM_005269920.3 XM_006717903.3 XM_006717905.3 XM_011539876.2 XM_011539877.2 XM_011539878.2 XM_011539879.2 XM_011539881.2 XM_011539882.2 XM_011539883.2 XM_017016349.1 XM_017016350.1 XM_017016351.1 XM_017016352.1 XM_017016353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4181797 c.1883A>G p.E628G Chr.10: 87170894-87170894 on Build GRCh38
4181796 c.1882G>A p.E628K Chr.10: 87170893-87170893 on Build GRCh38
6316949 c.1831G>T p.A611S Chr.10: 87170842-87170842 on Build GRCh38
6316948 c.1831G>T p.A611S Chr.10: 87170842-87170842 on Build GRCh38
1967966 c.1883A>G p.E628G Chr.10: 87170894-87170894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057276261 Chr.10: 87171063-87171063 on Build GRCh38
rs576470288 Chr.10: 87170919-87170919 on Build GRCh38
rs986186648 Chr.10: 87170991-87170991 on Build GRCh38
rs905423730 Chr.10: 87171156-87171156 on Build GRCh38
rs1032434042 Chr.10: 87171191-87171192 on Build GRCh38
rs574616229 Chr.10: 87170872-87170872 on Build GRCh38
rs954864398 Chr.10: 87170982-87170982 on Build GRCh38
rs895606122 Chr.10: 87171072-87171072 on Build GRCh38
rs375331825 Chr.10: 87171035-87171035 on Build GRCh38
rs117800846 Chr.10: 87171219-87171219 on Build GRCh38
rs767354402 Chr.10: 87170855-87170855 on Build GRCh38
rs543806968 Chr.10: 87170958-87170958 on Build GRCh38
rs532833531 Chr.10: 87171018-87171018 on Build GRCh38
rs760683134 Chr.10: 87170867-87170867 on Build GRCh38
rs759207880 Chr.10: 87170851-87170851 on Build GRCh38
rs956764958 Chr.10: 87170934-87170934 on Build GRCh38
rs541782713 Chr.10: 87171062-87171062 on Build GRCh38
rs910451392 Chr.10: 87171008-87171008 on Build GRCh38
rs764182714 Chr.10: 87170868-87170868 on Build GRCh38
rs748571234 Chr.10: 87170918-87170918 on Build GRCh38
rs548502808 Chr.10: 87171157-87171157 on Build GRCh38
rs1044070932 Chr.10: 87171133-87171133 on Build GRCh38
rs565613899 Chr.10: 87171183-87171183 on Build GRCh38
rs1032484860 Chr.10: 87170910-87170910 on Build GRCh38
rs778389466 Chr.10: 87170889-87170889 on Build GRCh38
rs570146442 Chr.10: 87171307-87171307 on Build GRCh38
rs892635503 Chr.10: 87171213-87171213 on Build GRCh38
rs941941704 Chr.10: 87171012-87171012 on Build GRCh38
rs750113564 Chr.10: 87170890-87170890 on Build GRCh38
rs948373250 Chr.10: 87171075-87171075 on Build GRCh38
rs200421513 Chr.10: 87170912-87170912 on Build GRCh38
rs373802734 Chr.10: 87170898-87170898 on Build GRCh38
rs954839600 Chr.10: 87171293-87171293 on Build GRCh38
rs71477647 Chr.10: 87171298-87171298 on Build GRCh38
rs752507591 Chr.10: 87170865-87170865 on Build GRCh38
rs923261027 Chr.10: 87170978-87170978 on Build GRCh38
rs368106548 Chr.10: 87170893-87170893 on Build GRCh38
rs530367818 Chr.10: 87171142-87171142 on Build GRCh38
rs988633336 Chr.10: 87170954-87170954 on Build GRCh38
rs770265650 Chr.10: 87170931-87170931 on Build GRCh38
rs530540114 Chr.10: 87171286-87171286 on Build GRCh38
rs559685467 Chr.10: 87171108-87171108 on Build GRCh38
rs1030827149 Chr.10: 87171267-87171267 on Build GRCh38
rs551731244 Chr.10: 87171297-87171297 on Build GRCh38
rs565301910 Chr.10: 87171013-87171013 on Build GRCh38

View Full Product Details