Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91460252-91460762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD2
Entrez Gene ID
143279
Gene Name
HECT domain E3 ubiquitin protein ligase 2
Gene Aliases
-
UniGene
Hs.596096
Gene Chromosome Location
Chr.10: 91409280-91514829 on Build GRCh38
Transcripts
NM_001284274.1 NM_173497.3 NM_182765.4 NR_104291.1 XM_017015750.1 XM_017015751.1 NR_024467.1

Gene Symbol
HECTD2-AS1
Entrez Gene ID
100188947
Gene Name
HECTD2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.535293
Gene Chromosome Location
Chr.10: 91306962-91611460 on Build GRCh38
Transcripts
NM_001284274.1 NM_173497.3 NM_182765.4 NR_104291.1 XM_017015750.1 XM_017015751.1 NR_024467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6678701 c.314G>A p.R105H Chr.10: 91460472-91460472 on Build GRCh38
198466 c.340G>A p.E114K Chr.10: 91460498-91460498 on Build GRCh38
6067279 c.362C>A p.P121Q Chr.10: 91460520-91460520 on Build GRCh38
6067281 c.349G>T p.A117S Chr.10: 91460507-91460507 on Build GRCh38
4779991 c.333A>G p.S111S Chr.10: 91460491-91460491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558049885 Chr.10: 91460676-91460676 on Build GRCh38
rs759123338 Chr.10: 91460326-91460326 on Build GRCh38
rs779528097 Chr.10: 91460605-91460605 on Build GRCh38
rs751162122 Chr.10: 91460549-91460549 on Build GRCh38
rs753761920 Chr.10: 91460416-91460416 on Build GRCh38
rs537212342 Chr.10: 91460411-91460411 on Build GRCh38
rs752770990 Chr.10: 91460387-91460389 on Build GRCh38
rs904238749 Chr.10: 91460435-91460435 on Build GRCh38
rs963134891 Chr.10: 91460286-91460286 on Build GRCh38
rs61754654 Chr.10: 91460485-91460485 on Build GRCh38
rs992405059 Chr.10: 91460343-91460343 on Build GRCh38
rs758577340 Chr.10: 91460594-91460595 on Build GRCh38
rs148048715 Chr.10: 91460489-91460489 on Build GRCh38
rs368348007 Chr.10: 91460439-91460439 on Build GRCh38
rs757057019 Chr.10: 91460516-91460516 on Build GRCh38
rs763973629 Chr.10: 91460332-91460332 on Build GRCh38
rs771503582 Chr.10: 91460479-91460479 on Build GRCh38
rs746716912 Chr.10: 91460471-91460471 on Build GRCh38
rs970035916 Chr.10: 91460480-91460480 on Build GRCh38
rs980765481 Chr.10: 91460310-91460310 on Build GRCh38
rs574666940 Chr.10: 91460390-91460390 on Build GRCh38
rs185438597 Chr.10: 91460734-91460734 on Build GRCh38
rs115084987 Chr.10: 91460581-91460581 on Build GRCh38
rs914125790 Chr.10: 91460714-91460714 on Build GRCh38
rs765112994 Chr.10: 91460526-91460526 on Build GRCh38
rs372063843 Chr.10: 91460544-91460544 on Build GRCh38
rs1043615966 Chr.10: 91460311-91460311 on Build GRCh38
rs896321741 Chr.10: 91460422-91460422 on Build GRCh38
rs11186575 Chr.10: 91460347-91460347 on Build GRCh38
rs41286930 Chr.10: 91460472-91460472 on Build GRCh38
rs1017189632 Chr.10: 91460604-91460604 on Build GRCh38
rs921711039 Chr.10: 91460698-91460698 on Build GRCh38
rs182084837 Chr.10: 91460586-91460586 on Build GRCh38
rs201470639 Chr.10: 91460491-91460491 on Build GRCh38
rs200710574 Chr.10: 91460492-91460492 on Build GRCh38
rs530535213 Chr.10: 91460703-91460703 on Build GRCh38
rs752363576 Chr.10: 91460478-91460478 on Build GRCh38
rs569296139 Chr.10: 91460380-91460380 on Build GRCh38
rs1043884950 Chr.10: 91460398-91460398 on Build GRCh38
rs368934451 Chr.10: 91460483-91460483 on Build GRCh38
rs908008015 Chr.10: 91460583-91460583 on Build GRCh38
rs931646816 Chr.10: 91460729-91460729 on Build GRCh38
rs753564446 Chr.10: 91460508-91460508 on Build GRCh38
rs761015822 Chr.10: 91460498-91460498 on Build GRCh38
rs981303678 Chr.10: 91460619-91460619 on Build GRCh38
rs771450758 Chr.10: 91460593-91460593 on Build GRCh38
rs900146581 Chr.10: 91460291-91460291 on Build GRCh38
rs372075310 Chr.10: 91460442-91460442 on Build GRCh38
rs1033677358 Chr.10: 91460634-91460637 on Build GRCh38
rs754729832 Chr.10: 91460565-91460565 on Build GRCh38
rs779535332 Chr.10: 91460470-91460470 on Build GRCh38
rs939617281 Chr.10: 91460342-91460342 on Build GRCh38
rs763792046 Chr.10: 91460505-91460505 on Build GRCh38
rs1017008973 Chr.10: 91460274-91460274 on Build GRCh38
rs745324269 Chr.10: 91460443-91460443 on Build GRCh38
rs561481989 Chr.10: 91460664-91460664 on Build GRCh38
rs368901450 Chr.10: 91460541-91460541 on Build GRCh38
rs759697775 Chr.10: 91460376-91460376 on Build GRCh38
rs961021313 Chr.10: 91460669-91460669 on Build GRCh38
rs200442528 Chr.10: 91460528-91460528 on Build GRCh38
rs772030060 Chr.10: 91460612-91460612 on Build GRCh38
rs999398993 Chr.10: 91460354-91460354 on Build GRCh38
rs755833365 Chr.10: 91460395-91460395 on Build GRCh38
rs773774144 Chr.10: 91460474-91460474 on Build GRCh38
rs548800940 Chr.10: 91460646-91460646 on Build GRCh38
rs757949507 Chr.10: 91460444-91460444 on Build GRCh38
rs931677851 Chr.10: 91460308-91460308 on Build GRCh38
rs746434702 Chr.10: 91460606-91460606 on Build GRCh38

View Full Product Details