Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91477946-91478449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD2
Entrez Gene ID
143279
Gene Name
HECT domain E3 ubiquitin protein ligase 2
Gene Aliases
-
UniGene
Hs.596096
Gene Chromosome Location
Chr.10: 91409280-91514829 on Build GRCh38
Transcripts
NM_001284274.1 NM_182765.4 NR_104291.1 XM_017015750.1 XM_017015751.1 NR_024467.1

Gene Symbol
HECTD2-AS1
Entrez Gene ID
100188947
Gene Name
HECTD2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.535293
Gene Chromosome Location
Chr.10: 91306962-91611460 on Build GRCh38
Transcripts
NM_001284274.1 NM_182765.4 NR_104291.1 XM_017015750.1 XM_017015751.1 NR_024467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6461390 c.647T>C p.L216P Chr.10: 91478247-91478247 on Build GRCh38
6897699 c.610G>A p.V204I Chr.10: 91478210-91478210 on Build GRCh38
5448059 c.643A>C p.S215R Chr.10: 91478243-91478243 on Build GRCh38
4689901 c.632G>A p.G211E Chr.10: 91478232-91478232 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533267334 Chr.10: 91478189-91478189 on Build GRCh38
rs889354306 Chr.10: 91478149-91478149 on Build GRCh38
rs369284664 Chr.10: 91478210-91478210 on Build GRCh38
rs868725967 Chr.10: 91478396-91478396 on Build GRCh38
rs201945945 Chr.10: 91478294-91478294 on Build GRCh38
rs777771105 Chr.10: 91478253-91478253 on Build GRCh38
rs929361784 Chr.10: 91478133-91478133 on Build GRCh38
rs776326489 Chr.10: 91478195-91478195 on Build GRCh38
rs778601867 Chr.10: 91478242-91478242 on Build GRCh38
rs191367638 Chr.10: 91478393-91478393 on Build GRCh38
rs1039263555 Chr.10: 91478205-91478205 on Build GRCh38
rs748333373 Chr.10: 91478319-91478319 on Build GRCh38
rs186877594 Chr.10: 91478134-91478134 on Build GRCh38
rs376510687 Chr.10: 91478204-91478204 on Build GRCh38
rs764376822 Chr.10: 91478198-91478199 on Build GRCh38
rs1009076800 Chr.10: 91478149-91478149 on Build GRCh38
rs972968167 Chr.10: 91478100-91478100 on Build GRCh38
rs772024766 Chr.10: 91478297-91478297 on Build GRCh38
rs774079201 Chr.10: 91478154-91478154 on Build GRCh38
rs776073775 Chr.10: 91478307-91478307 on Build GRCh38
rs893888129 Chr.10: 91478097-91478098 on Build GRCh38
rs935176650 Chr.10: 91478070-91478070 on Build GRCh38
rs754717871 Chr.10: 91478182-91478182 on Build GRCh38
rs745662262 Chr.10: 91478160-91478160 on Build GRCh38
rs909723827 Chr.10: 91478064-91478064 on Build GRCh38
rs561528869 Chr.10: 91478074-91478074 on Build GRCh38
rs746993986 Chr.10: 91478300-91478300 on Build GRCh38
rs751858400 Chr.10: 91478207-91478207 on Build GRCh38
rs570367558 Chr.10: 91478172-91478172 on Build GRCh38
rs113450507 Chr.10: 91478110-91478110 on Build GRCh38
rs927753194 Chr.10: 91478032-91478032 on Build GRCh38
rs756193649 Chr.10: 91478237-91478237 on Build GRCh38
rs779208551 Chr.10: 91478284-91478284 on Build GRCh38
rs374348428 Chr.10: 91478352-91478352 on Build GRCh38
rs149453116 Chr.10: 91478023-91478023 on Build GRCh38
rs1046248825 Chr.10: 91478025-91478025 on Build GRCh38
rs947974248 Chr.10: 91478009-91478009 on Build GRCh38
rs756986752 Chr.10: 91478277-91478277 on Build GRCh38
rs768201149 Chr.10: 91478302-91478302 on Build GRCh38
rs757353542 Chr.10: 91478283-91478283 on Build GRCh38
rs761433244 Chr.10: 91478198-91478198 on Build GRCh38
rs879036242 Chr.10: 91478101-91478101 on Build GRCh38
rs546827469 Chr.10: 91478305-91478305 on Build GRCh38
rs1052489453 Chr.10: 91478092-91478092 on Build GRCh38
rs529365103 Chr.10: 91477990-91477990 on Build GRCh38
rs754084276 Chr.10: 91478271-91478271 on Build GRCh38
rs916497099 Chr.10: 91477968-91477969 on Build GRCh38
rs919375686 Chr.10: 91478122-91478122 on Build GRCh38
rs894042831 Chr.10: 91478209-91478209 on Build GRCh38
rs751875620 Chr.10: 91478251-91478251 on Build GRCh38
rs776547956 Chr.10: 91478353-91478353 on Build GRCh38
rs952610406 Chr.10: 91478337-91478338 on Build GRCh38
rs79168137 Chr.10: 91478006-91478006 on Build GRCh38
rs1010897640 Chr.10: 91478102-91478102 on Build GRCh38
rs775591511 Chr.10: 91478176-91478176 on Build GRCh38
rs546270886 Chr.10: 91478193-91478193 on Build GRCh38
rs200421586 Chr.10: 91478236-91478236 on Build GRCh38
rs1025385147 Chr.10: 91478322-91478323 on Build GRCh38
rs369872084 Chr.10: 91478350-91478350 on Build GRCh38
rs780109529 Chr.10: 91478299-91478299 on Build GRCh38
rs143008675 Chr.10: 91478150-91478150 on Build GRCh38
rs755687291 Chr.10: 91477980-91477980 on Build GRCh38
rs759911720 Chr.10: 91478208-91478208 on Build GRCh38
rs1011104695 Chr.10: 91478316-91478316 on Build GRCh38

View Full Product Details