Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92893271-92893773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6
Entrez Gene ID
54536
Gene Name
exocyst complex component 6
Gene Aliases
EXOC6A, SEC15, SEC15L, SEC15L1, SEC15L3, Sec15p
UniGene
Hs.655657
Gene Chromosome Location
Chr.10: 92826831-93059494 on Build GRCh38
Transcripts
NM_001013848.3 NM_001319194.1 NM_001319195.1 NM_001319200.1 NM_019053.5 XM_017016344.1 XM_017016345.1 XM_017016346.1 XM_017016347.1 XM_017016348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807998 c.247G>T p.E83* Chr.10: 92893509-92893509 on Build GRCh38
1661722 c.172C>A p.H58N Chr.10: 92893419-92893419 on Build GRCh38
7261560 c.175G>A p.A59T Chr.10: 92893374-92893374 on Build GRCh38
5524918 c.288C>T p.L96L Chr.10: 92893487-92893487 on Build GRCh38
921493 c.272A>C p.K91T Chr.10: 92893519-92893519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748815462 Chr.10: 92893414-92893414 on Build GRCh38
rs973826119 Chr.10: 92893723-92893723 on Build GRCh38
rs968690367 Chr.10: 92893631-92893631 on Build GRCh38
rs575416565 Chr.10: 92893638-92893638 on Build GRCh38
rs899749219 Chr.10: 92893591-92893591 on Build GRCh38
rs745468180 Chr.10: 92893468-92893468 on Build GRCh38
rs773642694 Chr.10: 92893501-92893501 on Build GRCh38
rs908185010 Chr.10: 92893661-92893661 on Build GRCh38
rs762344356 Chr.10: 92893349-92893349 on Build GRCh38
rs374679977 Chr.10: 92893360-92893360 on Build GRCh38
rs767767281 Chr.10: 92893529-92893529 on Build GRCh38
rs185410835 Chr.10: 92893583-92893583 on Build GRCh38
rs201253450 Chr.10: 92893520-92893520 on Build GRCh38
rs754834070 Chr.10: 92893376-92893376 on Build GRCh38
rs755910097 Chr.10: 92893550-92893550 on Build GRCh38
rs957534580 Chr.10: 92893506-92893506 on Build GRCh38
rs1021667627 Chr.10: 92893623-92893623 on Build GRCh38
rs539465103 Chr.10: 92893334-92893334 on Build GRCh38
rs138507234 Chr.10: 92893394-92893394 on Build GRCh38
rs955621036 Chr.10: 92893604-92893604 on Build GRCh38
rs781202291 Chr.10: 92893321-92893321 on Build GRCh38
rs766590934 Chr.10: 92893372-92893372 on Build GRCh38
rs748794925 Chr.10: 92893338-92893338 on Build GRCh38
rs766678586 Chr.10: 92893537-92893538 on Build GRCh38
rs555709067 Chr.10: 92893588-92893588 on Build GRCh38
rs754661026 Chr.10: 92893317-92893317 on Build GRCh38
rs780890935 Chr.10: 92893379-92893379 on Build GRCh38
rs576007847 Chr.10: 92893620-92893620 on Build GRCh38
rs372961243 Chr.10: 92893522-92893522 on Build GRCh38
rs145875144 Chr.10: 92893500-92893500 on Build GRCh38
rs200297922 Chr.10: 92893400-92893400 on Build GRCh38
rs770510321 Chr.10: 92893435-92893435 on Build GRCh38
rs751673313 Chr.10: 92893375-92893375 on Build GRCh38
rs141813516 Chr.10: 92893514-92893514 on Build GRCh38
rs528958776 Chr.10: 92893343-92893343 on Build GRCh38
rs763016416 Chr.10: 92893369-92893369 on Build GRCh38
rs771722053 Chr.10: 92893474-92893474 on Build GRCh38
rs202055138 Chr.10: 92893471-92893471 on Build GRCh38
rs753782586 Chr.10: 92893560-92893560 on Build GRCh38
rs756178975 Chr.10: 92893389-92893389 on Build GRCh38
rs747997207 Chr.10: 92893333-92893333 on Build GRCh38
rs201788696 Chr.10: 92893508-92893508 on Build GRCh38
rs201775806 Chr.10: 92893406-92893406 on Build GRCh38
rs143952914 Chr.10: 92893457-92893457 on Build GRCh38
rs776905084 Chr.10: 92893476-92893476 on Build GRCh38
rs746888356 Chr.10: 92893299-92893299 on Build GRCh38
rs762148486 Chr.10: 92893479-92893479 on Build GRCh38
rs1028598805 Chr.10: 92893590-92893590 on Build GRCh38
rs1045170377 Chr.10: 92893391-92893391 on Build GRCh38
rs538014835 Chr.10: 92893737-92893737 on Build GRCh38
rs919766667 Chr.10: 92893732-92893732 on Build GRCh38
rs368246632 Chr.10: 92893567-92893567 on Build GRCh38
rs750559663 Chr.10: 92893367-92893367 on Build GRCh38
rs998834646 Chr.10: 92893731-92893731 on Build GRCh38
rs752501853 Chr.10: 92893531-92893531 on Build GRCh38
rs886836195 Chr.10: 92893505-92893505 on Build GRCh38
rs777870455 Chr.10: 92893554-92893554 on Build GRCh38
rs371363561 Chr.10: 92893346-92893346 on Build GRCh38
rs752514599 Chr.10: 92893383-92893383 on Build GRCh38
rs939776470 Chr.10: 92893690-92893690 on Build GRCh38
rs772898237 Chr.10: 92893336-92893336 on Build GRCh38

View Full Product Details