The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181836.5 | NP_861974.1 | 3 - 3 | 2058 | 134 |
GenBank mRNA | AB002450.1 | - | 1 - 1 | 852 | 134 |
AK074962.1 | - | 3 - 3 | 1878 | 134 | |
BC019349.2 | - | 3 - 3 | 1904 | 134 | |
BC107426.1 | - | 2 - 2 | 135 |