The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033117.2 | NP_001028289.1 | 20 - 21 | 3223 | 73 |
NM_014850.3 | NP_055665.1 | 20 - 21 | 3295 | 73 | |
XM_011534295.1 | XP_011532597.1 | 21 - 22 | 3310 | 73 | |
XM_011534296.1 | XP_011532598.1 | 22 - 23 | 3427 | 73 | |
XM_011534301.2 | XP_011532603.1 | 18 - 19 | 5139 | 73 | |
XM_011534303.1 | XP_011532605.1 | 16 - 17 | 2035 | 73 | |
XM_017007574.1 | XP_016863063.1 | 20 - 21 | 3633 | 73 | |
XM_017007575.1 | XP_016863064.1 | 20 - 21 | 3561 | 73 | |
XM_017007576.1 | XP_016863065.1 | 22 - 23 | 2802 | 73 | |
XM_017007577.1 | XP_016863066.1 | 22 - 23 | 2741 | 73 | |
XM_017007578.1 | XP_016863067.1 | 20 - 21 | 3661 | 73 | |
GenBank mRNA | AB007871.2 | - | 20 - 21 | 2447 | 73 |
AF427144.1 | - | 20 - 21 | 2663 | 73 | |
AF464189.1 | - | 20 - 21 | 2751 | 73 |