Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNMAL2
Entrez Gene ID
57469
Gene Name
paraneoplastic Ma antigen family-like 2
Gene Aliases
-
Chromosome Location
Chr.19: 46491191 - 46495912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020709.2 NP_065760.1 1 - 1 1372 90
GenBank mRNA AB033009.1 - 1 - 1 49 90
BX390603.1 - 1 - 1 581 90

View Full Product Details