The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258415.1 | NP_001245344.1 | 2 - 3 | 479 | 95 |
NM_001258416.1 | NP_001245345.1 | 3 - 4 | 365 | 95 | |
NM_001258417.1 | NP_001245346.1 | 2 - 3 | 179 | 95 | |
NM_001258418.1 | NP_001245347.1 | 2 - 3 | 297 | 95 | |
NM_020809.3 | NP_065860.2 | 3 - 4 | 476 | 95 | |
XM_006718893.3 | XP_006718956.1 | 2 - 3 | 385 | 95 | |
GenBank mRNA | AB037812.1 | - | 2 - 3 | 201 | 95 |
AK307602.1 | - | 2 - 3 | 385 | 95 | |
AY496263.1 | - | 3 - 4 | 476 | 95 | |
AY496264.1 | - | 2 - 3 | 179 | 95 | |
AY496265.1 | - | 2 - 3 | 297 | 95 | |
AY496266.1 | - | 2 - 3 | 479 | 95 | |
AY496267.1 | - | 3 - 4 | 365 | 95 |