Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 417852-418349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
UniGene
Hs.501622
Gene Chromosome Location
Chr.11: 417930-442011 on Build GRCh38
Transcripts
NM_001012302.2 XM_011520053.2

SNP Detail

SNP ID Chromosome Location
rs546395889 Chr.11: 418301-418301 on Build GRCh38
rs371329952 Chr.11: 418069-418069 on Build GRCh38
rs577353461 Chr.11: 418300-418300 on Build GRCh38
rs555969920 Chr.11: 418078-418078 on Build GRCh38
rs770891121 Chr.11: 418120-418120 on Build GRCh38
rs541394174 Chr.11: 418093-418093 on Build GRCh38
rs921257257 Chr.11: 417917-417917 on Build GRCh38
rs1044944161 Chr.11: 418290-418290 on Build GRCh38
rs911741033 Chr.11: 417990-417990 on Build GRCh38
rs1050364306 Chr.11: 418225-418225 on Build GRCh38
rs1028350586 Chr.11: 418177-418177 on Build GRCh38
rs11539437 Chr.11: 418074-418074 on Build GRCh38
rs533854526 Chr.11: 417963-417963 on Build GRCh38
rs987174748 Chr.11: 418180-418180 on Build GRCh38
rs769357973 Chr.11: 418222-418222 on Build GRCh38
rs994516900 Chr.11: 418319-418320 on Build GRCh38
rs918146407 Chr.11: 417918-417918 on Build GRCh38
rs930602165 Chr.11: 418252-418252 on Build GRCh38
rs868868537 Chr.11: 418304-418304 on Build GRCh38
rs780787277 Chr.11: 418322-418322 on Build GRCh38
rs968267992 Chr.11: 417908-417908 on Build GRCh38
rs907541195 Chr.11: 418186-418186 on Build GRCh38
rs562943054 Chr.11: 418317-418317 on Build GRCh38
rs7228 Chr.11: 418025-418025 on Build GRCh38
rs141340877 Chr.11: 417888-417888 on Build GRCh38
rs931479232 Chr.11: 418220-418220 on Build GRCh38
rs568314803 Chr.11: 417923-417923 on Build GRCh38
rs920421863 Chr.11: 418251-418251 on Build GRCh38
rs971928971 Chr.11: 418249-418249 on Build GRCh38
rs986884539 Chr.11: 417947-417947 on Build GRCh38
rs961720514 Chr.11: 418236-418236 on Build GRCh38
rs138448794 Chr.11: 418098-418098 on Build GRCh38
rs75767088 Chr.11: 418079-418079 on Build GRCh38
rs775763071 Chr.11: 418308-418308 on Build GRCh38
rs898661474 Chr.11: 417997-417997 on Build GRCh38
rs183826257 Chr.11: 418072-418072 on Build GRCh38
rs930527036 Chr.11: 418059-418059 on Build GRCh38
rs1034080945 Chr.11: 418147-418147 on Build GRCh38
rs15185 Chr.11: 418142-418142 on Build GRCh38
rs570550547 Chr.11: 418063-418063 on Build GRCh38
rs940547964 Chr.11: 418006-418006 on Build GRCh38
rs1022847853 Chr.11: 418129-418129 on Build GRCh38
rs759023553 Chr.11: 418195-418195 on Build GRCh38
rs889847047 Chr.11: 418247-418247 on Build GRCh38
rs10331 Chr.11: 418179-418179 on Build GRCh38
rs960074176 Chr.11: 418162-418162 on Build GRCh38
rs975658408 Chr.11: 418140-418140 on Build GRCh38
rs540569891 Chr.11: 418240-418240 on Build GRCh38
rs778102064 Chr.11: 417932-417932 on Build GRCh38
rs531704965 Chr.11: 417887-417887 on Build GRCh38
rs1009758094 Chr.11: 418124-418124 on Build GRCh38
rs975415193 Chr.11: 417913-417913 on Build GRCh38
rs560869682 Chr.11: 418299-418299 on Build GRCh38
rs940263280 Chr.11: 417989-417989 on Build GRCh38
rs908643338 Chr.11: 418205-418205 on Build GRCh38
rs777578762 Chr.11: 417950-417950 on Build GRCh38
rs745324544 Chr.11: 418326-418326 on Build GRCh38
rs1021533236 Chr.11: 418100-418103 on Build GRCh38
rs972972168 Chr.11: 418204-418204 on Build GRCh38
rs899084142 Chr.11: 418049-418049 on Build GRCh38
rs968298978 Chr.11: 418130-418130 on Build GRCh38

View Full Product Details