Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE5
Entrez Gene ID
57699
Gene Name
copine 5
Gene Aliases
COPN5, CPN5
Chromosome Location
Chr.6: 36740773 - 36839444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314017.1 NP_001300946.1 1 - 2 167 72
NM_020939.1 NP_065990.1 1 - 2 167 72
XM_005249247.2 XP_005249304.1 1 - 2 167 72
XM_011514768.1 XP_011513070.1 1 - 2 167 72
XM_011514769.1 XP_011513071.1 1 - 2 167 72
XM_011514772.1 XP_011513074.1 1 - 2 167 72
GenBank mRNA AB046819.1 - 1 - 2 725 72
DA265730.1 - 1 - 2 167 72

View Full Product Details