Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CES5A
Entrez Gene ID
221223
Gene Name
carboxylesterase 5A
Gene Aliases
CAUXIN, CES4C1, CES5, CES7, HEL126
Chromosome Location
Chr.16: 55846154 - 55956031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143685.1 NP_001137157.1 9 - 10 1277 63
NM_001190158.1 NP_001177087.1 10 - 11 1361 63
NM_145024.2 NP_659461.1 9 - 10 1277 63
GenBank mRNA AK056109.1 - 9 - 10 1277 63
AK090997.1 - 9 - 10 1203 63
AK294334.1 - 10 - 11 1361 63
AY907669.1 - 9 - 10 1204 63
BC039073.1 - 9 - 10 1290 63
BC069501.1 - 9 - 10 1248 63
BC069548.1 - 9 - 10 1247 63
BC117126.1 - 9 - 10 1250 63
BC143692.1 - 9 - 10 1250 63
EU794678.1 - 9 - 10 1277 63

View Full Product Details