Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31519890-31520404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP4
Entrez Gene ID
26610
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
UniGene
Hs.175534
Gene Chromosome Location
Chr.11: 31509729-31784525 on Build GRCh38
Transcripts
NM_001288725.1 NM_001288726.1 NM_019040.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4954460 c.251T>G p.L84R Chr.11: 31520083-31520083 on Build GRCh38
6335896 c.238G>T p.V80F Chr.11: 31520070-31520070 on Build GRCh38
6653551 c.236C>T p.A79V Chr.11: 31520068-31520068 on Build GRCh38
6653549 c.250C>A p.L84I Chr.11: 31520082-31520082 on Build GRCh38
6653555 c.241G>T p.G81* Chr.11: 31520073-31520073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773211070 Chr.11: 31520131-31520131 on Build GRCh38
rs1019977960 Chr.11: 31520310-31520310 on Build GRCh38
rs772561401 Chr.11: 31520075-31520075 on Build GRCh38
rs34556981 Chr.11: 31520316-31520317 on Build GRCh38
rs188498302 Chr.11: 31520197-31520197 on Build GRCh38
rs1013359610 Chr.11: 31520360-31520360 on Build GRCh38
rs373853950 Chr.11: 31520069-31520069 on Build GRCh38
rs939528948 Chr.11: 31520208-31520212 on Build GRCh38
rs940870077 Chr.11: 31520027-31520027 on Build GRCh38
rs1010368676 Chr.11: 31520300-31520300 on Build GRCh38
rs1043656584 Chr.11: 31520193-31520193 on Build GRCh38
rs760791371 Chr.11: 31520078-31520078 on Build GRCh38
rs767222314 Chr.11: 31520014-31520014 on Build GRCh38
rs571540938 Chr.11: 31520138-31520138 on Build GRCh38
rs987323929 Chr.11: 31520376-31520376 on Build GRCh38
rs768804650 Chr.11: 31520098-31520098 on Build GRCh38
rs761976402 Chr.11: 31520124-31520124 on Build GRCh38
rs895571693 Chr.11: 31520266-31520266 on Build GRCh38
rs868018085 Chr.11: 31520256-31520256 on Build GRCh38
rs184900808 Chr.11: 31519948-31519948 on Build GRCh38
rs753352984 Chr.11: 31520077-31520077 on Build GRCh38
rs889136804 Chr.11: 31520142-31520142 on Build GRCh38
rs547529355 Chr.11: 31520351-31520351 on Build GRCh38
rs550363782 Chr.11: 31520128-31520128 on Build GRCh38
rs192277325 Chr.11: 31520372-31520372 on Build GRCh38
rs1031279754 Chr.11: 31520309-31520309 on Build GRCh38
rs775446929 Chr.11: 31519981-31519981 on Build GRCh38
rs1000141109 Chr.11: 31520338-31520338 on Build GRCh38
rs776894002 Chr.11: 31520114-31520114 on Build GRCh38
rs371555226 Chr.11: 31520155-31520155 on Build GRCh38
rs201656765 Chr.11: 31520033-31520033 on Build GRCh38
rs202180164 Chr.11: 31520070-31520070 on Build GRCh38
rs765893575 Chr.11: 31519988-31519988 on Build GRCh38
rs759484723 Chr.11: 31520133-31520133 on Build GRCh38
rs1057324987 Chr.11: 31520212-31520212 on Build GRCh38
rs978648595 Chr.11: 31519964-31519964 on Build GRCh38
rs752452109 Chr.11: 31520365-31520365 on Build GRCh38
rs895505738 Chr.11: 31519935-31519937 on Build GRCh38
rs1035432553 Chr.11: 31519926-31519926 on Build GRCh38
rs923371135 Chr.11: 31520072-31520072 on Build GRCh38
rs755782101 Chr.11: 31520022-31520022 on Build GRCh38
rs930914312 Chr.11: 31520162-31520162 on Build GRCh38
rs370482596 Chr.11: 31520237-31520237 on Build GRCh38
rs149823111 Chr.11: 31519957-31519957 on Build GRCh38
rs747383466 Chr.11: 31520094-31520094 on Build GRCh38
rs764920621 Chr.11: 31520346-31520346 on Build GRCh38
rs904167533 Chr.11: 31520330-31520330 on Build GRCh38
rs746405426 Chr.11: 31520074-31520074 on Build GRCh38
rs991809616 Chr.11: 31519943-31519943 on Build GRCh38
rs779939365 Chr.11: 31520128-31520128 on Build GRCh38
rs972679346 Chr.11: 31520064-31520064 on Build GRCh38
rs754537171 Chr.11: 31520194-31520194 on Build GRCh38
rs567952199 Chr.11: 31520036-31520036 on Build GRCh38
rs985054077 Chr.11: 31519972-31519972 on Build GRCh38
rs752479738 Chr.11: 31520015-31520015 on Build GRCh38

View Full Product Details