Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31647651-31648154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP4
Entrez Gene ID
26610
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
UniGene
Hs.175534
Gene Chromosome Location
Chr.11: 31509729-31784525 on Build GRCh38
Transcripts
NM_001288725.1 NM_001288726.1 NM_019040.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7102808 c.1000G>C p.E334Q Chr.11: 31647813-31647813 on Build GRCh38
466736 c.928-1G>T p.? Chr.11: 31647740-31647740 on Build GRCh38
3446751 c.1016C>T p.P339L Chr.11: 31647829-31647829 on Build GRCh38
5001943 c.964G>A p.D322N Chr.11: 31647777-31647777 on Build GRCh38
3446752 c.1016C>T p.P339L Chr.11: 31647829-31647829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556086865 Chr.11: 31647768-31647768 on Build GRCh38
rs906080583 Chr.11: 31647942-31647942 on Build GRCh38
rs567102192 Chr.11: 31648040-31648040 on Build GRCh38
rs751972970 Chr.11: 31647778-31647778 on Build GRCh38
rs954044444 Chr.11: 31647989-31647989 on Build GRCh38
rs769315762 Chr.11: 31647859-31647859 on Build GRCh38
rs538301162 Chr.11: 31647959-31647959 on Build GRCh38
rs745833536 Chr.11: 31647730-31647730 on Build GRCh38
rs777968972 Chr.11: 31647825-31647825 on Build GRCh38
rs1019645656 Chr.11: 31648030-31648030 on Build GRCh38
rs372439263 Chr.11: 31647842-31647842 on Build GRCh38
rs765864843 Chr.11: 31647896-31647896 on Build GRCh38
rs537621638 Chr.11: 31648062-31648062 on Build GRCh38
rs773010532 Chr.11: 31647754-31647754 on Build GRCh38
rs202114087 Chr.11: 31647743-31647743 on Build GRCh38
rs369921889 Chr.11: 31647738-31647738 on Build GRCh38
rs775421034 Chr.11: 31647734-31647734 on Build GRCh38
rs1017083398 Chr.11: 31647994-31647994 on Build GRCh38
rs761539164 Chr.11: 31647858-31647858 on Build GRCh38
rs377086142 Chr.11: 31648059-31648059 on Build GRCh38
rs759777429 Chr.11: 31647830-31647830 on Build GRCh38
rs1006881183 Chr.11: 31648021-31648021 on Build GRCh38
rs756234913 Chr.11: 31647824-31647824 on Build GRCh38
rs762349112 Chr.11: 31647895-31647895 on Build GRCh38
rs764805051 Chr.11: 31647759-31647759 on Build GRCh38
rs898687851 Chr.11: 31648067-31648067 on Build GRCh38
rs766767508 Chr.11: 31647777-31647777 on Build GRCh38
rs896189449 Chr.11: 31648041-31648041 on Build GRCh38
rs369889993 Chr.11: 31647810-31647810 on Build GRCh38
rs1048978214 Chr.11: 31647724-31647724 on Build GRCh38
rs770895253 Chr.11: 31647883-31647883 on Build GRCh38
rs375515034 Chr.11: 31647713-31647713 on Build GRCh38
rs751919719 Chr.11: 31647696-31647696 on Build GRCh38
rs750968466 Chr.11: 31647692-31647692 on Build GRCh38
rs771888940 Chr.11: 31647732-31647732 on Build GRCh38
rs777648639 Chr.11: 31647728-31647728 on Build GRCh38
rs1052886012 Chr.11: 31647881-31647881 on Build GRCh38
rs919058664 Chr.11: 31648116-31648116 on Build GRCh38
rs373360320 Chr.11: 31647749-31647749 on Build GRCh38
rs772083794 Chr.11: 31647828-31647828 on Build GRCh38
rs114605813 Chr.11: 31648106-31648106 on Build GRCh38
rs755236226 Chr.11: 31647701-31647701 on Build GRCh38
rs745442570 Chr.11: 31648089-31648089 on Build GRCh38
rs746889942 Chr.11: 31647834-31647834 on Build GRCh38
rs375519266 Chr.11: 31647750-31647750 on Build GRCh38
rs781373805 Chr.11: 31647708-31647708 on Build GRCh38
rs1001490982 Chr.11: 31647927-31647927 on Build GRCh38
rs7102844 Chr.11: 31647691-31647691 on Build GRCh38
rs372481468 Chr.11: 31648074-31648074 on Build GRCh38
rs758762353 Chr.11: 31647693-31647693 on Build GRCh38
rs763586359 Chr.11: 31647774-31647774 on Build GRCh38
rs574668037 Chr.11: 31647882-31647882 on Build GRCh38
rs772820950 Chr.11: 31647884-31647884 on Build GRCh38
rs1045428551 Chr.11: 31647910-31647910 on Build GRCh38
rs902692650 Chr.11: 31647869-31647869 on Build GRCh38
rs768602850 Chr.11: 31647835-31647835 on Build GRCh38
rs780478390 Chr.11: 31647695-31647695 on Build GRCh38
rs755362706 Chr.11: 31647806-31647806 on Build GRCh38
rs533838732 Chr.11: 31647823-31647823 on Build GRCh38
rs776500513 Chr.11: 31647847-31647847 on Build GRCh38
rs533474275 Chr.11: 31647715-31647715 on Build GRCh38
rs1017528539 Chr.11: 31647688-31647688 on Build GRCh38
rs767672001 Chr.11: 31647909-31647909 on Build GRCh38
rs749842817 Chr.11: 31647760-31647760 on Build GRCh38
rs749321679 Chr.11: 31647827-31647827 on Build GRCh38
rs769362365 Chr.11: 31648100-31648100 on Build GRCh38
rs7117653 Chr.11: 31647964-31647964 on Build GRCh38

View Full Product Details