The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033224.4 | NP_150093.1 | 1 - 1 | 3396 | 65 |
GenBank mRNA | AK056651.1 | - | 1 - 1 | 450 | 65 |