Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44260993-44261498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

SNP Detail

SNP ID Chromosome Location
rs892913904 Chr.11: 44261308-44261308 on Build GRCh38
rs554409058 Chr.11: 44261070-44261070 on Build GRCh38
rs972120441 Chr.11: 44261116-44261116 on Build GRCh38
rs147497663 Chr.11: 44261062-44261062 on Build GRCh38
rs373307279 Chr.11: 44261049-44261049 on Build GRCh38
rs940765757 Chr.11: 44261114-44261114 on Build GRCh38
rs917907442 Chr.11: 44261163-44261163 on Build GRCh38
rs552414721 Chr.11: 44261078-44261078 on Build GRCh38
rs1016172724 Chr.11: 44261279-44261279 on Build GRCh38
rs1010381955 Chr.11: 44261309-44261309 on Build GRCh38
rs1045684178 Chr.11: 44261250-44261250 on Build GRCh38
rs748214374 Chr.11: 44261472-44261472 on Build GRCh38
rs561534323 Chr.11: 44261333-44261333 on Build GRCh38
rs886048284 Chr.11: 44261094-44261099 on Build GRCh38
rs142004753 Chr.11: 44261469-44261469 on Build GRCh38
rs1030060422 Chr.11: 44261328-44261328 on Build GRCh38
rs758694942 Chr.11: 44261422-44261422 on Build GRCh38
rs745494340 Chr.11: 44261325-44261325 on Build GRCh38
rs143536201 Chr.11: 44261042-44261042 on Build GRCh38
rs771229047 Chr.11: 44261074-44261074 on Build GRCh38
rs756954535 Chr.11: 44261217-44261217 on Build GRCh38
rs747580754 Chr.11: 44261445-44261445 on Build GRCh38
rs1047507687 Chr.11: 44261121-44261121 on Build GRCh38
rs753339429 Chr.11: 44261131-44261131 on Build GRCh38
rs901530755 Chr.11: 44261420-44261420 on Build GRCh38
rs866761428 Chr.11: 44261279-44261279 on Build GRCh38
rs569151215 Chr.11: 44261277-44261277 on Build GRCh38
rs538499078 Chr.11: 44261029-44261029 on Build GRCh38
rs995980785 Chr.11: 44261315-44261315 on Build GRCh38
rs1031076826 Chr.11: 44261426-44261426 on Build GRCh38
rs886048285 Chr.11: 44261348-44261348 on Build GRCh38
rs953628584 Chr.11: 44261450-44261450 on Build GRCh38
rs116662493 Chr.11: 44261085-44261085 on Build GRCh38
rs11037919 Chr.11: 44261258-44261258 on Build GRCh38
rs185781739 Chr.11: 44261327-44261327 on Build GRCh38
rs778340814 Chr.11: 44261388-44261388 on Build GRCh38
rs575864402 Chr.11: 44261323-44261323 on Build GRCh38
rs558403646 Chr.11: 44261148-44261148 on Build GRCh38
rs114755169 Chr.11: 44261081-44261081 on Build GRCh38
rs113734133 Chr.11: 44261455-44261455 on Build GRCh38
rs531931217 Chr.11: 44261473-44261473 on Build GRCh38
rs1009937911 Chr.11: 44261020-44261020 on Build GRCh38
rs576862101 Chr.11: 44261191-44261191 on Build GRCh38
rs374150602 Chr.11: 44261414-44261415 on Build GRCh38
rs949344500 Chr.11: 44261234-44261234 on Build GRCh38
rs7115841 Chr.11: 44261449-44261449 on Build GRCh38
rs879062512 Chr.11: 44261291-44261291 on Build GRCh38
rs780412869 Chr.11: 44261441-44261441 on Build GRCh38
rs999168943 Chr.11: 44261350-44261350 on Build GRCh38

View Full Product Details