Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44263246-44263748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

SNP Detail

SNP ID Chromosome Location
rs986850014 Chr.11: 44263389-44263389 on Build GRCh38
rs572968250 Chr.11: 44263286-44263287 on Build GRCh38
rs543762689 Chr.11: 44263718-44263718 on Build GRCh38
rs565623792 Chr.11: 44263330-44263330 on Build GRCh38
rs929937466 Chr.11: 44263697-44263697 on Build GRCh38
rs544187857 Chr.11: 44263508-44263508 on Build GRCh38
rs895225599 Chr.11: 44263680-44263680 on Build GRCh38
rs913386142 Chr.11: 44263411-44263411 on Build GRCh38
rs11037920 Chr.11: 44263407-44263407 on Build GRCh38
rs552492615 Chr.11: 44263388-44263388 on Build GRCh38
rs546675090 Chr.11: 44263510-44263510 on Build GRCh38
rs10458913 Chr.11: 44263723-44263723 on Build GRCh38
rs886048303 Chr.11: 44263719-44263719 on Build GRCh38
rs947565798 Chr.11: 44263446-44263446 on Build GRCh38
rs937416547 Chr.11: 44263672-44263672 on Build GRCh38
rs1038775201 Chr.11: 44263402-44263402 on Build GRCh38
rs947451318 Chr.11: 44263265-44263265 on Build GRCh38
rs968689081 Chr.11: 44263383-44263383 on Build GRCh38
rs776401651 Chr.11: 44263636-44263636 on Build GRCh38
rs924126870 Chr.11: 44263387-44263387 on Build GRCh38
rs556404097 Chr.11: 44263483-44263483 on Build GRCh38
rs538156753 Chr.11: 44263457-44263457 on Build GRCh38
rs953931896 Chr.11: 44263555-44263555 on Build GRCh38
rs978333334 Chr.11: 44263548-44263548 on Build GRCh38
rs1015375565 Chr.11: 44263339-44263339 on Build GRCh38
rs959070756 Chr.11: 44263310-44263310 on Build GRCh38
rs886048300 Chr.11: 44263446-44263446 on Build GRCh38
rs965097966 Chr.11: 44263620-44263620 on Build GRCh38
rs978261428 Chr.11: 44263385-44263385 on Build GRCh38
rs955546865 Chr.11: 44263575-44263575 on Build GRCh38
rs534446074 Chr.11: 44263336-44263336 on Build GRCh38
rs1003306156 Chr.11: 44263293-44263293 on Build GRCh38
rs572446886 Chr.11: 44263517-44263517 on Build GRCh38
rs771684261 Chr.11: 44263391-44263391 on Build GRCh38
rs1026133216 Chr.11: 44263291-44263291 on Build GRCh38
rs886290871 Chr.11: 44263476-44263476 on Build GRCh38
rs897094228 Chr.11: 44263706-44263706 on Build GRCh38
rs1022578069 Chr.11: 44263377-44263377 on Build GRCh38
rs866343400 Chr.11: 44263544-44263544 on Build GRCh38
rs113781536 Chr.11: 44263509-44263509 on Build GRCh38
rs768453519 Chr.11: 44263563-44263563 on Build GRCh38
rs886048298 Chr.11: 44263384-44263384 on Build GRCh38
rs76501131 Chr.11: 44263627-44263627 on Build GRCh38
rs777781257 Chr.11: 44263269-44263269 on Build GRCh38
rs1034742024 Chr.11: 44263308-44263308 on Build GRCh38
rs1036878135 Chr.11: 44263701-44263701 on Build GRCh38
rs564284430 Chr.11: 44263635-44263635 on Build GRCh38
rs942751053 Chr.11: 44263396-44263396 on Build GRCh38
rs761727621 Chr.11: 44263648-44263648 on Build GRCh38
rs529798397 Chr.11: 44263489-44263489 on Build GRCh38
rs886048302 Chr.11: 44263697-44263697 on Build GRCh38
rs373805569 Chr.11: 44263322-44263322 on Build GRCh38
rs986923481 Chr.11: 44263579-44263579 on Build GRCh38
rs1013388072 Chr.11: 44263323-44263323 on Build GRCh38
rs927369185 Chr.11: 44263658-44263658 on Build GRCh38
rs562110335 Chr.11: 44263264-44263264 on Build GRCh38
rs964458023 Chr.11: 44263632-44263632 on Build GRCh38
rs995310919 Chr.11: 44263525-44263525 on Build GRCh38
rs886048301 Chr.11: 44263498-44263498 on Build GRCh38
rs937321240 Chr.11: 44263487-44263487 on Build GRCh38
rs111964563 Chr.11: 44263292-44263292 on Build GRCh38
rs760091330 Chr.11: 44263673-44263673 on Build GRCh38
rs967713224 Chr.11: 44263518-44263518 on Build GRCh38
rs886048299 Chr.11: 44263410-44263410 on Build GRCh38
rs961232466 Chr.11: 44263371-44263371 on Build GRCh38
rs917218614 Chr.11: 44263690-44263690 on Build GRCh38
rs930533277 Chr.11: 44263410-44263410 on Build GRCh38
rs966021272 Chr.11: 44263403-44263403 on Build GRCh38
rs576272082 Chr.11: 44263707-44263707 on Build GRCh38

View Full Product Details