Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44263646-44264136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

SNP Detail

SNP ID Chromosome Location
rs151214135 Chr.11: 44263882-44263882 on Build GRCh38
rs543762689 Chr.11: 44263718-44263718 on Build GRCh38
rs911161091 Chr.11: 44264010-44264010 on Build GRCh38
rs776365654 Chr.11: 44263851-44263851 on Build GRCh38
rs929937466 Chr.11: 44263697-44263697 on Build GRCh38
rs951437284 Chr.11: 44263998-44263998 on Build GRCh38
rs543627488 Chr.11: 44263955-44263955 on Build GRCh38
rs1043461284 Chr.11: 44263753-44263753 on Build GRCh38
rs895225599 Chr.11: 44263680-44263680 on Build GRCh38
rs1031249586 Chr.11: 44263778-44263778 on Build GRCh38
rs10458913 Chr.11: 44263723-44263723 on Build GRCh38
rs886048303 Chr.11: 44263719-44263719 on Build GRCh38
rs937416547 Chr.11: 44263672-44263672 on Build GRCh38
rs955619452 Chr.11: 44263783-44263783 on Build GRCh38
rs555740434 Chr.11: 44263981-44263981 on Build GRCh38
rs962108525 Chr.11: 44263986-44263986 on Build GRCh38
rs185632337 Chr.11: 44263932-44263932 on Build GRCh38
rs1008375147 Chr.11: 44263785-44263785 on Build GRCh38
rs1056695823 Chr.11: 44263732-44263732 on Build GRCh38
rs117526668 Chr.11: 44263881-44263881 on Build GRCh38
rs532888348 Chr.11: 44263806-44263806 on Build GRCh38
rs960007733 Chr.11: 44264028-44264028 on Build GRCh38
rs552527026 Chr.11: 44263892-44263892 on Build GRCh38
rs768051621 Chr.11: 44263872-44263872 on Build GRCh38
rs964561896 Chr.11: 44263979-44263979 on Build GRCh38
rs746147000 Chr.11: 44263987-44263987 on Build GRCh38
rs1009925999 Chr.11: 44263804-44263804 on Build GRCh38
rs1041513805 Chr.11: 44264075-44264075 on Build GRCh38
rs142207322 Chr.11: 44263829-44263829 on Build GRCh38
rs192668810 Chr.11: 44263857-44263857 on Build GRCh38
rs369798593 Chr.11: 44264016-44264016 on Build GRCh38
rs999113525 Chr.11: 44263777-44263777 on Build GRCh38
rs184957641 Chr.11: 44263864-44263864 on Build GRCh38
rs897094228 Chr.11: 44263706-44263706 on Build GRCh38
rs149094042 Chr.11: 44263766-44263766 on Build GRCh38
rs889604969 Chr.11: 44263893-44263893 on Build GRCh38
rs972041200 Chr.11: 44263884-44263884 on Build GRCh38
rs916420953 Chr.11: 44263909-44263909 on Build GRCh38
rs763305324 Chr.11: 44263941-44263941 on Build GRCh38
rs759202883 Chr.11: 44263807-44263807 on Build GRCh38
rs781008520 Chr.11: 44264098-44264098 on Build GRCh38
rs1036878135 Chr.11: 44263701-44263701 on Build GRCh38
rs190383779 Chr.11: 44263805-44263805 on Build GRCh38
rs566027488 Chr.11: 44263926-44263926 on Build GRCh38
rs939198909 Chr.11: 44263731-44263731 on Build GRCh38
rs964214818 Chr.11: 44263828-44263828 on Build GRCh38
rs1024127317 Chr.11: 44263736-44263736 on Build GRCh38
rs747955576 Chr.11: 44264116-44264116 on Build GRCh38
rs1034633563 Chr.11: 44263954-44263954 on Build GRCh38
rs765943711 Chr.11: 44263754-44263754 on Build GRCh38
rs573850141 Chr.11: 44263999-44263999 on Build GRCh38
rs140400459 Chr.11: 44263891-44263891 on Build GRCh38
rs181642223 Chr.11: 44263931-44263931 on Build GRCh38
rs886048302 Chr.11: 44263697-44263697 on Build GRCh38
rs76229477 Chr.11: 44263952-44263952 on Build GRCh38
rs146483327 Chr.11: 44263733-44263733 on Build GRCh38
rs373562733 Chr.11: 44264072-44264072 on Build GRCh38
rs760091330 Chr.11: 44263673-44263673 on Build GRCh38
rs149719812 Chr.11: 44264045-44264047 on Build GRCh38
rs1012425450 Chr.11: 44263750-44263750 on Build GRCh38
rs1008908597 Chr.11: 44263962-44263962 on Build GRCh38
rs942561945 Chr.11: 44263908-44263908 on Build GRCh38
rs1018504484 Chr.11: 44263978-44263978 on Build GRCh38
rs758619819 Chr.11: 44263995-44263995 on Build GRCh38
rs917218614 Chr.11: 44263690-44263690 on Build GRCh38
rs188954464 Chr.11: 44263910-44263910 on Build GRCh38
rs576272082 Chr.11: 44263707-44263707 on Build GRCh38

View Full Product Details