Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44264036-44264540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

SNP Detail

SNP ID Chromosome Location
rs925131509 Chr.11: 44264321-44264321 on Build GRCh38
rs544795542 Chr.11: 44264234-44264234 on Build GRCh38
rs1039366158 Chr.11: 44264423-44264423 on Build GRCh38
rs879258985 Chr.11: 44264187-44264187 on Build GRCh38
rs887217228 Chr.11: 44264290-44264290 on Build GRCh38
rs113592690 Chr.11: 44264429-44264430 on Build GRCh38
rs960323235 Chr.11: 44264293-44264293 on Build GRCh38
rs410592 Chr.11: 44264141-44264141 on Build GRCh38
rs967178204 Chr.11: 44264514-44264514 on Build GRCh38
rs775765158 Chr.11: 44264503-44264503 on Build GRCh38
rs1040368269 Chr.11: 44264188-44264188 on Build GRCh38
rs890809009 Chr.11: 44264185-44264185 on Build GRCh38
rs4755797 Chr.11: 44264337-44264337 on Build GRCh38
rs897969515 Chr.11: 44264396-44264396 on Build GRCh38
rs1041513805 Chr.11: 44264075-44264075 on Build GRCh38
rs900072756 Chr.11: 44264240-44264240 on Build GRCh38
rs1017602865 Chr.11: 44264463-44264463 on Build GRCh38
rs945893970 Chr.11: 44264295-44264295 on Build GRCh38
rs1004352060 Chr.11: 44264492-44264492 on Build GRCh38
rs454205 Chr.11: 44264140-44264140 on Build GRCh38
rs979287863 Chr.11: 44264119-44264119 on Build GRCh38
rs558317757 Chr.11: 44264457-44264457 on Build GRCh38
rs4755796 Chr.11: 44264307-44264307 on Build GRCh38
rs1052104662 Chr.11: 44264182-44264182 on Build GRCh38
rs113482355 Chr.11: 44264482-44264482 on Build GRCh38
rs781008520 Chr.11: 44264098-44264098 on Build GRCh38
rs554752826 Chr.11: 44264281-44264281 on Build GRCh38
rs993645498 Chr.11: 44264416-44264416 on Build GRCh38
rs747955576 Chr.11: 44264116-44264116 on Build GRCh38
rs1014505562 Chr.11: 44264356-44264356 on Build GRCh38
rs574340639 Chr.11: 44264291-44264291 on Build GRCh38
rs969314126 Chr.11: 44264310-44264310 on Build GRCh38
rs539799951 Chr.11: 44264447-44264447 on Build GRCh38
rs935625791 Chr.11: 44264327-44264327 on Build GRCh38
rs570155774 Chr.11: 44264507-44264507 on Build GRCh38
rs549951730 Chr.11: 44264308-44264308 on Build GRCh38
rs147563621 Chr.11: 44264195-44264195 on Build GRCh38
rs1035739473 Chr.11: 44264292-44264292 on Build GRCh38
rs962224127 Chr.11: 44264154-44264154 on Build GRCh38
rs534068592 Chr.11: 44264330-44264330 on Build GRCh38
rs1045986678 Chr.11: 44264357-44264357 on Build GRCh38
rs373562733 Chr.11: 44264072-44264072 on Build GRCh38
rs866751592 Chr.11: 44264254-44264254 on Build GRCh38
rs532906858 Chr.11: 44264199-44264199 on Build GRCh38
rs993524376 Chr.11: 44264155-44264155 on Build GRCh38
rs1008092384 Chr.11: 44264459-44264459 on Build GRCh38
rs935069701 Chr.11: 44264177-44264177 on Build GRCh38
rs542591808 Chr.11: 44264417-44264417 on Build GRCh38
rs887277360 Chr.11: 44264490-44264490 on Build GRCh38
rs900106013 Chr.11: 44264433-44264433 on Build GRCh38
rs996114763 Chr.11: 44264280-44264280 on Build GRCh38
rs568270311 Chr.11: 44264311-44264311 on Build GRCh38
rs986152359 Chr.11: 44264278-44264278 on Build GRCh38
rs912279375 Chr.11: 44264388-44264388 on Build GRCh38

View Full Product Details