Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44309915-44310420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX4
Entrez Gene ID
60529
Gene Name
ALX homeobox 4
Gene Aliases
CRS5, FND2
UniGene
Hs.436055
Gene Chromosome Location
Chr.11: 44260728-44310166 on Build GRCh38
Transcripts
NM_021926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3752366 c.104G>C p.R35T Chr.11: 44309959-44309959 on Build GRCh38
306892 c.69G>C p.P23P Chr.11: 44309994-44309994 on Build GRCh38
1746231 c.79A>T p.S27C Chr.11: 44309984-44309984 on Build GRCh38
4032841 c.2T>C p.M1T Chr.11: 44310061-44310061 on Build GRCh38
1979331 c.129C>T p.F43F Chr.11: 44309934-44309934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015731069 Chr.11: 44310393-44310393 on Build GRCh38
rs868555366 Chr.11: 44309957-44309957 on Build GRCh38
rs1009644339 Chr.11: 44310077-44310077 on Build GRCh38
rs990702443 Chr.11: 44309972-44309972 on Build GRCh38
rs377130834 Chr.11: 44310107-44310107 on Build GRCh38
rs753099218 Chr.11: 44309958-44309958 on Build GRCh38
rs373566188 Chr.11: 44309942-44309942 on Build GRCh38
rs748167884 Chr.11: 44310092-44310092 on Build GRCh38
rs746177266 Chr.11: 44310288-44310289 on Build GRCh38
rs370055325 Chr.11: 44309974-44309974 on Build GRCh38
rs567317334 Chr.11: 44310180-44310180 on Build GRCh38
rs767919976 Chr.11: 44309954-44309954 on Build GRCh38
rs374650044 Chr.11: 44310001-44310001 on Build GRCh38
rs961774593 Chr.11: 44310395-44310395 on Build GRCh38
rs746753665 Chr.11: 44309996-44309996 on Build GRCh38
rs764565331 Chr.11: 44310113-44310113 on Build GRCh38
rs1040745274 Chr.11: 44310227-44310227 on Build GRCh38
rs778531428 Chr.11: 44309988-44309988 on Build GRCh38
rs949272605 Chr.11: 44310002-44310002 on Build GRCh38
rs756783968 Chr.11: 44309981-44309981 on Build GRCh38
rs281865153 Chr.11: 44310044-44310044 on Build GRCh38
rs1006138426 Chr.11: 44310344-44310344 on Build GRCh38
rs901312887 Chr.11: 44310087-44310087 on Build GRCh38
rs1028571716 Chr.11: 44310332-44310332 on Build GRCh38
rs775495740 Chr.11: 44309995-44309995 on Build GRCh38
rs776324451 Chr.11: 44310289-44310289 on Build GRCh38
rs763035360 Chr.11: 44309944-44309944 on Build GRCh38
rs779947973 Chr.11: 44310187-44310187 on Build GRCh38
rs3824915 Chr.11: 44309959-44309959 on Build GRCh38
rs549253712 Chr.11: 44310285-44310286 on Build GRCh38
rs774417073 Chr.11: 44310094-44310094 on Build GRCh38
rs370141684 Chr.11: 44310074-44310074 on Build GRCh38
rs534829417 Chr.11: 44310204-44310204 on Build GRCh38
rs767405336 Chr.11: 44310287-44310289 on Build GRCh38
rs940008482 Chr.11: 44309999-44309999 on Build GRCh38
rs551131922 Chr.11: 44310286-44310286 on Build GRCh38
rs892212276 Chr.11: 44310213-44310213 on Build GRCh38
rs945073880 Chr.11: 44310224-44310224 on Build GRCh38
rs796988365 Chr.11: 44310287-44310287 on Build GRCh38
rs961447352 Chr.11: 44310148-44310148 on Build GRCh38
rs997130653 Chr.11: 44310331-44310331 on Build GRCh38
rs766617978 Chr.11: 44310333-44310333 on Build GRCh38
rs1041884317 Chr.11: 44310328-44310328 on Build GRCh38
rs775418839 Chr.11: 44310251-44310252 on Build GRCh38
rs551356786 Chr.11: 44310102-44310102 on Build GRCh38
rs559427156 Chr.11: 44310056-44310056 on Build GRCh38
rs373304528 Chr.11: 44310082-44310082 on Build GRCh38
rs749945782 Chr.11: 44310062-44310062 on Build GRCh38
rs748769921 Chr.11: 44309980-44309980 on Build GRCh38
rs971077214 Chr.11: 44310179-44310179 on Build GRCh38
rs980635977 Chr.11: 44310177-44310177 on Build GRCh38
rs746807473 Chr.11: 44310073-44310073 on Build GRCh38
rs533107251 Chr.11: 44310098-44310098 on Build GRCh38
rs116381450 Chr.11: 44310153-44310153 on Build GRCh38
rs984256325 Chr.11: 44310143-44310143 on Build GRCh38
rs1044890514 Chr.11: 44310027-44310027 on Build GRCh38
rs774394460 Chr.11: 44310024-44310024 on Build GRCh38
rs936055169 Chr.11: 44310338-44310338 on Build GRCh38
rs540061419 Chr.11: 44310214-44310215 on Build GRCh38
rs976437741 Chr.11: 44310279-44310279 on Build GRCh38
rs963724598 Chr.11: 44310213-44310214 on Build GRCh38
rs766486576 Chr.11: 44310019-44310019 on Build GRCh38
rs946324140 Chr.11: 44310293-44310293 on Build GRCh38
rs754405082 Chr.11: 44310047-44310047 on Build GRCh38
rs1041331414 Chr.11: 44310055-44310055 on Build GRCh38
rs762975194 Chr.11: 44310011-44310011 on Build GRCh38
rs949324499 Chr.11: 44310163-44310163 on Build GRCh38
rs758130420 Chr.11: 44310066-44310066 on Build GRCh38
rs71035686 Chr.11: 44310295-44310295 on Build GRCh38
rs530695931 Chr.11: 44310122-44310122 on Build GRCh38
rs746479735 Chr.11: 44310046-44310047 on Build GRCh38
rs755162732 Chr.11: 44309951-44309951 on Build GRCh38
rs992798837 Chr.11: 44310151-44310151 on Build GRCh38
rs922864150 Chr.11: 44310291-44310291 on Build GRCh38
rs764539399 Chr.11: 44310045-44310045 on Build GRCh38
rs984594878 Chr.11: 44309991-44309991 on Build GRCh38
rs936491740 Chr.11: 44310200-44310200 on Build GRCh38
rs61737298 Chr.11: 44310000-44310000 on Build GRCh38
rs772303422 Chr.11: 44310099-44310099 on Build GRCh38
rs115968657 Chr.11: 44309994-44309994 on Build GRCh38
rs886048305 Chr.11: 44310143-44310143 on Build GRCh38
rs779702556 Chr.11: 44310072-44310072 on Build GRCh38
rs868447607 Chr.11: 44310147-44310147 on Build GRCh38
rs767658097 Chr.11: 44310033-44310033 on Build GRCh38
rs766611438 Chr.11: 44309949-44309949 on Build GRCh38
rs778621319 Chr.11: 44310058-44310058 on Build GRCh38
rs904596139 Chr.11: 44310118-44310118 on Build GRCh38
rs375641880 Chr.11: 44309964-44309964 on Build GRCh38
rs952439034 Chr.11: 44310197-44310197 on Build GRCh38
rs370875089 Chr.11: 44310293-44310295 on Build GRCh38
rs773173346 Chr.11: 44309943-44309943 on Build GRCh38
rs772754759 Chr.11: 44310045-44310046 on Build GRCh38
rs755972714 Chr.11: 44310039-44310039 on Build GRCh38
rs931085887 Chr.11: 44310008-44310008 on Build GRCh38
rs369346595 Chr.11: 44309937-44309937 on Build GRCh38
rs78306887 Chr.11: 44310287-44310288 on Build GRCh38
rs900815008 Chr.11: 44310228-44310228 on Build GRCh38
rs1053577568 Chr.11: 44310201-44310201 on Build GRCh38

View Full Product Details